Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 71696212-71696644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E3
Entrez Gene ID
317649
Gene Name
eukaryotic translation initiation factor 4E family member 3
Gene Aliases
eIF-4E3, eIF4E-3
UniGene
Hs.581355
Gene Chromosome Location
Chr.3: 71679289-71754831 on Build GRCh38
Transcripts
NM_001134649.2 NM_001134650.1 NM_001134651.1 NM_001282886.1 NM_173359.4 XM_011533651.2 XM_017006278.1 XM_017006279.1 XM_017006280.1 XM_017006281.1 XM_017006282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1149790 c.352G>T p.E118* Chr.3: 71696513-71696513 on Build GRCh38
6011084 c.33G>A p.E11E Chr.3: 71696514-71696514 on Build GRCh38
5599462 c.391C>T p.P131S Chr.3: 71696474-71696474 on Build GRCh38
6011083 c.351G>A p.E117E Chr.3: 71696514-71696514 on Build GRCh38
731639 c.34G>T p.E12* Chr.3: 71696513-71696513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770648457 Chr.3: 71696492-71696492 on Build GRCh38
rs750907767 Chr.3: 71696470-71696470 on Build GRCh38
rs775027686 Chr.3: 71696511-71696511 on Build GRCh38
rs968637028 Chr.3: 71696472-71696472 on Build GRCh38
rs546519899 Chr.3: 71696598-71696598 on Build GRCh38
rs920465078 Chr.3: 71696435-71696435 on Build GRCh38
rs9876950 Chr.3: 71696249-71696249 on Build GRCh38
rs766963638 Chr.3: 71696558-71696558 on Build GRCh38
rs142557730 Chr.3: 71696493-71696493 on Build GRCh38
rs756639426 Chr.3: 71696473-71696473 on Build GRCh38
rs897480708 Chr.3: 71696399-71696399 on Build GRCh38
rs750733610 Chr.3: 71696389-71696389 on Build GRCh38
rs762417927 Chr.3: 71696513-71696513 on Build GRCh38
rs1052462219 Chr.3: 71696528-71696528 on Build GRCh38
rs904268191 Chr.3: 71696340-71696340 on Build GRCh38
rs757571848 Chr.3: 71696555-71696555 on Build GRCh38
rs373339454 Chr.3: 71696582-71696582 on Build GRCh38
rs942336466 Chr.3: 71696617-71696617 on Build GRCh38
rs552037155 Chr.3: 71696507-71696507 on Build GRCh38
rs751958629 Chr.3: 71696461-71696461 on Build GRCh38
rs781517526 Chr.3: 71696415-71696415 on Build GRCh38
rs780340330 Chr.3: 71696475-71696475 on Build GRCh38
rs568500933 Chr.3: 71696599-71696599 on Build GRCh38
rs376929795 Chr.3: 71696424-71696424 on Build GRCh38
rs759464935 Chr.3: 71696443-71696443 on Build GRCh38
rs1052939278 Chr.3: 71696240-71696240 on Build GRCh38
rs774014261 Chr.3: 71696524-71696524 on Build GRCh38
rs78161596 Chr.3: 71696426-71696426 on Build GRCh38
rs559389462 Chr.3: 71696397-71696397 on Build GRCh38
rs749358048 Chr.3: 71696480-71696480 on Build GRCh38
rs1037394914 Chr.3: 71696370-71696370 on Build GRCh38
rs906562226 Chr.3: 71696237-71696237 on Build GRCh38
rs752486443 Chr.3: 71696271-71696271 on Build GRCh38
rs770722374 Chr.3: 71696434-71696434 on Build GRCh38
rs777344087 Chr.3: 71696529-71696529 on Build GRCh38
rs1028749183 Chr.3: 71696381-71696381 on Build GRCh38
rs939302277 Chr.3: 71696517-71696517 on Build GRCh38
rs148884167 Chr.3: 71696343-71696343 on Build GRCh38
rs755292451 Chr.3: 71696481-71696481 on Build GRCh38
rs200481951 Chr.3: 71696430-71696430 on Build GRCh38
rs779312149 Chr.3: 71696483-71696483 on Build GRCh38
rs769755377 Chr.3: 71696509-71696509 on Build GRCh38
rs575962715 Chr.3: 71696241-71696241 on Build GRCh38
rs74589814 Chr.3: 71696258-71696258 on Build GRCh38
rs764653823 Chr.3: 71696446-71696446 on Build GRCh38
rs369768758 Chr.3: 71696442-71696442 on Build GRCh38
rs754359365 Chr.3: 71696565-71696565 on Build GRCh38
rs889342408 Chr.3: 71696286-71696286 on Build GRCh38
rs892734037 Chr.3: 71696281-71696281 on Build GRCh38
rs1050682864 Chr.3: 71696274-71696274 on Build GRCh38
rs9877458 Chr.3: 71696537-71696537 on Build GRCh38
rs762648303 Chr.3: 71696449-71696449 on Build GRCh38
rs760517928 Chr.3: 71696433-71696433 on Build GRCh38
rs528287934 Chr.3: 71696576-71696576 on Build GRCh38
rs757593083 Chr.3: 71696465-71696465 on Build GRCh38
rs748334973 Chr.3: 71696484-71696484 on Build GRCh38
rs1043890923 Chr.3: 71696573-71696573 on Build GRCh38
rs910704979 Chr.3: 71696615-71696615 on Build GRCh38
rs540516657 Chr.3: 71696379-71696379 on Build GRCh38
rs750787005 Chr.3: 71696538-71696538 on Build GRCh38
rs752195959 Chr.3: 71696448-71696448 on Build GRCh38
rs535956262 Chr.3: 71696600-71696600 on Build GRCh38
rs766369312 Chr.3: 71696275-71696275 on Build GRCh38
rs146976107 Chr.3: 71696427-71696427 on Build GRCh38
rs763554441 Chr.3: 71696522-71696522 on Build GRCh38
rs768988956 Chr.3: 71696429-71696429 on Build GRCh38
rs530682446 Chr.3: 71696460-71696460 on Build GRCh38
rs1006871987 Chr.3: 71696351-71696351 on Build GRCh38

View Full Product Details