Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 48576063-48576567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A5
Entrez Gene ID
25817
Gene Name
family with sequence similarity 19 member A5, C-C motif chemokine like
Gene Aliases
QLLK5208, TAFA-5, TAFA5, UNQ5208
UniGene
Hs.436854
Gene Chromosome Location
Chr.22: 48489460-48751935 on Build GRCh38
Transcripts
NM_001082967.2 NM_015381.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7201656 c.61C>T p.L21L Chr.22: 48576541-48576541 on Build GRCh38
3064021 c.40G>A p.A14T Chr.22: 48576520-48576520 on Build GRCh38
6559481 c.23G>T p.W8L Chr.22: 48576503-48576503 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753515423 Chr.22: 48576479-48576479 on Build GRCh38
rs573207972 Chr.22: 48576457-48576457 on Build GRCh38
rs750529065 Chr.22: 48576444-48576444 on Build GRCh38
rs778675506 Chr.22: 48576496-48576496 on Build GRCh38
rs768127953 Chr.22: 48576518-48576518 on Build GRCh38
rs923877374 Chr.22: 48576292-48576292 on Build GRCh38
rs375758320 Chr.22: 48576482-48576482 on Build GRCh38
rs933980930 Chr.22: 48576344-48576344 on Build GRCh38
rs111704292 Chr.22: 48576149-48576149 on Build GRCh38
rs202051146 Chr.22: 48576509-48576509 on Build GRCh38
rs550750077 Chr.22: 48576355-48576355 on Build GRCh38
rs773751241 Chr.22: 48576541-48576541 on Build GRCh38
rs1014041440 Chr.22: 48576297-48576297 on Build GRCh38
rs967358955 Chr.22: 48576281-48576281 on Build GRCh38
rs994716407 Chr.22: 48576292-48576293 on Build GRCh38
rs967935570 Chr.22: 48576131-48576131 on Build GRCh38
rs546610655 Chr.22: 48576166-48576166 on Build GRCh38
rs757241535 Chr.22: 48576486-48576486 on Build GRCh38
rs921225966 Chr.22: 48576206-48576206 on Build GRCh38
rs892877649 Chr.22: 48576537-48576537 on Build GRCh38
rs1055706726 Chr.22: 48576428-48576428 on Build GRCh38
rs898973572 Chr.22: 48576423-48576423 on Build GRCh38
rs890324082 Chr.22: 48576245-48576245 on Build GRCh38
rs191207490 Chr.22: 48576305-48576305 on Build GRCh38
rs936035540 Chr.22: 48576158-48576158 on Build GRCh38
rs908619591 Chr.22: 48576364-48576364 on Build GRCh38
rs748416322 Chr.22: 48576535-48576535 on Build GRCh38
rs370029176 Chr.22: 48576520-48576520 on Build GRCh38
rs375149926 Chr.22: 48576543-48576543 on Build GRCh38
rs964510767 Chr.22: 48576249-48576249 on Build GRCh38
rs755185178 Chr.22: 48576455-48576455 on Build GRCh38
rs894117118 Chr.22: 48576296-48576296 on Build GRCh38
rs957623061 Chr.22: 48576316-48576316 on Build GRCh38
rs1051589050 Chr.22: 48576239-48576240 on Build GRCh38
rs371861843 Chr.22: 48576434-48576434 on Build GRCh38
rs940501188 Chr.22: 48576257-48576257 on Build GRCh38
rs921279830 Chr.22: 48576336-48576336 on Build GRCh38
rs1054556391 Chr.22: 48576442-48576442 on Build GRCh38
rs139257794 Chr.22: 48576392-48576392 on Build GRCh38
rs990164680 Chr.22: 48576253-48576253 on Build GRCh38
rs533906401 Chr.22: 48576508-48576508 on Build GRCh38
rs751834078 Chr.22: 48576453-48576453 on Build GRCh38
rs975615252 Chr.22: 48576325-48576325 on Build GRCh38
rs1018347200 Chr.22: 48576243-48576243 on Build GRCh38
rs930559471 Chr.22: 48576425-48576425 on Build GRCh38
rs977949306 Chr.22: 48576285-48576285 on Build GRCh38
rs766418738 Chr.22: 48576452-48576452 on Build GRCh38
rs1036321879 Chr.22: 48576268-48576268 on Build GRCh38
rs926193984 Chr.22: 48576157-48576158 on Build GRCh38
rs986927412 Chr.22: 48576354-48576355 on Build GRCh38
rs186743544 Chr.22: 48576298-48576298 on Build GRCh38
rs1024489529 Chr.22: 48576112-48576112 on Build GRCh38
rs184126678 Chr.22: 48576540-48576540 on Build GRCh38
rs766511060 Chr.22: 48576542-48576542 on Build GRCh38
rs914520013 Chr.22: 48576274-48576274 on Build GRCh38
rs200506227 Chr.22: 48576153-48576153 on Build GRCh38
rs558062009 Chr.22: 48576445-48576445 on Build GRCh38
rs899070758 Chr.22: 48576272-48576272 on Build GRCh38
rs774593296 Chr.22: 48576465-48576465 on Build GRCh38
rs940569323 Chr.22: 48576385-48576385 on Build GRCh38
rs367548223 Chr.22: 48576433-48576433 on Build GRCh38
rs992554210 Chr.22: 48576110-48576110 on Build GRCh38
rs934098630 Chr.22: 48576215-48576215 on Build GRCh38
rs758029970 Chr.22: 48576506-48576506 on Build GRCh38
rs1034033698 Chr.22: 48576293-48576293 on Build GRCh38
rs762387102 Chr.22: 48576443-48576443 on Build GRCh38
rs745768555 Chr.22: 48576497-48576497 on Build GRCh38
rs958445116 Chr.22: 48576083-48576083 on Build GRCh38
rs977984015 Chr.22: 48576300-48576300 on Build GRCh38

View Full Product Details