Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18039216-18039737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GID4
Entrez Gene ID
79018
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C17orf39, VID24
UniGene
Hs.187422
Gene Chromosome Location
Chr.17: 18039297-18068405 on Build GRCh38
Transcripts
NM_024052.4 XM_005256799.4 XM_005256800.1 XM_011524008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3514837 c.272C>T p.P91L Chr.17: 18039736-18039736 on Build GRCh38
5385803 c.74_75CC>TT p.S25F Chr.17: 18039538-18039539 on Build GRCh38
6411849 c.31A>G p.T11A Chr.17: 18039495-18039495 on Build GRCh38
3983293 c.216A>C p.P72P Chr.17: 18039680-18039680 on Build GRCh38
4129638 c.46G>A p.G16R Chr.17: 18039510-18039510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895420718 Chr.17: 18039581-18039581 on Build GRCh38
rs1040392769 Chr.17: 18039279-18039279 on Build GRCh38
rs747617947 Chr.17: 18039407-18039408 on Build GRCh38
rs909144033 Chr.17: 18039577-18039577 on Build GRCh38
rs979620100 Chr.17: 18039294-18039294 on Build GRCh38
rs775970305 Chr.17: 18039688-18039688 on Build GRCh38
rs1001096676 Chr.17: 18039479-18039479 on Build GRCh38
rs377757723 Chr.17: 18039426-18039426 on Build GRCh38
rs564429104 Chr.17: 18039664-18039664 on Build GRCh38
rs147791971 Chr.17: 18039298-18039298 on Build GRCh38
rs545188753 Chr.17: 18039656-18039656 on Build GRCh38
rs566501754 Chr.17: 18039701-18039701 on Build GRCh38
rs918009122 Chr.17: 18039305-18039305 on Build GRCh38
rs1049247858 Chr.17: 18039601-18039601 on Build GRCh38
rs949474472 Chr.17: 18039337-18039337 on Build GRCh38
rs939375480 Chr.17: 18039329-18039329 on Build GRCh38
rs901476160 Chr.17: 18039687-18039687 on Build GRCh38
rs988567617 Chr.17: 18039528-18039528 on Build GRCh38
rs865971612 Chr.17: 18039418-18039418 on Build GRCh38
rs748773460 Chr.17: 18039684-18039684 on Build GRCh38
rs747919270 Chr.17: 18039669-18039669 on Build GRCh38
rs956950340 Chr.17: 18039492-18039492 on Build GRCh38
rs186674984 Chr.17: 18039516-18039516 on Build GRCh38
rs1049002626 Chr.17: 18039369-18039369 on Build GRCh38
rs769053973 Chr.17: 18039697-18039697 on Build GRCh38
rs998105190 Chr.17: 18039266-18039266 on Build GRCh38
rs942901007 Chr.17: 18039578-18039578 on Build GRCh38
rs759244626 Chr.17: 18039406-18039407 on Build GRCh38
rs971226127 Chr.17: 18039556-18039556 on Build GRCh38
rs531574702 Chr.17: 18039610-18039610 on Build GRCh38
rs887463459 Chr.17: 18039370-18039370 on Build GRCh38
rs966263930 Chr.17: 18039246-18039246 on Build GRCh38
rs1045979514 Chr.17: 18039341-18039341 on Build GRCh38
rs765575003 Chr.17: 18039455-18039455 on Build GRCh38
rs770486184 Chr.17: 18039686-18039686 on Build GRCh38
rs889280811 Chr.17: 18039475-18039475 on Build GRCh38
rs894559468 Chr.17: 18039658-18039658 on Build GRCh38
rs906349882 Chr.17: 18039710-18039710 on Build GRCh38
rs988273537 Chr.17: 18039303-18039303 on Build GRCh38
rs933099340 Chr.17: 18039703-18039703 on Build GRCh38
rs1023996048 Chr.17: 18039409-18039418 on Build GRCh38
rs922879032 Chr.17: 18039602-18039602 on Build GRCh38
rs143459469 Chr.17: 18039405-18039406 on Build GRCh38
rs533427067 Chr.17: 18039678-18039678 on Build GRCh38
rs762399154 Chr.17: 18039656-18039657 on Build GRCh38
rs927080295 Chr.17: 18039344-18039344 on Build GRCh38
rs535142636 Chr.17: 18039670-18039670 on Build GRCh38
rs770105827 Chr.17: 18039719-18039719 on Build GRCh38
rs887078264 Chr.17: 18039363-18039363 on Build GRCh38
rs780983555 Chr.17: 18039665-18039665 on Build GRCh38
rs577619186 Chr.17: 18039425-18039425 on Build GRCh38
rs953631943 Chr.17: 18039277-18039277 on Build GRCh38
rs7406982 Chr.17: 18039299-18039299 on Build GRCh38
rs369896915 Chr.17: 18039552-18039552 on Build GRCh38
rs1005660043 Chr.17: 18039368-18039368 on Build GRCh38
rs940335977 Chr.17: 18039374-18039374 on Build GRCh38
rs762112011 Chr.17: 18039706-18039706 on Build GRCh38
rs759157782 Chr.17: 18039454-18039455 on Build GRCh38
rs1047321605 Chr.17: 18039620-18039620 on Build GRCh38
rs751616570 Chr.17: 18039582-18039582 on Build GRCh38
rs899003669 Chr.17: 18039398-18039398 on Build GRCh38
rs980842230 Chr.17: 18039566-18039566 on Build GRCh38
rs747481250 Chr.17: 18039694-18039694 on Build GRCh38
rs927153372 Chr.17: 18039567-18039567 on Build GRCh38
rs901607230 Chr.17: 18039408-18039408 on Build GRCh38
rs997234849 Chr.17: 18039412-18039412 on Build GRCh38
rs1038896729 Chr.17: 18039654-18039654 on Build GRCh38
rs941340088 Chr.17: 18039635-18039635 on Build GRCh38
rs1029092478 Chr.17: 18039273-18039273 on Build GRCh38
rs1005076763 Chr.17: 18039693-18039693 on Build GRCh38
rs562752755 Chr.17: 18039605-18039605 on Build GRCh38
rs1035998429 Chr.17: 18039405-18039405 on Build GRCh38
rs956334028 Chr.17: 18039467-18039467 on Build GRCh38
rs557614810 Chr.17: 18039349-18039349 on Build GRCh38
rs937141107 Chr.17: 18039568-18039568 on Build GRCh38
rs1030640064 Chr.17: 18039428-18039428 on Build GRCh38
rs1056611486 Chr.17: 18039407-18039407 on Build GRCh38
rs984736907 Chr.17: 18039574-18039574 on Build GRCh38
rs970604465 Chr.17: 18039421-18039428 on Build GRCh38
rs908548965 Chr.17: 18039623-18039623 on Build GRCh38
rs748829625 Chr.17: 18039271-18039271 on Build GRCh38
rs546695787 Chr.17: 18039681-18039681 on Build GRCh38
rs553766703 Chr.17: 18039495-18039495 on Build GRCh38
rs555563723 Chr.17: 18039238-18039238 on Build GRCh38
rs1032543574 Chr.17: 18039486-18039486 on Build GRCh38
rs1012350685 Chr.17: 18039591-18039591 on Build GRCh38

View Full Product Details