Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57641503-57642075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG1
Entrez Gene ID
9289
Gene Name
adhesion G protein-coupled receptor G1
Gene Aliases
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
UniGene
Hs.513633
Gene Chromosome Location
Chr.16: 57619535-57665039 on Build GRCh38
Transcripts
NM_001145770.2 NM_001145771.2 NM_001145772.2 NM_001145773.2 NM_001145774.2 NM_001290142.1 NM_001290143.1 NM_001290144.1 NM_005682.6 NM_201524.3 NM_201525.3 XM_005256237.2 XM_005256238.2 XM_005256239.2 XM_005256240.4 XM_005256242.4 XM_005256244.3 XM_005256245.2 XM_005256246.2 XM_005256247.2 XM_005256248.2 XM_005256249.3 XM_005256251.4 XM_005256254.2 XM_005256255.2 XM_006721338.2 XM_006721339.3 XM_006721340.3 XM_006721341.2 XM_006721342.2 XM_006721343.3 XM_006721344.2 XM_006721345.3 XM_006721347.2 XM_011523461.2 XM_011523463.2 XM_011523464.2 XM_011523465.2 XM_017023892.1

SNP Detail

SNP ID Chromosome Location
rs7184374 Chr.16: 57641841-57641841 on Build GRCh38
rs573010528 Chr.16: 57641663-57641663 on Build GRCh38
rs768957806 Chr.16: 57641844-57641844 on Build GRCh38
rs923223137 Chr.16: 57641835-57641835 on Build GRCh38
rs10468288 Chr.16: 57641526-57641526 on Build GRCh38
rs1021798620 Chr.16: 57642044-57642044 on Build GRCh38
rs776074596 Chr.16: 57641954-57641954 on Build GRCh38
rs780139864 Chr.16: 57641664-57641664 on Build GRCh38
rs149722665 Chr.16: 57641820-57641820 on Build GRCh38
rs1031309751 Chr.16: 57642017-57642017 on Build GRCh38
rs573743798 Chr.16: 57641594-57641594 on Build GRCh38
rs771148084 Chr.16: 57641724-57641724 on Build GRCh38
rs533821881 Chr.16: 57641728-57641728 on Build GRCh38
rs543955937 Chr.16: 57641948-57641948 on Build GRCh38
rs140329167 Chr.16: 57642014-57642014 on Build GRCh38
rs772600916 Chr.16: 57641952-57641952 on Build GRCh38
rs151010444 Chr.16: 57641658-57641658 on Build GRCh38
rs1016461822 Chr.16: 57641643-57641643 on Build GRCh38
rs886563689 Chr.16: 57641629-57641629 on Build GRCh38
rs1011283416 Chr.16: 57642037-57642037 on Build GRCh38
rs886510310 Chr.16: 57641851-57641851 on Build GRCh38
rs1047745903 Chr.16: 57641845-57641845 on Build GRCh38
rs981146299 Chr.16: 57641680-57641680 on Build GRCh38
rs35725310 Chr.16: 57641565-57641566 on Build GRCh38
rs587780952 Chr.16: 57641607-57641607 on Build GRCh38
rs184329440 Chr.16: 57641944-57641944 on Build GRCh38
rs990320907 Chr.16: 57641744-57641744 on Build GRCh38
rs551156836 Chr.16: 57641970-57641970 on Build GRCh38
rs555536742 Chr.16: 57641582-57641582 on Build GRCh38
rs554593338 Chr.16: 57641732-57641733 on Build GRCh38
rs1012465975 Chr.16: 57641659-57641659 on Build GRCh38
rs1038194683 Chr.16: 57641855-57641855 on Build GRCh38
rs573708671 Chr.16: 57641945-57641945 on Build GRCh38
rs544417966 Chr.16: 57641642-57641642 on Build GRCh38
rs561858971 Chr.16: 57641854-57641854 on Build GRCh38
rs747501086 Chr.16: 57641712-57641712 on Build GRCh38
rs1048268079 Chr.16: 57641625-57641625 on Build GRCh38
rs545017173 Chr.16: 57641690-57641690 on Build GRCh38
rs901810546 Chr.16: 57641542-57641542 on Build GRCh38
rs974595686 Chr.16: 57641829-57641829 on Build GRCh38
rs914385543 Chr.16: 57641784-57641784 on Build GRCh38
rs140860620 Chr.16: 57641803-57641803 on Build GRCh38
rs895282977 Chr.16: 57641887-57641887 on Build GRCh38
rs1046680970 Chr.16: 57641967-57641967 on Build GRCh38
rs186745179 Chr.16: 57642022-57642022 on Build GRCh38

View Full Product Details