Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 153999-154537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBZ
Entrez Gene ID
3050
Gene Name
hemoglobin subunit zeta
Gene Aliases
HBZ-T1, HBZ1
UniGene
Hs.585357
Gene Chromosome Location
Chr.16: 142422-154511 on Build GRCh38
Transcripts
NM_005332.2 XM_005255287.3 XM_005255288.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4489170 c.343C>T p.P115S Chr.16: 154314-154314 on Build GRCh38
1609016 c.424C>T p.R142C Chr.16: 154395-154395 on Build GRCh38
6430788 c.305T>A p.L102Q Chr.16: 154276-154276 on Build GRCh38
6400980 c.323T>C p.V108A Chr.16: 154294-154294 on Build GRCh38
4804137 c.373G>T p.A125S Chr.16: 154344-154344 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755680922 Chr.16: 154388-154389 on Build GRCh38
rs537524062 Chr.16: 154131-154131 on Build GRCh38
rs750836028 Chr.16: 154126-154126 on Build GRCh38
rs777046880 Chr.16: 154482-154482 on Build GRCh38
rs769897159 Chr.16: 154264-154264 on Build GRCh38
rs768742258 Chr.16: 154286-154286 on Build GRCh38
rs780341332 Chr.16: 154280-154280 on Build GRCh38
rs192538076 Chr.16: 154184-154184 on Build GRCh38
rs746262034 Chr.16: 154501-154501 on Build GRCh38
rs772700418 Chr.16: 154304-154304 on Build GRCh38
rs752296813 Chr.16: 154419-154419 on Build GRCh38
rs554661254 Chr.16: 154116-154116 on Build GRCh38
rs1055283723 Chr.16: 154395-154395 on Build GRCh38
rs1001995715 Chr.16: 154386-154386 on Build GRCh38
rs752703713 Chr.16: 154148-154148 on Build GRCh38
rs894233807 Chr.16: 154514-154514 on Build GRCh38
rs762273268 Chr.16: 154298-154298 on Build GRCh38
rs111819749 Chr.16: 154217-154217 on Build GRCh38
rs764152551 Chr.16: 154442-154442 on Build GRCh38
rs765392465 Chr.16: 154101-154131 on Build GRCh38
rs751257941 Chr.16: 154448-154448 on Build GRCh38
rs753340727 Chr.16: 154134-154134 on Build GRCh38
rs531302321 Chr.16: 154127-154127 on Build GRCh38
rs751894883 Chr.16: 154139-154184 on Build GRCh38
rs999035037 Chr.16: 154468-154468 on Build GRCh38
rs765879765 Chr.16: 154314-154314 on Build GRCh38
rs929143630 Chr.16: 154032-154032 on Build GRCh38
rs986271698 Chr.16: 154507-154507 on Build GRCh38
rs967623074 Chr.16: 154444-154444 on Build GRCh38
rs893477807 Chr.16: 154403-154403 on Build GRCh38
rs746892224 Chr.16: 154259-154259 on Build GRCh38
rs758265794 Chr.16: 154434-154434 on Build GRCh38
rs866807448 Chr.16: 154162-154162 on Build GRCh38
rs530419464 Chr.16: 154351-154351 on Build GRCh38
rs778954767 Chr.16: 154140-154140 on Build GRCh38
rs767747643 Chr.16: 154121-154121 on Build GRCh38
rs184262115 Chr.16: 154218-154218 on Build GRCh38
rs769097751 Chr.16: 154110-154110 on Build GRCh38
rs774459822 Chr.16: 154297-154297 on Build GRCh38
rs750738743 Chr.16: 154100-154101 on Build GRCh38
rs906353426 Chr.16: 154341-154341 on Build GRCh38
rs199870026 Chr.16: 154157-154157 on Build GRCh38
rs1047332154 Chr.16: 154447-154447 on Build GRCh38
rs558812274 Chr.16: 154310-154310 on Build GRCh38
rs111716204 Chr.16: 154411-154411 on Build GRCh38
rs71382264 Chr.16: 154248-154248 on Build GRCh38
rs766812663 Chr.16: 154123-154127 on Build GRCh38
rs759067295 Chr.16: 154363-154363 on Build GRCh38
rs941076600 Chr.16: 154473-154473 on Build GRCh38
rs753148548 Chr.16: 154146-154175 on Build GRCh38
rs781669341 Chr.16: 154144-154147 on Build GRCh38
rs755333814 Chr.16: 154144-154174 on Build GRCh38
rs749303647 Chr.16: 154285-154285 on Build GRCh38
rs868845913 Chr.16: 154180-154180 on Build GRCh38
rs773433397 Chr.16: 154308-154308 on Build GRCh38
rs763081773 Chr.16: 154105-154105 on Build GRCh38
rs868564060 Chr.16: 154152-154152 on Build GRCh38
rs754539140 Chr.16: 154136-154136 on Build GRCh38
rs375118948 Chr.16: 154441-154441 on Build GRCh38
rs763050711 Chr.16: 154118-154127 on Build GRCh38

View Full Product Details