Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81656802-81657314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPA1
Entrez Gene ID
3612
Gene Name
inositol monophosphatase 1
Gene Aliases
IMP, IMPA
UniGene
Hs.656694
Gene Chromosome Location
Chr.8: 81656916-81686354 on Build GRCh38
Transcripts
NM_001144878.1 NM_001144879.1 NM_005536.3

SNP Detail

SNP ID Chromosome Location
rs892600685 Chr.8: 81657048-81657048 on Build GRCh38
rs982724606 Chr.8: 81657189-81657189 on Build GRCh38
rs563165987 Chr.8: 81656895-81656895 on Build GRCh38
rs1056849180 Chr.8: 81656907-81656907 on Build GRCh38
rs1046628009 Chr.8: 81656981-81656984 on Build GRCh38
rs1039451838 Chr.8: 81657255-81657256 on Build GRCh38
rs763172764 Chr.8: 81657017-81657017 on Build GRCh38
rs141693012 Chr.8: 81657143-81657143 on Build GRCh38
rs926992851 Chr.8: 81656846-81656846 on Build GRCh38
rs62512280 Chr.8: 81656834-81656834 on Build GRCh38
rs528905985 Chr.8: 81656912-81656912 on Build GRCh38
rs34007079 Chr.8: 81657186-81657186 on Build GRCh38
rs374158539 Chr.8: 81656977-81656977 on Build GRCh38
rs976682349 Chr.8: 81656910-81656910 on Build GRCh38
rs551599101 Chr.8: 81657063-81657063 on Build GRCh38
rs1010169280 Chr.8: 81657051-81657051 on Build GRCh38
rs770381514 Chr.8: 81656954-81656954 on Build GRCh38
rs746395387 Chr.8: 81656925-81656925 on Build GRCh38
rs897442876 Chr.8: 81656992-81656992 on Build GRCh38
rs949579332 Chr.8: 81657180-81657180 on Build GRCh38
rs929801496 Chr.8: 81657105-81657105 on Build GRCh38
rs571590619 Chr.8: 81657101-81657101 on Build GRCh38
rs1018082256 Chr.8: 81656890-81656890 on Build GRCh38
rs978649204 Chr.8: 81657132-81657132 on Build GRCh38
rs768608304 Chr.8: 81657093-81657093 on Build GRCh38
rs537370023 Chr.8: 81657108-81657108 on Build GRCh38
rs192893996 Chr.8: 81656975-81656975 on Build GRCh38
rs145959111 Chr.8: 81657288-81657288 on Build GRCh38
rs78050442 Chr.8: 81656843-81656843 on Build GRCh38
rs1033902128 Chr.8: 81657254-81657254 on Build GRCh38
rs892559558 Chr.8: 81657195-81657195 on Build GRCh38
rs774705061 Chr.8: 81657100-81657100 on Build GRCh38
rs147975053 Chr.8: 81656913-81656913 on Build GRCh38
rs981073178 Chr.8: 81657086-81657086 on Build GRCh38
rs936717298 Chr.8: 81656931-81656931 on Build GRCh38
rs1020192870 Chr.8: 81657099-81657099 on Build GRCh38
rs916677422 Chr.8: 81657178-81657178 on Build GRCh38
rs933986246 Chr.8: 81657176-81657176 on Build GRCh38
rs1051159174 Chr.8: 81657191-81657191 on Build GRCh38
rs904060576 Chr.8: 81657242-81657242 on Build GRCh38
rs909745619 Chr.8: 81657136-81657136 on Build GRCh38
rs1009684589 Chr.8: 81657240-81657240 on Build GRCh38
rs1033974141 Chr.8: 81657087-81657087 on Build GRCh38
rs1002403390 Chr.8: 81657244-81657244 on Build GRCh38
rs998266657 Chr.8: 81656997-81656997 on Build GRCh38
rs1039680115 Chr.8: 81657158-81657158 on Build GRCh38
rs878926936 Chr.8: 81657043-81657043 on Build GRCh38
rs968694046 Chr.8: 81657071-81657071 on Build GRCh38
rs983034027 Chr.8: 81657112-81657112 on Build GRCh38
rs916727642 Chr.8: 81656968-81656968 on Build GRCh38
rs888116828 Chr.8: 81657035-81657035 on Build GRCh38
rs899855103 Chr.8: 81656987-81656987 on Build GRCh38
rs958399263 Chr.8: 81657089-81657089 on Build GRCh38
rs917073373 Chr.8: 81656911-81656911 on Build GRCh38
rs778214420 Chr.8: 81656883-81656884 on Build GRCh38
rs77290168 Chr.8: 81657241-81657241 on Build GRCh38
rs958327557 Chr.8: 81657255-81657255 on Build GRCh38
rs565440148 Chr.8: 81656952-81656952 on Build GRCh38
rs899824237 Chr.8: 81657159-81657159 on Build GRCh38
rs930173146 Chr.8: 81657221-81657221 on Build GRCh38
rs937073601 Chr.8: 81656870-81656870 on Build GRCh38
rs929620395 Chr.8: 81657005-81657005 on Build GRCh38
rs1005522708 Chr.8: 81656973-81656973 on Build GRCh38

View Full Product Details