Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62236904-62237404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK4
Entrez Gene ID
163782
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
ANKRD38, dJ1078M7.1
UniGene
Hs.283398
Gene Chromosome Location
Chr.1: 62236165-62319414 on Build GRCh38
Transcripts
NM_001320269.1 NM_181712.4 XM_011540843.2 XM_011540845.2 XM_017000483.1 XM_017000484.1 XM_017000485.1

SNP Detail

SNP ID Chromosome Location
rs574905626 Chr.1: 62237084-62237084 on Build GRCh38
rs796810386 Chr.1: 62237147-62237148 on Build GRCh38
rs953441032 Chr.1: 62237061-62237061 on Build GRCh38
rs953565395 Chr.1: 62237122-62237122 on Build GRCh38
rs189616069 Chr.1: 62237296-62237296 on Build GRCh38
rs996594563 Chr.1: 62237365-62237365 on Build GRCh38
rs1054142052 Chr.1: 62237348-62237348 on Build GRCh38
rs919151832 Chr.1: 62237107-62237107 on Build GRCh38
rs930453389 Chr.1: 62237026-62237026 on Build GRCh38
rs573401862 Chr.1: 62237132-62237132 on Build GRCh38
rs930714979 Chr.1: 62237258-62237258 on Build GRCh38
rs1000969179 Chr.1: 62237243-62237243 on Build GRCh38
rs1057196454 Chr.1: 62237127-62237127 on Build GRCh38
rs6688986 Chr.1: 62237048-62237048 on Build GRCh38
rs2481688 Chr.1: 62236965-62236965 on Build GRCh38
rs768085338 Chr.1: 62237063-62237063 on Build GRCh38
rs57754916 Chr.1: 62237148-62237149 on Build GRCh38
rs112507659 Chr.1: 62237275-62237275 on Build GRCh38
rs963233698 Chr.1: 62237367-62237367 on Build GRCh38
rs370877595 Chr.1: 62237131-62237132 on Build GRCh38
rs894343253 Chr.1: 62237046-62237046 on Build GRCh38
rs747409672 Chr.1: 62237263-62237263 on Build GRCh38
rs765353223 Chr.1: 62237366-62237366 on Build GRCh38
rs569560714 Chr.1: 62237162-62237162 on Build GRCh38
rs953325460 Chr.1: 62237352-62237352 on Build GRCh38
rs776928555 Chr.1: 62237297-62237297 on Build GRCh38
rs767479887 Chr.1: 62237131-62237131 on Build GRCh38
rs998903694 Chr.1: 62237347-62237347 on Build GRCh38
rs1042758648 Chr.1: 62237193-62237193 on Build GRCh38
rs772554857 Chr.1: 62236948-62236948 on Build GRCh38
rs576030469 Chr.1: 62237016-62237016 on Build GRCh38
rs1045691081 Chr.1: 62237052-62237052 on Build GRCh38
rs938393276 Chr.1: 62237123-62237123 on Build GRCh38
rs904048357 Chr.1: 62237199-62237199 on Build GRCh38
rs956753404 Chr.1: 62236998-62236998 on Build GRCh38
rs943949946 Chr.1: 62237115-62237115 on Build GRCh38
rs186421720 Chr.1: 62237082-62237082 on Build GRCh38
rs909056202 Chr.1: 62237072-62237072 on Build GRCh38
rs768903494 Chr.1: 62237131-62237134 on Build GRCh38
rs1036818440 Chr.1: 62237001-62237001 on Build GRCh38
rs972107093 Chr.1: 62237215-62237215 on Build GRCh38
rs997566660 Chr.1: 62237292-62237292 on Build GRCh38
rs181495182 Chr.1: 62237362-62237362 on Build GRCh38
rs972025547 Chr.1: 62237083-62237083 on Build GRCh38
rs987744980 Chr.1: 62236944-62236944 on Build GRCh38
rs537704641 Chr.1: 62237248-62237248 on Build GRCh38
rs987713927 Chr.1: 62237172-62237172 on Build GRCh38
rs1011051714 Chr.1: 62237094-62237094 on Build GRCh38
rs912154760 Chr.1: 62237183-62237183 on Build GRCh38
rs35485154 Chr.1: 62236946-62236947 on Build GRCh38
rs760537672 Chr.1: 62237132-62237135 on Build GRCh38
rs948479037 Chr.1: 62237184-62237184 on Build GRCh38
rs1031280258 Chr.1: 62237269-62237269 on Build GRCh38
rs575320862 Chr.1: 62237135-62237136 on Build GRCh38
rs894104194 Chr.1: 62237178-62237178 on Build GRCh38
rs1046026101 Chr.1: 62237294-62237294 on Build GRCh38
rs976477115 Chr.1: 62237029-62237029 on Build GRCh38
rs1055565492 Chr.1: 62237032-62237032 on Build GRCh38

View Full Product Details