Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39412759-39413252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0754
Entrez Gene ID
643314
Gene Name
KIAA0754
Gene Aliases
-
UniGene
Hs.658760
Gene Chromosome Location
Chr.1: 39409504-39416482 on Build GRCh38
Transcripts
NM_015038.1 NM_012090.5 XM_005270694.1 XM_005270695.1 XM_005270696.1 XM_005270697.1 XM_005270698.1 XM_006710519.1 XM_006710520.2 XM_006710521.1 XM_006710522.1 XM_006710524.2 XM_006710526.3 XM_006710528.3 XM_006710529.3 XM_006710530.3 XM_006710531.3 XM_006710532.3 XM_006710533.2 XM_006710535.3 XM_006710536.2 XM_011541141.2 XM_011541142.2 XM_017000873.1 XM_017000874.1 XM_017000875.1 XM_017000876.1 XM_017000877.1 XM_017000878.1 XM_017000879.1 XM_017000880.1 XM_017000881.1 XM_017000882.1

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
UniGene
Hs.472475
Gene Chromosome Location
Chr.1: 39084167-39487138 on Build GRCh38
Transcripts
NM_015038.1 NM_012090.5 XM_005270694.1 XM_005270695.1 XM_005270696.1 XM_005270697.1 XM_005270698.1 XM_006710519.1 XM_006710520.2 XM_006710521.1 XM_006710522.1 XM_006710524.2 XM_006710526.3 XM_006710528.3 XM_006710529.3 XM_006710530.3 XM_006710531.3 XM_006710532.3 XM_006710533.2 XM_006710535.3 XM_006710536.2 XM_011541141.2 XM_011541142.2 XM_017000873.1 XM_017000874.1 XM_017000875.1 XM_017000876.1 XM_017000877.1 XM_017000878.1 XM_017000879.1 XM_017000880.1 XM_017000881.1 XM_017000882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1646127 c.2245G>A p.V749I Chr.1: 39412918-39412918 on Build GRCh38
5478609 c.2573T>C p.V858A Chr.1: 39413246-39413246 on Build GRCh38
5799753 c.2096C>T p.S699F Chr.1: 39412769-39412769 on Build GRCh38
4945653 c.2461G>T p.A821S Chr.1: 39413134-39413134 on Build GRCh38
1646184 c.2297C>G p.S766* Chr.1: 39412970-39412970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761696935 Chr.1: 39413063-39413063 on Build GRCh38
rs201860546 Chr.1: 39413113-39413113 on Build GRCh38
rs145511340 Chr.1: 39413069-39413069 on Build GRCh38
rs752889576 Chr.1: 39412998-39412998 on Build GRCh38
rs778702944 Chr.1: 39412871-39412871 on Build GRCh38
rs753925820 Chr.1: 39413098-39413098 on Build GRCh38
rs760649455 Chr.1: 39412843-39412843 on Build GRCh38
rs755298631 Chr.1: 39412987-39412987 on Build GRCh38
rs770477858 Chr.1: 39412902-39412902 on Build GRCh38
rs540075106 Chr.1: 39413039-39413039 on Build GRCh38
rs776051331 Chr.1: 39413129-39413129 on Build GRCh38
rs1746842 Chr.1: 39413143-39413143 on Build GRCh38
rs74066781 Chr.1: 39412895-39412895 on Build GRCh38
rs746694571 Chr.1: 39413127-39413127 on Build GRCh38
rs771771927 Chr.1: 39413014-39413014 on Build GRCh38
rs562590902 Chr.1: 39413087-39413087 on Build GRCh38
rs376849966 Chr.1: 39412795-39412795 on Build GRCh38
rs755186662 Chr.1: 39412881-39412881 on Build GRCh38
rs758698472 Chr.1: 39412787-39412787 on Build GRCh38
rs772936286 Chr.1: 39413080-39413080 on Build GRCh38
rs555128110 Chr.1: 39413058-39413058 on Build GRCh38
rs752977560 Chr.1: 39413083-39413083 on Build GRCh38
rs547690826 Chr.1: 39413184-39413184 on Build GRCh38
rs550484193 Chr.1: 39413191-39413191 on Build GRCh38
rs553896360 Chr.1: 39413042-39413042 on Build GRCh38
rs766207387 Chr.1: 39413082-39413082 on Build GRCh38
rs1045374631 Chr.1: 39412796-39412796 on Build GRCh38
rs754642915 Chr.1: 39413198-39413198 on Build GRCh38
rs370273391 Chr.1: 39412999-39412999 on Build GRCh38
rs201118698 Chr.1: 39412813-39412813 on Build GRCh38
rs781548547 Chr.1: 39412997-39412997 on Build GRCh38
rs749784888 Chr.1: 39413003-39413003 on Build GRCh38
rs764357114 Chr.1: 39413161-39413161 on Build GRCh38
rs753725380 Chr.1: 39412963-39412963 on Build GRCh38
rs372642125 Chr.1: 39413128-39413128 on Build GRCh38
rs774338508 Chr.1: 39413224-39413224 on Build GRCh38
rs375863714 Chr.1: 39413134-39413134 on Build GRCh38
rs747575023 Chr.1: 39412954-39412954 on Build GRCh38
rs747686098 Chr.1: 39413200-39413200 on Build GRCh38
rs567834622 Chr.1: 39413185-39413185 on Build GRCh38
rs767040694 Chr.1: 39412837-39412837 on Build GRCh38
rs680528 Chr.1: 39412807-39412807 on Build GRCh38
rs761023006 Chr.1: 39413159-39413159 on Build GRCh38
rs757355981 Chr.1: 39413176-39413176 on Build GRCh38
rs753726510 Chr.1: 39412855-39412855 on Build GRCh38
rs753926462 Chr.1: 39413164-39413164 on Build GRCh38
rs764085645 Chr.1: 39412956-39412956 on Build GRCh38
rs775121634 Chr.1: 39413031-39413031 on Build GRCh38
rs369036896 Chr.1: 39413135-39413135 on Build GRCh38
rs770964363 Chr.1: 39413223-39413223 on Build GRCh38
rs759146003 Chr.1: 39412933-39412933 on Build GRCh38
rs764972948 Chr.1: 39412968-39412968 on Build GRCh38
rs776166898 Chr.1: 39413052-39413052 on Build GRCh38
rs776856717 Chr.1: 39413114-39413114 on Build GRCh38
rs773757866 Chr.1: 39412824-39412824 on Build GRCh38
rs906818102 Chr.1: 39413104-39413104 on Build GRCh38
rs531168361 Chr.1: 39413140-39413140 on Build GRCh38
rs752165466 Chr.1: 39412838-39412838 on Build GRCh38
rs778416857 Chr.1: 39412912-39412912 on Build GRCh38
rs779187368 Chr.1: 39413007-39413007 on Build GRCh38
rs375115807 Chr.1: 39413032-39413032 on Build GRCh38
rs763212123 Chr.1: 39412827-39412827 on Build GRCh38
rs762834563 Chr.1: 39413081-39413081 on Build GRCh38
rs760167168 Chr.1: 39412951-39412951 on Build GRCh38
rs372153671 Chr.1: 39413094-39413094 on Build GRCh38
rs770679544 Chr.1: 39412830-39412830 on Build GRCh38
rs557574081 Chr.1: 39412945-39412945 on Build GRCh38
rs747352331 Chr.1: 39413194-39413194 on Build GRCh38
rs756998787 Chr.1: 39412861-39412861 on Build GRCh38
rs1028835499 Chr.1: 39413149-39413149 on Build GRCh38
rs542491711 Chr.1: 39413086-39413086 on Build GRCh38
rs766295348 Chr.1: 39413144-39413144 on Build GRCh38
rs747181233 Chr.1: 39412797-39412797 on Build GRCh38
rs768630961 Chr.1: 39412801-39412801 on Build GRCh38
rs759395254 Chr.1: 39413228-39413228 on Build GRCh38
rs768829056 Chr.1: 39413229-39413229 on Build GRCh38
rs773876653 Chr.1: 39412831-39412831 on Build GRCh38
rs987442764 Chr.1: 39413017-39413017 on Build GRCh38
rs771433555 Chr.1: 39413005-39413005 on Build GRCh38
rs963948282 Chr.1: 39412918-39412918 on Build GRCh38
rs955996999 Chr.1: 39412916-39412916 on Build GRCh38
rs770391968 Chr.1: 39412815-39412815 on Build GRCh38
rs866089901 Chr.1: 39413008-39413008 on Build GRCh38
rs907928434 Chr.1: 39413048-39413048 on Build GRCh38
rs748694710 Chr.1: 39412814-39412814 on Build GRCh38
rs530448143 Chr.1: 39413186-39413186 on Build GRCh38
rs748460886 Chr.1: 39412819-39412819 on Build GRCh38
rs577618906 Chr.1: 39413011-39413011 on Build GRCh38
rs771684328 Chr.1: 39412943-39412943 on Build GRCh38
rs751751124 Chr.1: 39412873-39412873 on Build GRCh38
rs774252275 Chr.1: 39413146-39413146 on Build GRCh38
rs759174248 Chr.1: 39412834-39412834 on Build GRCh38
rs757756785 Chr.1: 39413109-39413109 on Build GRCh38
rs750199737 Chr.1: 39412982-39412982 on Build GRCh38
rs777365833 Chr.1: 39413212-39413212 on Build GRCh38
rs768988546 Chr.1: 39413000-39413000 on Build GRCh38
rs749733969 Chr.1: 39412915-39412915 on Build GRCh38
rs770173852 Chr.1: 39412851-39412889 on Build GRCh38
rs773641011 Chr.1: 39412939-39412941 on Build GRCh38
rs555439185 Chr.1: 39412854-39412854 on Build GRCh38
rs199819105 Chr.1: 39412927-39412927 on Build GRCh38
rs72928320 Chr.1: 39413221-39413221 on Build GRCh38
rs570319082 Chr.1: 39413202-39413202 on Build GRCh38
rs771216792 Chr.1: 39412924-39412924 on Build GRCh38
rs780891493 Chr.1: 39413199-39413199 on Build GRCh38
rs1002563002 Chr.1: 39412869-39412869 on Build GRCh38
rs377071603 Chr.1: 39413110-39413110 on Build GRCh38

View Full Product Details