Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132696233-132696778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3A
Entrez Gene ID
11127
Gene Name
kinesin family member 3A
Gene Aliases
FLA10, KLP-20
UniGene
Hs.43670
Gene Chromosome Location
Chr.5: 132688808-132737633 on Build GRCh38
Transcripts
NM_001300791.1 NM_001300792.1 NM_007054.6 XM_006714526.3 XM_017008995.1 XM_017008996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6695474 c.2066G>A p.R689H Chr.5: 132696677-132696677 on Build GRCh38
6695473 c.2057G>A p.R686H Chr.5: 132696677-132696677 on Build GRCh38
1727358 c.2068C>T p.P690S Chr.5: 132696666-132696666 on Build GRCh38
1727359 c.2077C>T p.P693S Chr.5: 132696666-132696666 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs528081117 Chr.5: 132696723-132696723 on Build GRCh38
rs765021859 Chr.5: 132696687-132696687 on Build GRCh38
rs535753698 Chr.5: 132696277-132696277 on Build GRCh38
rs1007243663 Chr.5: 132696466-132696466 on Build GRCh38
rs879534366 Chr.5: 132696470-132696470 on Build GRCh38
rs1048954266 Chr.5: 132696575-132696575 on Build GRCh38
rs998020490 Chr.5: 132696398-132696398 on Build GRCh38
rs988517721 Chr.5: 132696402-132696402 on Build GRCh38
rs397760002 Chr.5: 132696276-132696277 on Build GRCh38
rs200200989 Chr.5: 132696584-132696584 on Build GRCh38
rs13355720 Chr.5: 132696719-132696719 on Build GRCh38
rs17166194 Chr.5: 132696330-132696330 on Build GRCh38
rs567802528 Chr.5: 132696396-132696396 on Build GRCh38
rs543655884 Chr.5: 132696426-132696426 on Build GRCh38
rs1050963276 Chr.5: 132696437-132696437 on Build GRCh38
rs938296424 Chr.5: 132696347-132696347 on Build GRCh38
rs557175585 Chr.5: 132696517-132696517 on Build GRCh38
rs374045109 Chr.5: 132696716-132696716 on Build GRCh38
rs565927162 Chr.5: 132696322-132696322 on Build GRCh38
rs772455451 Chr.5: 132696639-132696639 on Build GRCh38
rs183467237 Chr.5: 132696378-132696378 on Build GRCh38
rs776548288 Chr.5: 132696604-132696619 on Build GRCh38
rs919397838 Chr.5: 132696316-132696316 on Build GRCh38
rs777053647 Chr.5: 132696469-132696469 on Build GRCh38
rs928228421 Chr.5: 132696340-132696340 on Build GRCh38
rs1015628822 Chr.5: 132696281-132696281 on Build GRCh38
rs756542237 Chr.5: 132696724-132696724 on Build GRCh38
rs58963671 Chr.5: 132696266-132696267 on Build GRCh38
rs143233559 Chr.5: 132696677-132696677 on Build GRCh38
rs139191778 Chr.5: 132696678-132696678 on Build GRCh38
rs752754929 Chr.5: 132696599-132696599 on Build GRCh38
rs779064367 Chr.5: 132696597-132696597 on Build GRCh38
rs574008530 Chr.5: 132696521-132696521 on Build GRCh38
rs973490315 Chr.5: 132696296-132696296 on Build GRCh38
rs774029743 Chr.5: 132696264-132696264 on Build GRCh38
rs748130062 Chr.5: 132696633-132696633 on Build GRCh38
rs370686985 Chr.5: 132696403-132696403 on Build GRCh38
rs975066627 Chr.5: 132696460-132696460 on Build GRCh38
rs773535182 Chr.5: 132696656-132696656 on Build GRCh38
rs765728118 Chr.5: 132696571-132696571 on Build GRCh38
rs1026359100 Chr.5: 132696581-132696581 on Build GRCh38
rs749801541 Chr.5: 132696585-132696585 on Build GRCh38
rs367715945 Chr.5: 132696596-132696596 on Build GRCh38
rs558132997 Chr.5: 132696265-132696266 on Build GRCh38
rs747665841 Chr.5: 132696495-132696495 on Build GRCh38
rs946391210 Chr.5: 132696741-132696741 on Build GRCh38
rs762326709 Chr.5: 132696710-132696710 on Build GRCh38
rs747603660 Chr.5: 132696326-132696326 on Build GRCh38
rs888505259 Chr.5: 132696260-132696261 on Build GRCh38
rs186688533 Chr.5: 132696664-132696664 on Build GRCh38
rs1054795129 Chr.5: 132696358-132696358 on Build GRCh38
rs12514976 Chr.5: 132696280-132696280 on Build GRCh38
rs950853382 Chr.5: 132696616-132696616 on Build GRCh38
rs930474272 Chr.5: 132696487-132696487 on Build GRCh38
rs1014954555 Chr.5: 132696318-132696318 on Build GRCh38
rs770835605 Chr.5: 132696673-132696673 on Build GRCh38
rs78683393 Chr.5: 132696376-132696376 on Build GRCh38
rs889710223 Chr.5: 132696461-132696461 on Build GRCh38
rs940128963 Chr.5: 132696665-132696665 on Build GRCh38
rs921847395 Chr.5: 132696477-132696477 on Build GRCh38
rs914915026 Chr.5: 132696361-132696361 on Build GRCh38
rs895816291 Chr.5: 132696313-132696313 on Build GRCh38
rs775085316 Chr.5: 132696696-132696696 on Build GRCh38
rs982239120 Chr.5: 132696644-132696644 on Build GRCh38

View Full Product Details