Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11786303-11786845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf167
Entrez Gene ID
284498
Gene Name
chromosome 1 open reading frame 167
Gene Aliases
-
UniGene
Hs.585415
Gene Chromosome Location
Chr.1: 11762188-11789585 on Build GRCh38
Transcripts
NM_001010881.1 XM_011541269.2 XM_011541271.2 XM_011541272.2 XM_011541274.2 XM_011541275.2 XM_011541276.2 XM_011541277.2 XM_011541278.2 XM_011541280.2 XM_011541281.2 XM_017001062.1 XM_017001063.1 XM_017001064.1 NM_005957.4

Gene Symbol
MTHFR
Entrez Gene ID
4524
Gene Name
methylenetetrahydrofolate reductase (NAD(P)H)
Gene Aliases
-
UniGene
Hs.214142
Gene Chromosome Location
Chr.1: 11785730-11806103 on Build GRCh38
Transcripts
NM_001010881.1 XM_011541269.2 XM_011541271.2 XM_011541272.2 XM_011541274.2 XM_011541275.2 XM_011541276.2 XM_011541277.2 XM_011541278.2 XM_011541280.2 XM_011541281.2 XM_017001062.1 XM_017001063.1 XM_017001064.1 NM_005957.4

SNP Detail

SNP ID Chromosome Location
rs373669592 Chr.1: 11786722-11786722 on Build GRCh38
rs573867846 Chr.1: 11786426-11786427 on Build GRCh38
rs57739320 Chr.1: 11786405-11786405 on Build GRCh38
rs921590221 Chr.1: 11786538-11786541 on Build GRCh38
rs957381558 Chr.1: 11786491-11786491 on Build GRCh38
rs896385823 Chr.1: 11786342-11786342 on Build GRCh38
rs774051202 Chr.1: 11786603-11786610 on Build GRCh38
rs553378191 Chr.1: 11786384-11786384 on Build GRCh38
rs534760603 Chr.1: 11786378-11786378 on Build GRCh38
rs372228149 Chr.1: 11786736-11786736 on Build GRCh38
rs746375674 Chr.1: 11786410-11786410 on Build GRCh38
rs4845884 Chr.1: 11786390-11786390 on Build GRCh38
rs781205127 Chr.1: 11786606-11786610 on Build GRCh38
rs565962962 Chr.1: 11786753-11786754 on Build GRCh38
rs550133360 Chr.1: 11786809-11786809 on Build GRCh38
rs1029877485 Chr.1: 11786805-11786805 on Build GRCh38
rs541259931 Chr.1: 11786475-11786475 on Build GRCh38
rs908781340 Chr.1: 11786782-11786782 on Build GRCh38
rs919677776 Chr.1: 11786595-11786595 on Build GRCh38
rs545991628 Chr.1: 11786679-11786679 on Build GRCh38
rs748502645 Chr.1: 11786385-11786385 on Build GRCh38
rs142369131 Chr.1: 11786706-11786706 on Build GRCh38
rs755027235 Chr.1: 11786602-11786603 on Build GRCh38
rs55780505 Chr.1: 11786603-11786606 on Build GRCh38
rs2184227 Chr.1: 11786685-11786685 on Build GRCh38
rs372468963 Chr.1: 11786791-11786791 on Build GRCh38
rs886045172 Chr.1: 11786619-11786622 on Build GRCh38
rs896407305 Chr.1: 11786607-11786607 on Build GRCh38
rs1055165654 Chr.1: 11786602-11786602 on Build GRCh38
rs1033297009 Chr.1: 11786474-11786474 on Build GRCh38
rs142884651 Chr.1: 11786327-11786327 on Build GRCh38
rs886045173 Chr.1: 11786639-11786639 on Build GRCh38
rs561382029 Chr.1: 11786781-11786781 on Build GRCh38
rs775738162 Chr.1: 11786502-11786502 on Build GRCh38
rs975814376 Chr.1: 11786498-11786498 on Build GRCh38
rs563550540 Chr.1: 11786587-11786587 on Build GRCh38
rs189783792 Chr.1: 11786707-11786707 on Build GRCh38
rs764347204 Chr.1: 11786538-11786538 on Build GRCh38
rs932956142 Chr.1: 11786560-11786560 on Build GRCh38
rs1038110415 Chr.1: 11786592-11786592 on Build GRCh38
rs1013428487 Chr.1: 11786346-11786346 on Build GRCh38
rs1042256525 Chr.1: 11786670-11786670 on Build GRCh38
rs1000393582 Chr.1: 11786723-11786723 on Build GRCh38
rs542177264 Chr.1: 11786580-11786580 on Build GRCh38
rs866057115 Chr.1: 11786740-11786740 on Build GRCh38
rs908688875 Chr.1: 11786564-11786564 on Build GRCh38
rs78722668 Chr.1: 11786619-11786623 on Build GRCh38
rs1020393173 Chr.1: 11786713-11786713 on Build GRCh38
rs899118838 Chr.1: 11786655-11786655 on Build GRCh38
rs112469003 Chr.1: 11786427-11786427 on Build GRCh38
rs1033080590 Chr.1: 11786443-11786443 on Build GRCh38
rs976080950 Chr.1: 11786800-11786800 on Build GRCh38
rs923707579 Chr.1: 11786591-11786591 on Build GRCh38
rs575577732 Chr.1: 11786596-11786596 on Build GRCh38
rs1013898996 Chr.1: 11786639-11786639 on Build GRCh38
rs954397595 Chr.1: 11786815-11786815 on Build GRCh38
rs184978486 Chr.1: 11786436-11786436 on Build GRCh38
rs1014353928 Chr.1: 11786397-11786397 on Build GRCh38
rs886045171 Chr.1: 11786413-11786413 on Build GRCh38
rs1037919263 Chr.1: 11786637-11786637 on Build GRCh38
rs774295533 Chr.1: 11786512-11786512 on Build GRCh38
rs886045174 Chr.1: 11786717-11786717 on Build GRCh38
rs961563543 Chr.1: 11786819-11786819 on Build GRCh38

View Full Product Details