Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21395902-21396390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-4
Entrez Gene ID
644524
Gene Name
NK2 homeobox 4
Gene Aliases
NKX2.4, NKX2D
UniGene
Hs.456662
Gene Chromosome Location
Chr.20: 21395367-21397409 on Build GRCh38
Transcripts
NM_033176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6726621 c.1023C>T p.G341G Chr.20: 21395953-21395953 on Build GRCh38
6726620 c.722G>A p.R241Q Chr.20: 21396254-21396254 on Build GRCh38
5365925 c.999C>T p.D333D Chr.20: 21395977-21395977 on Build GRCh38
6726623 c.753G>C p.K251N Chr.20: 21396223-21396223 on Build GRCh38
6726622 c.609G>T p.E203D Chr.20: 21396367-21396367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754977478 Chr.20: 21395955-21395955 on Build GRCh38
rs767040814 Chr.20: 21396318-21396318 on Build GRCh38
rs774228726 Chr.20: 21396369-21396369 on Build GRCh38
rs977074895 Chr.20: 21396087-21396087 on Build GRCh38
rs769037418 Chr.20: 21395987-21395987 on Build GRCh38
rs760982092 Chr.20: 21395996-21395996 on Build GRCh38
rs777268983 Chr.20: 21395973-21395973 on Build GRCh38
rs371741137 Chr.20: 21396343-21396343 on Build GRCh38
rs936774350 Chr.20: 21396127-21396127 on Build GRCh38
rs6035869 Chr.20: 21396223-21396223 on Build GRCh38
rs1053777756 Chr.20: 21396131-21396131 on Build GRCh38
rs746690552 Chr.20: 21395974-21395974 on Build GRCh38
rs767132536 Chr.20: 21396226-21396226 on Build GRCh38
rs747141397 Chr.20: 21396181-21396181 on Build GRCh38
rs771186731 Chr.20: 21396268-21396268 on Build GRCh38
rs748717551 Chr.20: 21396175-21396175 on Build GRCh38
rs922509670 Chr.20: 21396092-21396092 on Build GRCh38
rs1009503627 Chr.20: 21396355-21396355 on Build GRCh38
rs752135229 Chr.20: 21396336-21396336 on Build GRCh38
rs772210720 Chr.20: 21395991-21395991 on Build GRCh38
rs755099896 Chr.20: 21396166-21396177 on Build GRCh38
rs775255385 Chr.20: 21396217-21396217 on Build GRCh38
rs780210342 Chr.20: 21396357-21396357 on Build GRCh38
rs751148824 Chr.20: 21396150-21396150 on Build GRCh38
rs556447303 Chr.20: 21395995-21395995 on Build GRCh38
rs747055518 Chr.20: 21396258-21396258 on Build GRCh38
rs758509573 Chr.20: 21396175-21396176 on Build GRCh38
rs775372230 Chr.20: 21396284-21396284 on Build GRCh38
rs983816903 Chr.20: 21396061-21396061 on Build GRCh38
rs755417325 Chr.20: 21396168-21396168 on Build GRCh38
rs779339429 Chr.20: 21396239-21396239 on Build GRCh38
rs766005689 Chr.20: 21396328-21396328 on Build GRCh38
rs760493283 Chr.20: 21395932-21395932 on Build GRCh38
rs761260962 Chr.20: 21396307-21396307 on Build GRCh38
rs756797212 Chr.20: 21396225-21396225 on Build GRCh38
rs963682882 Chr.20: 21396074-21396074 on Build GRCh38
rs753099139 Chr.20: 21396331-21396331 on Build GRCh38
rs765351890 Chr.20: 21396169-21396170 on Build GRCh38
rs906783211 Chr.20: 21396167-21396175 on Build GRCh38
rs768459805 Chr.20: 21396364-21396364 on Build GRCh38
rs781429920 Chr.20: 21396347-21396347 on Build GRCh38
rs181422340 Chr.20: 21396277-21396277 on Build GRCh38
rs770046656 Chr.20: 21396352-21396352 on Build GRCh38
rs762957553 Chr.20: 21396218-21396218 on Build GRCh38
rs927371693 Chr.20: 21396167-21396167 on Build GRCh38
rs1025853564 Chr.20: 21396059-21396059 on Build GRCh38
rs771978741 Chr.20: 21396177-21396177 on Build GRCh38
rs750107897 Chr.20: 21396228-21396228 on Build GRCh38
rs533976698 Chr.20: 21395931-21395931 on Build GRCh38
rs776944437 Chr.20: 21396271-21396271 on Build GRCh38
rs778079425 Chr.20: 21396182-21396183 on Build GRCh38
rs757797817 Chr.20: 21396136-21396136 on Build GRCh38
rs777377857 Chr.20: 21396149-21396149 on Build GRCh38
rs766217523 Chr.20: 21396001-21396001 on Build GRCh38
rs762017490 Chr.20: 21395921-21395921 on Build GRCh38
rs767600318 Chr.20: 21395930-21395930 on Build GRCh38
rs771275071 Chr.20: 21396183-21396183 on Build GRCh38
rs765056337 Chr.20: 21396097-21396097 on Build GRCh38
rs1017966253 Chr.20: 21396073-21396073 on Build GRCh38
rs780883605 Chr.20: 21396156-21396156 on Build GRCh38
rs376119111 Chr.20: 21396253-21396253 on Build GRCh38
rs749618382 Chr.20: 21396157-21396157 on Build GRCh38
rs757373462 Chr.20: 21396344-21396344 on Build GRCh38
rs755804118 Chr.20: 21396326-21396326 on Build GRCh38
rs753670858 Chr.20: 21396014-21396014 on Build GRCh38
rs748545142 Chr.20: 21396247-21396247 on Build GRCh38
rs777036252 Chr.20: 21396192-21396192 on Build GRCh38
rs539931571 Chr.20: 21395989-21395989 on Build GRCh38
rs375642949 Chr.20: 21396319-21396319 on Build GRCh38
rs576733232 Chr.20: 21396018-21396018 on Build GRCh38
rs1012473160 Chr.20: 21395956-21395956 on Build GRCh38
rs752145952 Chr.20: 21395966-21395966 on Build GRCh38
rs749740599 Chr.20: 21395985-21395985 on Build GRCh38
rs774318763 Chr.20: 21396303-21396303 on Build GRCh38
rs371025178 Chr.20: 21396299-21396299 on Build GRCh38
rs556131053 Chr.20: 21396209-21396209 on Build GRCh38
rs577220230 Chr.20: 21395965-21395965 on Build GRCh38
rs759444663 Chr.20: 21396036-21396036 on Build GRCh38
rs746024545 Chr.20: 21396350-21396350 on Build GRCh38
rs762716640 Chr.20: 21396285-21396285 on Build GRCh38
rs199964105 Chr.20: 21396256-21396256 on Build GRCh38
rs940603948 Chr.20: 21395935-21395935 on Build GRCh38
rs779236107 Chr.20: 21396173-21396173 on Build GRCh38
rs753760933 Chr.20: 21395939-21395939 on Build GRCh38
rs780403586 Chr.20: 21395984-21395984 on Build GRCh38
rs1047213545 Chr.20: 21396174-21396174 on Build GRCh38
rs754465489 Chr.20: 21395971-21395990 on Build GRCh38
rs770108018 Chr.20: 21395980-21395980 on Build GRCh38
rs755254863 Chr.20: 21396236-21396236 on Build GRCh38
rs554235844 Chr.20: 21395959-21395959 on Build GRCh38
rs967308951 Chr.20: 21396297-21396297 on Build GRCh38
rs112136447 Chr.20: 21395970-21395970 on Build GRCh38
rs1046373530 Chr.20: 21396358-21396358 on Build GRCh38
rs756919817 Chr.20: 21396155-21396155 on Build GRCh38
rs763540180 Chr.20: 21396221-21396221 on Build GRCh38
rs752730897 Chr.20: 21396116-21396116 on Build GRCh38
rs773438879 Chr.20: 21396178-21396178 on Build GRCh38
rs752830472 Chr.20: 21396176-21396184 on Build GRCh38
rs977266550 Chr.20: 21396112-21396112 on Build GRCh38
rs542520213 Chr.20: 21396048-21396048 on Build GRCh38
rs1025185706 Chr.20: 21395994-21395994 on Build GRCh38
rs766307645 Chr.20: 21395936-21395936 on Build GRCh38
rs916584520 Chr.20: 21396141-21396141 on Build GRCh38
rs757967753 Chr.20: 21395968-21395968 on Build GRCh38
rs769411856 Chr.20: 21396280-21396280 on Build GRCh38
rs759230500 Chr.20: 21396197-21396197 on Build GRCh38
rs367598502 Chr.20: 21396332-21396332 on Build GRCh38
rs1020428813 Chr.20: 21396370-21396370 on Build GRCh38

View Full Product Details