Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3941038-3941579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMRK2
Entrez Gene ID
27231
Gene Name
nicotinamide riboside kinase 2
Gene Aliases
ITGB1BP3, MIBP, NRK2
UniGene
Hs.135458
Gene Chromosome Location
Chr.19: 3933103-3942416 on Build GRCh38
Transcripts
NM_001289117.1 NM_170678.2 NR_110316.1 XM_006722725.3 XM_006722726.3 XM_017026597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306238 c.445G>A p.V149M Chr.19: 3941120-3941120 on Build GRCh38
4077891 c.502G>T p.V168F Chr.19: 3941177-3941177 on Build GRCh38
6151179 c.444C>A p.H148Q Chr.19: 3941119-3941119 on Build GRCh38
257187 c.435C>T p.F145F Chr.19: 3941110-3941110 on Build GRCh38
7313473 c.428G>T p.G143V Chr.19: 3941103-3941103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952321426 Chr.19: 3941406-3941406 on Build GRCh38
rs771373832 Chr.19: 3941217-3941218 on Build GRCh38
rs759186965 Chr.19: 3941220-3941220 on Build GRCh38
rs763352175 Chr.19: 3941064-3941064 on Build GRCh38
rs767503223 Chr.19: 3941178-3941178 on Build GRCh38
rs773019612 Chr.19: 3941063-3941063 on Build GRCh38
rs751431174 Chr.19: 3941102-3941102 on Build GRCh38
rs776003143 Chr.19: 3941134-3941134 on Build GRCh38
rs776759065 Chr.19: 3941147-3941147 on Build GRCh38
rs200196118 Chr.19: 3941193-3941193 on Build GRCh38
rs992325064 Chr.19: 3941452-3941452 on Build GRCh38
rs61099599 Chr.19: 3941228-3941228 on Build GRCh38
rs746172678 Chr.19: 3941209-3941209 on Build GRCh38
rs979336111 Chr.19: 3941492-3941492 on Build GRCh38
rs545220327 Chr.19: 3941516-3941516 on Build GRCh38
rs373956389 Chr.19: 3941447-3941447 on Build GRCh38
rs34077521 Chr.19: 3941250-3941250 on Build GRCh38
rs754077458 Chr.19: 3941091-3941091 on Build GRCh38
rs748875251 Chr.19: 3941194-3941194 on Build GRCh38
rs199748020 Chr.19: 3941196-3941196 on Build GRCh38
rs891855380 Chr.19: 3941266-3941267 on Build GRCh38
rs553426804 Chr.19: 3941486-3941486 on Build GRCh38
rs867722530 Chr.19: 3941384-3941384 on Build GRCh38
rs748522982 Chr.19: 3941124-3941124 on Build GRCh38
rs774554563 Chr.19: 3941125-3941125 on Build GRCh38
rs913238630 Chr.19: 3941501-3941501 on Build GRCh38
rs756182995 Chr.19: 3941190-3941190 on Build GRCh38
rs761929973 Chr.19: 3941159-3941159 on Build GRCh38
rs112432096 Chr.19: 3941214-3941214 on Build GRCh38
rs1001743722 Chr.19: 3941233-3941233 on Build GRCh38
rs184161969 Chr.19: 3941482-3941482 on Build GRCh38
rs1002174779 Chr.19: 3941532-3941532 on Build GRCh38
rs201360962 Chr.19: 3941116-3941116 on Build GRCh38
rs911177831 Chr.19: 3941416-3941416 on Build GRCh38
rs191710653 Chr.19: 3941288-3941288 on Build GRCh38
rs1015019922 Chr.19: 3941410-3941410 on Build GRCh38
rs779709567 Chr.19: 3941101-3941101 on Build GRCh38
rs781309104 Chr.19: 3941110-3941110 on Build GRCh38
rs567168930 Chr.19: 3941273-3941274 on Build GRCh38
rs397776693 Chr.19: 3941266-3941266 on Build GRCh38
rs754356500 Chr.19: 3941298-3941298 on Build GRCh38
rs201431061 Chr.19: 3941217-3941217 on Build GRCh38
rs960050472 Chr.19: 3941104-3941104 on Build GRCh38
rs534847810 Chr.19: 3941224-3941224 on Build GRCh38
rs199941317 Chr.19: 3941120-3941120 on Build GRCh38
rs747611563 Chr.19: 3941111-3941111 on Build GRCh38
rs965249997 Chr.19: 3941307-3941307 on Build GRCh38
rs557329045 Chr.19: 3941395-3941395 on Build GRCh38
rs976638260 Chr.19: 3941437-3941437 on Build GRCh38
rs755698982 Chr.19: 3941115-3941115 on Build GRCh38
rs760693075 Chr.19: 3941141-3941141 on Build GRCh38
rs565857828 Chr.19: 3941218-3941218 on Build GRCh38
rs1025325452 Chr.19: 3941394-3941394 on Build GRCh38
rs778661754 Chr.19: 3941206-3941206 on Build GRCh38
rs1036254409 Chr.19: 3941364-3941364 on Build GRCh38
rs142587749 Chr.19: 3941119-3941119 on Build GRCh38
rs865900807 Chr.19: 3941266-3941266 on Build GRCh38
rs984039035 Chr.19: 3941411-3941411 on Build GRCh38
rs916376109 Chr.19: 3941476-3941476 on Build GRCh38
rs199684854 Chr.19: 3941127-3941127 on Build GRCh38
rs773495369 Chr.19: 3941219-3941219 on Build GRCh38
rs754838869 Chr.19: 3941105-3941105 on Build GRCh38
rs1057315887 Chr.19: 3941535-3941535 on Build GRCh38
rs111502787 Chr.19: 3941515-3941515 on Build GRCh38
rs960457867 Chr.19: 3941426-3941428 on Build GRCh38
rs764255632 Chr.19: 3941080-3941080 on Build GRCh38
rs202157781 Chr.19: 3941164-3941164 on Build GRCh38
rs150918954 Chr.19: 3941165-3941165 on Build GRCh38
rs773464045 Chr.19: 3941205-3941210 on Build GRCh38
rs747510352 Chr.19: 3941213-3941213 on Build GRCh38
rs947431359 Chr.19: 3941514-3941514 on Build GRCh38
rs754823444 Chr.19: 3941167-3941167 on Build GRCh38
rs140282574 Chr.19: 3941075-3941075 on Build GRCh38
rs764827692 Chr.19: 3941251-3941251 on Build GRCh38
rs547623531 Chr.19: 3941095-3941096 on Build GRCh38
rs775034279 Chr.19: 3941226-3941229 on Build GRCh38
rs760778323 Chr.19: 3941076-3941076 on Build GRCh38
rs761196059 Chr.19: 3941208-3941209 on Build GRCh38
rs942589730 Chr.19: 3941176-3941176 on Build GRCh38
rs760193942 Chr.19: 3941250-3941251 on Build GRCh38
rs1019080648 Chr.19: 3941290-3941290 on Build GRCh38
rs768115503 Chr.19: 3941073-3941073 on Build GRCh38
rs759943907 Chr.19: 3941069-3941069 on Build GRCh38
rs868556298 Chr.19: 3941483-3941483 on Build GRCh38
rs752685316 Chr.19: 3941186-3941186 on Build GRCh38
rs573998386 Chr.19: 3941425-3941425 on Build GRCh38
rs563801293 Chr.19: 3941533-3941533 on Build GRCh38
rs931234996 Chr.19: 3941149-3941149 on Build GRCh38
rs377374741 Chr.19: 3941068-3941068 on Build GRCh38
rs758145718 Chr.19: 3941097-3941097 on Build GRCh38
rs769530595 Chr.19: 3941060-3941060 on Build GRCh38
rs537402774 Chr.19: 3941369-3941369 on Build GRCh38
rs79219472 Chr.19: 3941210-3941210 on Build GRCh38
rs578156628 Chr.19: 3941336-3941336 on Build GRCh38
rs933433437 Chr.19: 3941445-3941445 on Build GRCh38
rs772501742 Chr.19: 3941152-3941153 on Build GRCh38
rs764163881 Chr.19: 3941143-3941143 on Build GRCh38
rs200635027 Chr.19: 3941065-3941065 on Build GRCh38
rs922407754 Chr.19: 3941441-3941441 on Build GRCh38
rs1053099175 Chr.19: 3941464-3941464 on Build GRCh38

View Full Product Details