Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42053113-42053608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKDCC
Entrez Gene ID
91461
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
SGK493, Vlk
UniGene
Hs.408542
Gene Chromosome Location
Chr.2: 42048021-42058528 on Build GRCh38
Transcripts
NM_138370.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583505 c.697A>G p.T233A Chr.2: 42053296-42053296 on Build GRCh38
5643251 c.761G>A p.R254Q Chr.2: 42053360-42053360 on Build GRCh38
6019774 c.238G>A p.E80K Chr.2: 42053266-42053266 on Build GRCh38
4094250 c.304C>T p.L102L Chr.2: 42053332-42053332 on Build GRCh38
4094247 c.660G>A p.Q220Q Chr.2: 42053259-42053259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201891287 Chr.2: 42053348-42053348 on Build GRCh38
rs887040985 Chr.2: 42053159-42053159 on Build GRCh38
rs1028465012 Chr.2: 42053544-42053544 on Build GRCh38
rs900045749 Chr.2: 42053156-42053156 on Build GRCh38
rs754356956 Chr.2: 42053359-42053359 on Build GRCh38
rs769800340 Chr.2: 42053461-42053461 on Build GRCh38
rs991944501 Chr.2: 42053437-42053437 on Build GRCh38
rs868610277 Chr.2: 42053586-42053586 on Build GRCh38
rs746047258 Chr.2: 42053297-42053297 on Build GRCh38
rs892180991 Chr.2: 42053135-42053135 on Build GRCh38
rs774377592 Chr.2: 42053226-42053226 on Build GRCh38
rs938771125 Chr.2: 42053434-42053434 on Build GRCh38
rs776585273 Chr.2: 42053413-42053413 on Build GRCh38
rs747965088 Chr.2: 42053360-42053360 on Build GRCh38
rs143773696 Chr.2: 42053337-42053337 on Build GRCh38
rs996527554 Chr.2: 42053513-42053513 on Build GRCh38
rs770328282 Chr.2: 42053199-42053199 on Build GRCh38
rs144458592 Chr.2: 42053584-42053584 on Build GRCh38
rs763936517 Chr.2: 42053243-42053243 on Build GRCh38
rs943819790 Chr.2: 42053136-42053136 on Build GRCh38
rs921014981 Chr.2: 42053479-42053479 on Build GRCh38
rs772243421 Chr.2: 42053307-42053307 on Build GRCh38
rs960380621 Chr.2: 42053242-42053242 on Build GRCh38
rs776496646 Chr.2: 42053314-42053314 on Build GRCh38
rs943778535 Chr.2: 42053431-42053431 on Build GRCh38
rs900802794 Chr.2: 42053503-42053503 on Build GRCh38
rs570224850 Chr.2: 42053483-42053483 on Build GRCh38
rs1023014127 Chr.2: 42053542-42053542 on Build GRCh38
rs760674135 Chr.2: 42053309-42053309 on Build GRCh38
rs773048540 Chr.2: 42053504-42053504 on Build GRCh38
rs778265917 Chr.2: 42053201-42053201 on Build GRCh38
rs368819157 Chr.2: 42053196-42053196 on Build GRCh38
rs140342791 Chr.2: 42053264-42053264 on Build GRCh38
rs878878186 Chr.2: 42053394-42053394 on Build GRCh38
rs931120185 Chr.2: 42053457-42053457 on Build GRCh38
rs267599376 Chr.2: 42053349-42053349 on Build GRCh38
rs552799618 Chr.2: 42053585-42053585 on Build GRCh38
rs753303221 Chr.2: 42053266-42053266 on Build GRCh38
rs746160665 Chr.2: 42053374-42053374 on Build GRCh38
rs1011736687 Chr.2: 42053564-42053564 on Build GRCh38
rs1040811880 Chr.2: 42053471-42053471 on Build GRCh38
rs35771566 Chr.2: 42053218-42053218 on Build GRCh38
rs34482969 Chr.2: 42053160-42053160 on Build GRCh38
rs749682229 Chr.2: 42053282-42053282 on Build GRCh38
rs377080718 Chr.2: 42053236-42053236 on Build GRCh38
rs757961879 Chr.2: 42053263-42053263 on Build GRCh38
rs754477865 Chr.2: 42053353-42053353 on Build GRCh38
rs528702645 Chr.2: 42053422-42053422 on Build GRCh38
rs180825578 Chr.2: 42053278-42053278 on Build GRCh38
rs758546442 Chr.2: 42053379-42053379 on Build GRCh38
rs553708782 Chr.2: 42053154-42053154 on Build GRCh38
rs916017580 Chr.2: 42053448-42053448 on Build GRCh38
rs551906257 Chr.2: 42053475-42053475 on Build GRCh38
rs761532715 Chr.2: 42053250-42053250 on Build GRCh38
rs201582962 Chr.2: 42053325-42053325 on Build GRCh38
rs762523736 Chr.2: 42053411-42053412 on Build GRCh38
rs768587129 Chr.2: 42053311-42053311 on Build GRCh38
rs144782111 Chr.2: 42053167-42053167 on Build GRCh38
rs189276164 Chr.2: 42053158-42053158 on Build GRCh38
rs759683595 Chr.2: 42053227-42053227 on Build GRCh38
rs988110190 Chr.2: 42053588-42053588 on Build GRCh38
rs1021825576 Chr.2: 42053582-42053582 on Build GRCh38
rs34853050 Chr.2: 42053308-42053308 on Build GRCh38
rs1028081831 Chr.2: 42053249-42053249 on Build GRCh38
rs779227016 Chr.2: 42053296-42053296 on Build GRCh38
rs749998497 Chr.2: 42053257-42053257 on Build GRCh38
rs749585178 Chr.2: 42053202-42053202 on Build GRCh38
rs988187574 Chr.2: 42053389-42053389 on Build GRCh38
rs879003550 Chr.2: 42053163-42053163 on Build GRCh38
rs1004493551 Chr.2: 42053487-42053487 on Build GRCh38
rs756612199 Chr.2: 42053486-42053486 on Build GRCh38
rs1019263876 Chr.2: 42053161-42053161 on Build GRCh38
rs772656705 Chr.2: 42053304-42053304 on Build GRCh38
rs566330548 Chr.2: 42053414-42053414 on Build GRCh38
rs199966849 Chr.2: 42053371-42053371 on Build GRCh38
rs778355868 Chr.2: 42053280-42053280 on Build GRCh38
rs764982138 Chr.2: 42053326-42053326 on Build GRCh38
rs765977836 Chr.2: 42053347-42053347 on Build GRCh38
rs893567700 Chr.2: 42053545-42053545 on Build GRCh38
rs764887140 Chr.2: 42053252-42053252 on Build GRCh38
rs541184823 Chr.2: 42053184-42053184 on Build GRCh38
rs148598102 Chr.2: 42053492-42053492 on Build GRCh38
rs769715486 Chr.2: 42053255-42053255 on Build GRCh38
rs747108914 Chr.2: 42053382-42053382 on Build GRCh38
rs780257888 Chr.2: 42053381-42053381 on Build GRCh38
rs772005408 Chr.2: 42053234-42053234 on Build GRCh38
rs760509862 Chr.2: 42053238-42053238 on Build GRCh38
rs970137597 Chr.2: 42053583-42053583 on Build GRCh38
rs1048244916 Chr.2: 42053467-42053467 on Build GRCh38
rs781735900 Chr.2: 42053195-42053195 on Build GRCh38
rs771372919 Chr.2: 42053290-42053290 on Build GRCh38
rs572180807 Chr.2: 42053175-42053175 on Build GRCh38
rs1041183718 Chr.2: 42053151-42053152 on Build GRCh38
rs768676983 Chr.2: 42053391-42053391 on Build GRCh38
rs1037425383 Chr.2: 42053499-42053499 on Build GRCh38

View Full Product Details