Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207823733-207824279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM3
Entrez Gene ID
389072
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
DAPR, PLEKHM1L
UniGene
Hs.159188
Gene Chromosome Location
Chr.2: 207821288-208025675 on Build GRCh38
Transcripts
NM_001080475.2 XM_005246545.3 XM_011511160.2 XM_017004072.1 XM_017004073.1

SNP Detail

SNP ID Chromosome Location
rs910805867 Chr.2: 207824071-207824071 on Build GRCh38
rs933576149 Chr.2: 207823779-207823779 on Build GRCh38
rs867101380 Chr.2: 207823980-207823980 on Build GRCh38
rs562747397 Chr.2: 207823950-207823950 on Build GRCh38
rs570546710 Chr.2: 207824168-207824168 on Build GRCh38
rs778655180 Chr.2: 207824024-207824024 on Build GRCh38
rs531495045 Chr.2: 207824047-207824047 on Build GRCh38
rs895813941 Chr.2: 207823856-207823856 on Build GRCh38
rs946590316 Chr.2: 207824080-207824080 on Build GRCh38
rs912051288 Chr.2: 207824227-207824227 on Build GRCh38
rs1003415376 Chr.2: 207824160-207824160 on Build GRCh38
rs943538414 Chr.2: 207823809-207823809 on Build GRCh38
rs1035801359 Chr.2: 207823853-207823853 on Build GRCh38
rs1057460403 Chr.2: 207824161-207824161 on Build GRCh38
rs542557659 Chr.2: 207823861-207823861 on Build GRCh38
rs78651674 Chr.2: 207823774-207823774 on Build GRCh38
rs943108666 Chr.2: 207824004-207824004 on Build GRCh38
rs911681025 Chr.2: 207823990-207823990 on Build GRCh38
rs934754281 Chr.2: 207823847-207823847 on Build GRCh38
rs16840057 Chr.2: 207823806-207823806 on Build GRCh38
rs1010833962 Chr.2: 207823797-207823797 on Build GRCh38
rs181568896 Chr.2: 207823907-207823907 on Build GRCh38
rs539567988 Chr.2: 207824173-207824173 on Build GRCh38
rs904996511 Chr.2: 207823883-207823883 on Build GRCh38
rs934700336 Chr.2: 207824113-207824113 on Build GRCh38
rs988143313 Chr.2: 207823805-207823805 on Build GRCh38
rs867451813 Chr.2: 207824051-207824051 on Build GRCh38
rs556750402 Chr.2: 207823794-207823794 on Build GRCh38
rs565341455 Chr.2: 207824123-207824123 on Build GRCh38
rs779692247 Chr.2: 207823865-207823865 on Build GRCh38
rs1020779298 Chr.2: 207823803-207823804 on Build GRCh38
rs1043131243 Chr.2: 207823839-207823839 on Build GRCh38
rs1012956073 Chr.2: 207824166-207824166 on Build GRCh38
rs987624773 Chr.2: 207823962-207823962 on Build GRCh38
rs571371762 Chr.2: 207824158-207824159 on Build GRCh38
rs79899719 Chr.2: 207823866-207823866 on Build GRCh38
rs73986348 Chr.2: 207824164-207824164 on Build GRCh38
rs368823442 Chr.2: 207824226-207824226 on Build GRCh38
rs902324014 Chr.2: 207824143-207824143 on Build GRCh38
rs974506269 Chr.2: 207824245-207824245 on Build GRCh38
rs189571341 Chr.2: 207823894-207823894 on Build GRCh38
rs777991973 Chr.2: 207824209-207824209 on Build GRCh38
rs965016025 Chr.2: 207824079-207824079 on Build GRCh38
rs755711301 Chr.2: 207823842-207823842 on Build GRCh38
rs954372726 Chr.2: 207824219-207824219 on Build GRCh38
rs541030554 Chr.2: 207823893-207823893 on Build GRCh38
rs771812801 Chr.2: 207824185-207824185 on Build GRCh38
rs550104389 Chr.2: 207824174-207824174 on Build GRCh38
rs978925485 Chr.2: 207824092-207824092 on Build GRCh38
rs1038681436 Chr.2: 207824044-207824044 on Build GRCh38
rs76854461 Chr.2: 207823872-207823872 on Build GRCh38
rs145517504 Chr.2: 207824048-207824048 on Build GRCh38
rs1017312592 Chr.2: 207824247-207824247 on Build GRCh38
rs926145078 Chr.2: 207824106-207824106 on Build GRCh38
rs16840060 Chr.2: 207823924-207823924 on Build GRCh38
rs547760544 Chr.2: 207824050-207824050 on Build GRCh38
rs561836224 Chr.2: 207824133-207824133 on Build GRCh38
rs987624265 Chr.2: 207824222-207824222 on Build GRCh38
rs1053094319 Chr.2: 207823855-207823855 on Build GRCh38
rs868267242 Chr.2: 207823929-207823929 on Build GRCh38
rs111280268 Chr.2: 207824228-207824228 on Build GRCh38
rs954527434 Chr.2: 207824013-207824013 on Build GRCh38
rs993166794 Chr.2: 207824195-207824195 on Build GRCh38
rs547431028 Chr.2: 207824167-207824167 on Build GRCh38
rs899009933 Chr.2: 207824191-207824191 on Build GRCh38

View Full Product Details