Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 103138365-103138826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIP5K2
Entrez Gene ID
23262
Gene Name
diphosphoinositol pentakisphosphate kinase 2
Gene Aliases
CFAP160, HISPPD1, IP7K2, VIP2
UniGene
Hs.212046
Gene Chromosome Location
Chr.5: 103120254-103203523 on Build GRCh38
Transcripts
NM_001276277.1 NM_001281471.1 NM_015216.3 XM_005271935.1 XM_005271936.1 XM_005271938.1 XM_005277534.1 XM_005277535.1 XM_005277536.1 XM_005277538.1 XM_005277539.1 XM_005277541.1 XM_006714576.1 XM_006714577.1 XM_011543290.2 XM_017009269.1 XM_017009270.1 XM_017009271.1 XM_017009272.1 XM_017009273.1 XM_017009274.1 XM_017009275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354785 c.478A>T p.N160Y Chr.5: 103138460-103138460 on Build GRCh38
5628827 c.433G>T p.G145C Chr.5: 103138415-103138415 on Build GRCh38
6237337 c.408A>G p.E136E Chr.5: 103138390-103138390 on Build GRCh38
4126965 c.449G>A p.R150H Chr.5: 103138431-103138431 on Build GRCh38
5401885 c.448C>T p.R150C Chr.5: 103138430-103138430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781988239 Chr.5: 103138467-103138467 on Build GRCh38
rs1013425422 Chr.5: 103138549-103138549 on Build GRCh38
rs568428822 Chr.5: 103138760-103138760 on Build GRCh38
rs782748656 Chr.5: 103138476-103138476 on Build GRCh38
rs782306162 Chr.5: 103138416-103138416 on Build GRCh38
rs561882901 Chr.5: 103138757-103138757 on Build GRCh38
rs896293855 Chr.5: 103138799-103138799 on Build GRCh38
rs1055327412 Chr.5: 103138498-103138498 on Build GRCh38
rs782397265 Chr.5: 103138449-103138449 on Build GRCh38
rs140784482 Chr.5: 103138552-103138552 on Build GRCh38
rs896806495 Chr.5: 103138580-103138580 on Build GRCh38
rs978723764 Chr.5: 103138721-103138721 on Build GRCh38
rs113784867 Chr.5: 103138699-103138699 on Build GRCh38
rs782631877 Chr.5: 103138789-103138789 on Build GRCh38
rs138963678 Chr.5: 103138614-103138614 on Build GRCh38
rs376644240 Chr.5: 103138520-103138520 on Build GRCh38
rs781895744 Chr.5: 103138494-103138494 on Build GRCh38
rs782159496 Chr.5: 103138516-103138516 on Build GRCh38
rs564817465 Chr.5: 103138513-103138513 on Build GRCh38
rs913276473 Chr.5: 103138694-103138694 on Build GRCh38
rs1026744582 Chr.5: 103138583-103138583 on Build GRCh38
rs953889950 Chr.5: 103138591-103138591 on Build GRCh38
rs782656158 Chr.5: 103138568-103138568 on Build GRCh38
rs782488252 Chr.5: 103138630-103138630 on Build GRCh38
rs576480866 Chr.5: 103138664-103138664 on Build GRCh38
rs371225804 Chr.5: 103138397-103138397 on Build GRCh38
rs78617252 Chr.5: 103138618-103138618 on Build GRCh38
rs1054978846 Chr.5: 103138798-103138798 on Build GRCh38
rs937398633 Chr.5: 103138766-103138766 on Build GRCh38
rs150102989 Chr.5: 103138597-103138597 on Build GRCh38
rs782491593 Chr.5: 103138393-103138393 on Build GRCh38
rs995255222 Chr.5: 103138582-103138582 on Build GRCh38
rs782580389 Chr.5: 103138430-103138430 on Build GRCh38
rs782190045 Chr.5: 103138415-103138415 on Build GRCh38
rs375563547 Chr.5: 103138448-103138448 on Build GRCh38
rs782095362 Chr.5: 103138470-103138470 on Build GRCh38

View Full Product Details