Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48875799-48876216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R15A
Entrez Gene ID
23645
Gene Name
protein phosphatase 1 regulatory subunit 15A
Gene Aliases
GADD34
UniGene
Hs.631593
Gene Chromosome Location
Chr.19: 48872392-48876062 on Build GRCh38
Transcripts
NM_014330.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1136222 c.1954G>A p.A652T Chr.19: 48875902-48875902 on Build GRCh38
163682 c.2024G>C p.*675S Chr.19: 48875972-48875972 on Build GRCh38
999128 c.1918_1929del12 p.P640_P643delPSSP Chr.19: 48875866-48875877 on Build GRCh38
1612553 c.1889T>A p.L630H Chr.19: 48875837-48875837 on Build GRCh38
3797353 c.1976C>T p.S659L Chr.19: 48875924-48875924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774080120 Chr.19: 48875941-48875945 on Build GRCh38
rs767917201 Chr.19: 48875921-48875921 on Build GRCh38
rs763211405 Chr.19: 48875979-48875979 on Build GRCh38
rs902037568 Chr.19: 48876158-48876158 on Build GRCh38
rs927823179 Chr.19: 48875962-48875962 on Build GRCh38
rs996315796 Chr.19: 48876168-48876168 on Build GRCh38
rs771964492 Chr.19: 48875944-48875944 on Build GRCh38
rs775218534 Chr.19: 48875889-48875889 on Build GRCh38
rs752038705 Chr.19: 48875916-48875916 on Build GRCh38
rs750361540 Chr.19: 48876004-48876005 on Build GRCh38
rs892190138 Chr.19: 48876020-48876020 on Build GRCh38
rs751881103 Chr.19: 48875843-48875843 on Build GRCh38
rs688644 Chr.19: 48876069-48876069 on Build GRCh38
rs771308583 Chr.19: 48875924-48875924 on Build GRCh38
rs35007147 Chr.19: 48875920-48875920 on Build GRCh38
rs746786955 Chr.19: 48875947-48875947 on Build GRCh38
rs763906526 Chr.19: 48875940-48875940 on Build GRCh38
rs772078537 Chr.19: 48875858-48875865 on Build GRCh38
rs780525681 Chr.19: 48876061-48876061 on Build GRCh38
rs755730793 Chr.19: 48875924-48875924 on Build GRCh38
rs758846641 Chr.19: 48875826-48875826 on Build GRCh38
rs774480066 Chr.19: 48875987-48875987 on Build GRCh38
rs953578810 Chr.19: 48875837-48875837 on Build GRCh38
rs763051525 Chr.19: 48875914-48875914 on Build GRCh38
rs760976406 Chr.19: 48876022-48876022 on Build GRCh38
rs373351648 Chr.19: 48875867-48875867 on Build GRCh38
rs770019481 Chr.19: 48875963-48875963 on Build GRCh38
rs764242076 Chr.19: 48875885-48875885 on Build GRCh38
rs745605469 Chr.19: 48875886-48875886 on Build GRCh38
rs767004843 Chr.19: 48876004-48876007 on Build GRCh38
rs370969118 Chr.19: 48875899-48875899 on Build GRCh38
rs777100478 Chr.19: 48875932-48875940 on Build GRCh38
rs186438232 Chr.19: 48875940-48875940 on Build GRCh38
rs766823908 Chr.19: 48875915-48875915 on Build GRCh38
rs759963976 Chr.19: 48875938-48875939 on Build GRCh38
rs773610977 Chr.19: 48875877-48875877 on Build GRCh38
rs767921711 Chr.19: 48876015-48876015 on Build GRCh38
rs755786656 Chr.19: 48875870-48875870 on Build GRCh38
rs191743959 Chr.19: 48876159-48876159 on Build GRCh38
rs747507817 Chr.19: 48875923-48875925 on Build GRCh38
rs755358847 Chr.19: 48875850-48875850 on Build GRCh38
rs753035746 Chr.19: 48876019-48876019 on Build GRCh38
rs1022571033 Chr.19: 48875831-48875831 on Build GRCh38
rs377547229 Chr.19: 48876191-48876196 on Build GRCh38
rs761422376 Chr.19: 48875998-48875998 on Build GRCh38
rs766877382 Chr.19: 48875828-48875828 on Build GRCh38
rs146665963 Chr.19: 48875945-48875945 on Build GRCh38
rs753354896 Chr.19: 48875939-48875939 on Build GRCh38
rs757438814 Chr.19: 48875912-48875912 on Build GRCh38
rs889374433 Chr.19: 48876068-48876068 on Build GRCh38
rs778609437 Chr.19: 48875941-48875941 on Build GRCh38
rs73589818 Chr.19: 48876100-48876100 on Build GRCh38
rs560195365 Chr.19: 48875871-48875871 on Build GRCh38
rs879898438 Chr.19: 48876138-48876138 on Build GRCh38
rs524 Chr.19: 48875910-48875910 on Build GRCh38
rs758633704 Chr.19: 48876060-48876060 on Build GRCh38
rs386810065 Chr.19: 48875910-48875925 on Build GRCh38
rs946386293 Chr.19: 48876059-48876059 on Build GRCh38
rs879109083 Chr.19: 48876090-48876090 on Build GRCh38
rs113181686 Chr.19: 48876167-48876167 on Build GRCh38
rs202191597 Chr.19: 48875888-48875888 on Build GRCh38
rs760073575 Chr.19: 48876006-48876006 on Build GRCh38
rs781458075 Chr.19: 48875863-48875863 on Build GRCh38
rs1016548847 Chr.19: 48875839-48875839 on Build GRCh38
rs527 Chr.19: 48875925-48875925 on Build GRCh38
rs770966592 Chr.19: 48875982-48875982 on Build GRCh38
rs773446345 Chr.19: 48875969-48875969 on Build GRCh38

View Full Product Details