Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111549422-111549825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGR1
Entrez Gene ID
22949
Gene Name
prostaglandin reductase 1
Gene Aliases
LTB4DH, PGR1, ZADH3
UniGene
Hs.584864
Gene Chromosome Location
Chr.9: 111549722-111599855 on Build GRCh38
Transcripts
NM_001146109.1 NM_001007169.3 XM_011518300.2 XM_017014337.1 XM_017014338.1

Gene Symbol
ZNF483
Entrez Gene ID
158399
Gene Name
zinc finger protein 483
Gene Aliases
ZKSCAN16, ZSCAN48
UniGene
Hs.584864
Gene Chromosome Location
Chr.9: 111525159-111577844 on Build GRCh38
Transcripts
NM_001146109.1 NM_001007169.3 XM_011518300.2 XM_017014337.1 XM_017014338.1

SNP Detail

SNP ID Chromosome Location
rs964289698 Chr.9: 111549750-111549750 on Build GRCh38
rs532587811 Chr.9: 111549729-111549729 on Build GRCh38
rs907464725 Chr.9: 111549637-111549637 on Build GRCh38
rs556334651 Chr.9: 111549496-111549496 on Build GRCh38
rs903255389 Chr.9: 111549500-111549500 on Build GRCh38
rs893143321 Chr.9: 111549542-111549542 on Build GRCh38
rs963950476 Chr.9: 111549545-111549545 on Build GRCh38
rs561126273 Chr.9: 111549665-111549665 on Build GRCh38
rs995212336 Chr.9: 111549475-111549475 on Build GRCh38
rs545347979 Chr.9: 111549608-111549608 on Build GRCh38
rs961128552 Chr.9: 111549633-111549633 on Build GRCh38
rs1054363684 Chr.9: 111549539-111549539 on Build GRCh38
rs550842995 Chr.9: 111549673-111549673 on Build GRCh38
rs368701223 Chr.9: 111549584-111549584 on Build GRCh38
rs776298774 Chr.9: 111549737-111549737 on Build GRCh38
rs750662805 Chr.9: 111549467-111549467 on Build GRCh38
rs949358557 Chr.9: 111549696-111549696 on Build GRCh38
rs573682505 Chr.9: 111549463-111549463 on Build GRCh38
rs922067155 Chr.9: 111549578-111549578 on Build GRCh38
rs184731389 Chr.9: 111549655-111549655 on Build GRCh38
rs556642955 Chr.9: 111549446-111549446 on Build GRCh38
rs1028527800 Chr.9: 111549579-111549579 on Build GRCh38
rs937872128 Chr.9: 111549449-111549449 on Build GRCh38
rs1045998334 Chr.9: 111549631-111549631 on Build GRCh38
rs76345666 Chr.9: 111549638-111549638 on Build GRCh38
rs559445101 Chr.9: 111549709-111549709 on Build GRCh38
rs542351891 Chr.9: 111549466-111549466 on Build GRCh38
rs530677259 Chr.9: 111549650-111549650 on Build GRCh38
rs1032607516 Chr.9: 111549775-111549775 on Build GRCh38
rs957854045 Chr.9: 111549776-111549776 on Build GRCh38
rs1019566286 Chr.9: 111549738-111549738 on Build GRCh38
rs766534334 Chr.9: 111549780-111549780 on Build GRCh38
rs1042638439 Chr.9: 111549492-111549492 on Build GRCh38
rs949245774 Chr.9: 111549490-111549493 on Build GRCh38
rs755485449 Chr.9: 111549536-111549536 on Build GRCh38
rs913236757 Chr.9: 111549593-111549593 on Build GRCh38
rs192544648 Chr.9: 111549586-111549586 on Build GRCh38
rs1007894744 Chr.9: 111549734-111549734 on Build GRCh38
rs761614724 Chr.9: 111549765-111549765 on Build GRCh38
rs753892937 Chr.9: 111549516-111549516 on Build GRCh38
rs139173446 Chr.9: 111549668-111549668 on Build GRCh38
rs975632832 Chr.9: 111549569-111549569 on Build GRCh38
rs115554295 Chr.9: 111549483-111549483 on Build GRCh38
rs546674390 Chr.9: 111549671-111549671 on Build GRCh38
rs923791201 Chr.9: 111549740-111549740 on Build GRCh38

View Full Product Details