Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137220849-137221351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF208
Entrez Gene ID
727800
Gene Name
ring finger protein 208
Gene Aliases
-
UniGene
Hs.512767
Gene Chromosome Location
Chr.9: 137220247-137221323 on Build GRCh38
Transcripts
NM_031297.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996468 c.264A>C p.A88A Chr.9: 137220949-137220949 on Build GRCh38
753053 c.358C>G p.Q120E Chr.9: 137220855-137220855 on Build GRCh38
4459553 c.112C>T p.P38S Chr.9: 137221101-137221101 on Build GRCh38
4407048 c.292C>T p.P98S Chr.9: 137220921-137220921 on Build GRCh38
4487859 c.323C>T p.P108L Chr.9: 137220890-137220890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759806317 Chr.9: 137221209-137221209 on Build GRCh38
rs758493588 Chr.9: 137220963-137220963 on Build GRCh38
rs769596695 Chr.9: 137220932-137220932 on Build GRCh38
rs770797630 Chr.9: 137221172-137221172 on Build GRCh38
rs771637039 Chr.9: 137221093-137221093 on Build GRCh38
rs765638765 Chr.9: 137221214-137221214 on Build GRCh38
rs752316592 Chr.9: 137221241-137221241 on Build GRCh38
rs371992033 Chr.9: 137221078-137221078 on Build GRCh38
rs757870967 Chr.9: 137220984-137220984 on Build GRCh38
rs746927197 Chr.9: 137221169-137221169 on Build GRCh38
rs771399696 Chr.9: 137220926-137220926 on Build GRCh38
rs866752508 Chr.9: 137221191-137221191 on Build GRCh38
rs1036834401 Chr.9: 137221268-137221268 on Build GRCh38
rs774807823 Chr.9: 137220933-137220933 on Build GRCh38
rs774319617 Chr.9: 137220940-137220940 on Build GRCh38
rs947139468 Chr.9: 137221330-137221330 on Build GRCh38
rs754164349 Chr.9: 137221138-137221138 on Build GRCh38
rs376748684 Chr.9: 137221042-137221042 on Build GRCh38
rs562245552 Chr.9: 137221197-137221197 on Build GRCh38
rs747659565 Chr.9: 137221090-137221090 on Build GRCh38
rs201379698 Chr.9: 137221119-137221119 on Build GRCh38
rs757882635 Chr.9: 137221164-137221164 on Build GRCh38
rs766147210 Chr.9: 137221040-137221040 on Build GRCh38
rs767255539 Chr.9: 137220949-137220949 on Build GRCh38
rs577867259 Chr.9: 137221073-137221073 on Build GRCh38
rs771491252 Chr.9: 137220877-137220877 on Build GRCh38
rs995568773 Chr.9: 137221319-137221319 on Build GRCh38
rs747403739 Chr.9: 137220876-137220876 on Build GRCh38
rs778808713 Chr.9: 137220917-137220917 on Build GRCh38
rs776411089 Chr.9: 137220880-137220880 on Build GRCh38
rs761983884 Chr.9: 137221116-137221116 on Build GRCh38
rs752928708 Chr.9: 137220961-137220961 on Build GRCh38
rs144152667 Chr.9: 137221102-137221102 on Build GRCh38
rs767766904 Chr.9: 137221117-137221117 on Build GRCh38
rs201921996 Chr.9: 137220921-137220921 on Build GRCh38
rs1041063137 Chr.9: 137221012-137221012 on Build GRCh38
rs542084326 Chr.9: 137221132-137221132 on Build GRCh38
rs148710367 Chr.9: 137221023-137221023 on Build GRCh38
rs374015550 Chr.9: 137221168-137221168 on Build GRCh38
rs988149956 Chr.9: 137220927-137220927 on Build GRCh38
rs747855233 Chr.9: 137221160-137221160 on Build GRCh38
rs374551537 Chr.9: 137220905-137220905 on Build GRCh38
rs199706212 Chr.9: 137221038-137221038 on Build GRCh38
rs759311292 Chr.9: 137220885-137220885 on Build GRCh38
rs910634269 Chr.9: 137221301-137221301 on Build GRCh38
rs763188074 Chr.9: 137221104-137221104 on Build GRCh38
rs767753321 Chr.9: 137220886-137220886 on Build GRCh38
rs745481176 Chr.9: 137220929-137220929 on Build GRCh38
rs778335886 Chr.9: 137221083-137221083 on Build GRCh38
rs774508036 Chr.9: 137221111-137221111 on Build GRCh38
rs768145419 Chr.9: 137221061-137221061 on Build GRCh38
rs539579214 Chr.9: 137221060-137221060 on Build GRCh38
rs577259960 Chr.9: 137221046-137221046 on Build GRCh38
rs751714852 Chr.9: 137221052-137221052 on Build GRCh38
rs754433754 Chr.9: 137221201-137221201 on Build GRCh38
rs768105611 Chr.9: 137220937-137220937 on Build GRCh38
rs200012061 Chr.9: 137220952-137220952 on Build GRCh38
rs764238930 Chr.9: 137221109-137221109 on Build GRCh38
rs1042851887 Chr.9: 137221331-137221331 on Build GRCh38
rs777766977 Chr.9: 137220973-137220973 on Build GRCh38
rs866718580 Chr.9: 137221068-137221068 on Build GRCh38
rs747529317 Chr.9: 137220924-137220924 on Build GRCh38
rs762163588 Chr.9: 137221196-137221196 on Build GRCh38
rs752082200 Chr.9: 137220978-137220978 on Build GRCh38
rs990264530 Chr.9: 137221300-137221300 on Build GRCh38
rs200719013 Chr.9: 137221053-137221053 on Build GRCh38
rs376968453 Chr.9: 137221198-137221198 on Build GRCh38
rs772922762 Chr.9: 137221033-137221033 on Build GRCh38
rs372342634 Chr.9: 137220904-137220904 on Build GRCh38
rs990160130 Chr.9: 137220976-137220976 on Build GRCh38
rs754590756 Chr.9: 137221145-137221145 on Build GRCh38
rs748939477 Chr.9: 137221013-137221013 on Build GRCh38
rs756606861 Chr.9: 137221131-137221131 on Build GRCh38
rs138418399 Chr.9: 137221287-137221287 on Build GRCh38
rs532221535 Chr.9: 137221317-137221317 on Build GRCh38
rs754605951 Chr.9: 137221079-137221079 on Build GRCh38
rs572743786 Chr.9: 137221199-137221199 on Build GRCh38
rs1031478066 Chr.9: 137221251-137221251 on Build GRCh38
rs552183223 Chr.9: 137220918-137220918 on Build GRCh38
rs556117554 Chr.9: 137221096-137221096 on Build GRCh38
rs765498717 Chr.9: 137220953-137220953 on Build GRCh38
rs769506491 Chr.9: 137221000-137221000 on Build GRCh38
rs764108601 Chr.9: 137220888-137220888 on Build GRCh38
rs746998757 Chr.9: 137221099-137221099 on Build GRCh38
rs200615267 Chr.9: 137220887-137220887 on Build GRCh38
rs965115238 Chr.9: 137221307-137221307 on Build GRCh38
rs761579621 Chr.9: 137220899-137220899 on Build GRCh38
rs947003393 Chr.9: 137220974-137220974 on Build GRCh38
rs781651645 Chr.9: 137220991-137220991 on Build GRCh38
rs568207994 Chr.9: 137221003-137221003 on Build GRCh38
rs745657357 Chr.9: 137221186-137221186 on Build GRCh38
rs778532193 Chr.9: 137221153-137221153 on Build GRCh38
rs758078592 Chr.9: 137221242-137221242 on Build GRCh38
rs770728640 Chr.9: 137221101-137221101 on Build GRCh38
rs976072197 Chr.9: 137221310-137221311 on Build GRCh38
rs554581992 Chr.9: 137220997-137220997 on Build GRCh38
rs750239611 Chr.9: 137220951-137220951 on Build GRCh38
rs541256094 Chr.9: 137221249-137221249 on Build GRCh38
rs774706066 Chr.9: 137221187-137221187 on Build GRCh38
rs761774045 Chr.9: 137220942-137220942 on Build GRCh38
rs988723015 Chr.9: 137221221-137221221 on Build GRCh38
rs764195215 Chr.9: 137221048-137221048 on Build GRCh38
rs751041581 Chr.9: 137221066-137221066 on Build GRCh38
rs768178758 Chr.9: 137221022-137221022 on Build GRCh38
rs912957658 Chr.9: 137221323-137221323 on Build GRCh38

View Full Product Details