Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 17961697-17962198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A12
Entrez Gene ID
221074
Gene Name
solute carrier family 39 member 12
Gene Aliases
LZT-Hs8, ZIP-12, bA570F3.1
UniGene
Hs.350895
Gene Chromosome Location
Chr.10: 17951839-18043292 on Build GRCh38
Transcripts
NM_001145195.1 NM_001282733.1 NM_001282734.1 NM_152725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7410933 c.438C>T p.S146S Chr.10: 17961757-17961757 on Build GRCh38
6487641 c.510C>T p.F170F Chr.10: 17961829-17961829 on Build GRCh38
5050082 c.486T>C p.N162N Chr.10: 17961805-17961805 on Build GRCh38
4013159 c.497A>G p.D166G Chr.10: 17961816-17961816 on Build GRCh38
4013157 c.479C>A p.S160* Chr.10: 17961798-17961798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781834709 Chr.10: 17961747-17961747 on Build GRCh38
rs562394019 Chr.10: 17962149-17962149 on Build GRCh38
rs782120297 Chr.10: 17961730-17961730 on Build GRCh38
rs374474314 Chr.10: 17961872-17961872 on Build GRCh38
rs143514081 Chr.10: 17961847-17961847 on Build GRCh38
rs782183115 Chr.10: 17961911-17961911 on Build GRCh38
rs782630024 Chr.10: 17961898-17961898 on Build GRCh38
rs569987735 Chr.10: 17961722-17961722 on Build GRCh38
rs782292023 Chr.10: 17961900-17961900 on Build GRCh38
rs782660395 Chr.10: 17961773-17961773 on Build GRCh38
rs782805699 Chr.10: 17961848-17961848 on Build GRCh38
rs781960824 Chr.10: 17961788-17961788 on Build GRCh38
rs994720172 Chr.10: 17961850-17961850 on Build GRCh38
rs80088795 Chr.10: 17962062-17962062 on Build GRCh38
rs200500691 Chr.10: 17961784-17961784 on Build GRCh38
rs987616367 Chr.10: 17962065-17962065 on Build GRCh38
rs1003349665 Chr.10: 17962133-17962133 on Build GRCh38
rs782189625 Chr.10: 17961777-17961777 on Build GRCh38
rs782718234 Chr.10: 17961825-17961825 on Build GRCh38
rs781868690 Chr.10: 17961829-17961829 on Build GRCh38
rs782382519 Chr.10: 17961803-17961803 on Build GRCh38
rs1056719054 Chr.10: 17962141-17962141 on Build GRCh38
rs1035140400 Chr.10: 17961928-17961928 on Build GRCh38
rs782814170 Chr.10: 17961733-17961733 on Build GRCh38
rs576027176 Chr.10: 17962093-17962093 on Build GRCh38
rs180702675 Chr.10: 17961943-17961943 on Build GRCh38
rs781790857 Chr.10: 17961816-17961816 on Build GRCh38
rs557020284 Chr.10: 17961922-17961922 on Build GRCh38
rs1022307524 Chr.10: 17962020-17962020 on Build GRCh38
rs782066994 Chr.10: 17962094-17962094 on Build GRCh38
rs1038638351 Chr.10: 17962089-17962089 on Build GRCh38
rs776917335 Chr.10: 17962070-17962070 on Build GRCh38
rs781915741 Chr.10: 17962091-17962091 on Build GRCh38
rs782403101 Chr.10: 17961904-17961904 on Build GRCh38
rs782413228 Chr.10: 17961767-17961767 on Build GRCh38
rs782039964 Chr.10: 17961804-17961804 on Build GRCh38
rs550320751 Chr.10: 17962110-17962110 on Build GRCh38
rs139205048 Chr.10: 17961754-17961754 on Build GRCh38
rs781884801 Chr.10: 17961855-17961855 on Build GRCh38
rs377176790 Chr.10: 17961844-17961844 on Build GRCh38
rs556080783 Chr.10: 17961871-17961871 on Build GRCh38
rs782132198 Chr.10: 17961793-17961793 on Build GRCh38
rs782637785 Chr.10: 17961752-17961752 on Build GRCh38
rs782097594 Chr.10: 17961819-17961819 on Build GRCh38
rs782477663 Chr.10: 17961749-17961749 on Build GRCh38
rs782006981 Chr.10: 17962023-17962023 on Build GRCh38
rs367590715 Chr.10: 17961888-17961888 on Build GRCh38
rs553538069 Chr.10: 17961983-17961983 on Build GRCh38
rs782220089 Chr.10: 17961879-17961879 on Build GRCh38
rs782578067 Chr.10: 17961740-17961740 on Build GRCh38
rs372299361 Chr.10: 17961805-17961805 on Build GRCh38
rs148007591 Chr.10: 17962069-17962069 on Build GRCh38
rs782455653 Chr.10: 17961739-17961739 on Build GRCh38
rs551808692 Chr.10: 17961972-17961972 on Build GRCh38
rs781888329 Chr.10: 17961738-17961738 on Build GRCh38
rs781938680 Chr.10: 17961905-17961905 on Build GRCh38
rs924105898 Chr.10: 17962057-17962057 on Build GRCh38
rs1887217 Chr.10: 17962024-17962024 on Build GRCh38
rs782714669 Chr.10: 17961815-17961815 on Build GRCh38

View Full Product Details