Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62881594-62882092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC3A2
Entrez Gene ID
6520
Gene Name
solute carrier family 3 member 2
Gene Aliases
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE
UniGene
Hs.502769
Gene Chromosome Location
Chr.11: 62856012-62888883 on Build GRCh38
Transcripts
NM_001012662.2 NM_001012664.2 NM_001013251.2 NM_002394.5 NR_037193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4855614 c.821-3C>T p.? Chr.11: 62881890-62881890 on Build GRCh38
6782219 c.824T>G p.L275R Chr.11: 62881896-62881896 on Build GRCh38
7322271 c.894C>T p.N298N Chr.11: 62881966-62881966 on Build GRCh38
2161973 c.984_985insA p.S332fs*15 Chr.11: 62882056-62882057 on Build GRCh38
7290794 c.851C>T p.S284F Chr.11: 62881923-62881923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368712824 Chr.11: 62881849-62881849 on Build GRCh38
rs764836432 Chr.11: 62882042-62882042 on Build GRCh38
rs897013786 Chr.11: 62881746-62881746 on Build GRCh38
rs778611933 Chr.11: 62881850-62881850 on Build GRCh38
rs770255364 Chr.11: 62882057-62882067 on Build GRCh38
rs747648124 Chr.11: 62882065-62882065 on Build GRCh38
rs1006989743 Chr.11: 62881837-62881837 on Build GRCh38
rs998830518 Chr.11: 62881784-62881784 on Build GRCh38
rs756468277 Chr.11: 62881945-62881945 on Build GRCh38
rs73487879 Chr.11: 62881676-62881676 on Build GRCh38
rs556782167 Chr.11: 62881678-62881678 on Build GRCh38
rs758143162 Chr.11: 62882054-62882054 on Build GRCh38
rs530651178 Chr.11: 62881905-62881905 on Build GRCh38
rs78298849 Chr.11: 62881955-62881955 on Build GRCh38
rs80190679 Chr.11: 62881991-62881991 on Build GRCh38
rs746906877 Chr.11: 62881969-62881969 on Build GRCh38
rs778699846 Chr.11: 62881948-62881948 on Build GRCh38
rs112685148 Chr.11: 62881654-62881654 on Build GRCh38
rs771551841 Chr.11: 62882052-62882055 on Build GRCh38
rs912772777 Chr.11: 62881648-62881648 on Build GRCh38
rs765733324 Chr.11: 62881893-62881893 on Build GRCh38
rs746119027 Chr.11: 62881974-62881974 on Build GRCh38
rs931081499 Chr.11: 62881704-62881704 on Build GRCh38
rs1013827168 Chr.11: 62881741-62881741 on Build GRCh38
rs765852673 Chr.11: 62881931-62881931 on Build GRCh38
rs559343780 Chr.11: 62881824-62881824 on Build GRCh38
rs551960879 Chr.11: 62881935-62881935 on Build GRCh38
rs11548759 Chr.11: 62881939-62881939 on Build GRCh38
rs919613436 Chr.11: 62881936-62881942 on Build GRCh38
rs754585041 Chr.11: 62881943-62881943 on Build GRCh38
rs987336075 Chr.11: 62881840-62881840 on Build GRCh38
rs545471809 Chr.11: 62881804-62881804 on Build GRCh38
rs750013503 Chr.11: 62881920-62881920 on Build GRCh38
rs912833742 Chr.11: 62881853-62881853 on Build GRCh38
rs542576739 Chr.11: 62881904-62881904 on Build GRCh38
rs1046315837 Chr.11: 62881756-62881756 on Build GRCh38
rs933788989 Chr.11: 62881788-62881788 on Build GRCh38
rs546675254 Chr.11: 62882022-62882022 on Build GRCh38
rs767616332 Chr.11: 62881796-62881796 on Build GRCh38
rs765905019 Chr.11: 62882033-62882033 on Build GRCh38
rs771243821 Chr.11: 62881879-62881879 on Build GRCh38
rs1050995660 Chr.11: 62881803-62881803 on Build GRCh38
rs756460371 Chr.11: 62881640-62881640 on Build GRCh38
rs753492632 Chr.11: 62881936-62881936 on Build GRCh38
rs927496927 Chr.11: 62881663-62881663 on Build GRCh38
rs1010006638 Chr.11: 62881894-62881894 on Build GRCh38
rs770920417 Chr.11: 62881972-62881972 on Build GRCh38
rs772769834 Chr.11: 62882056-62882057 on Build GRCh38
rs940775202 Chr.11: 62881709-62881709 on Build GRCh38
rs777560601 Chr.11: 62881959-62881959 on Build GRCh38
rs574307727 Chr.11: 62881652-62881652 on Build GRCh38
rs778629415 Chr.11: 62882039-62882039 on Build GRCh38
rs775937542 Chr.11: 62882069-62882076 on Build GRCh38
rs776816882 Chr.11: 62881973-62881973 on Build GRCh38
rs748115145 Chr.11: 62881851-62881851 on Build GRCh38
rs760089491 Chr.11: 62882057-62882058 on Build GRCh38
rs750842231 Chr.11: 62881862-62881862 on Build GRCh38
rs138578664 Chr.11: 62882038-62882038 on Build GRCh38
rs759766081 Chr.11: 62881884-62881884 on Build GRCh38
rs771854350 Chr.11: 62881856-62881856 on Build GRCh38
rs116161785 Chr.11: 62881901-62881901 on Build GRCh38
rs755562137 Chr.11: 62881927-62881927 on Build GRCh38
rs777007713 Chr.11: 62881881-62881881 on Build GRCh38
rs1031603255 Chr.11: 62881785-62881785 on Build GRCh38
rs895017493 Chr.11: 62881730-62881730 on Build GRCh38
rs369040136 Chr.11: 62882058-62882058 on Build GRCh38
rs576729985 Chr.11: 62881699-62881699 on Build GRCh38
rs939672000 Chr.11: 62881668-62881668 on Build GRCh38
rs570866379 Chr.11: 62881997-62881997 on Build GRCh38
rs770121813 Chr.11: 62881981-62881981 on Build GRCh38
rs759547952 Chr.11: 62881632-62881633 on Build GRCh38
rs145066571 Chr.11: 62881949-62881949 on Build GRCh38
rs377656242 Chr.11: 62881993-62881993 on Build GRCh38
rs1036378346 Chr.11: 62881728-62881728 on Build GRCh38
rs533509167 Chr.11: 62882005-62882005 on Build GRCh38
rs775951412 Chr.11: 62881897-62881897 on Build GRCh38
rs780448539 Chr.11: 62881759-62881759 on Build GRCh38
rs752547667 Chr.11: 62882049-62882049 on Build GRCh38
rs747057109 Chr.11: 62881877-62881877 on Build GRCh38

View Full Product Details