Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27199846-27200344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A6
Entrez Gene ID
8884
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
UniGene
Hs.435735
Gene Chromosome Location
Chr.2: 27199587-27212307 on Build GRCh38
Transcripts
NM_021095.2 NR_028323.1 XM_006712128.1 XM_006712129.1 XM_006712130.1 XM_011533146.2 XM_017005216.1

SNP Detail

SNP ID Chromosome Location
rs894672467 Chr.2: 27200184-27200184 on Build GRCh38
rs1031716679 Chr.2: 27200235-27200235 on Build GRCh38
rs537794663 Chr.2: 27200282-27200282 on Build GRCh38
rs891214529 Chr.2: 27200261-27200261 on Build GRCh38
rs535676667 Chr.2: 27200014-27200014 on Build GRCh38
rs998847630 Chr.2: 27200232-27200232 on Build GRCh38
rs900042488 Chr.2: 27200236-27200236 on Build GRCh38
rs989977278 Chr.2: 27199896-27199896 on Build GRCh38
rs556606410 Chr.2: 27200081-27200081 on Build GRCh38
rs898077233 Chr.2: 27200070-27200070 on Build GRCh38
rs927709076 Chr.2: 27199931-27199931 on Build GRCh38
rs576665544 Chr.2: 27200136-27200136 on Build GRCh38
rs886768751 Chr.2: 27200033-27200033 on Build GRCh38
rs55786105 Chr.2: 27200320-27200320 on Build GRCh38
rs879006827 Chr.2: 27200118-27200118 on Build GRCh38
rs941661807 Chr.2: 27200048-27200048 on Build GRCh38
rs1039010791 Chr.2: 27200148-27200148 on Build GRCh38
rs553288948 Chr.2: 27199913-27199913 on Build GRCh38
rs756529262 Chr.2: 27200209-27200209 on Build GRCh38
rs768977463 Chr.2: 27200317-27200317 on Build GRCh38
rs1031704368 Chr.2: 27200299-27200299 on Build GRCh38
rs972371901 Chr.2: 27200264-27200264 on Build GRCh38
rs568295034 Chr.2: 27199978-27199978 on Build GRCh38
rs912281847 Chr.2: 27200259-27200259 on Build GRCh38
rs921120023 Chr.2: 27199926-27199927 on Build GRCh38
rs1036665889 Chr.2: 27200062-27200062 on Build GRCh38
rs907288356 Chr.2: 27200225-27200225 on Build GRCh38
rs191510120 Chr.2: 27200245-27200245 on Build GRCh38
rs930935710 Chr.2: 27200080-27200080 on Build GRCh38
rs987042331 Chr.2: 27200256-27200256 on Build GRCh38
rs189176336 Chr.2: 27200200-27200200 on Build GRCh38
rs796463488 Chr.2: 27200290-27200290 on Build GRCh38
rs996984107 Chr.2: 27200244-27200244 on Build GRCh38
rs1046729059 Chr.2: 27199942-27199942 on Build GRCh38
rs749599074 Chr.2: 27200286-27200286 on Build GRCh38
rs932378851 Chr.2: 27199940-27199940 on Build GRCh38
rs939058132 Chr.2: 27199933-27199933 on Build GRCh38
rs376605944 Chr.2: 27200097-27200099 on Build GRCh38
rs140531837 Chr.2: 27200273-27200273 on Build GRCh38

View Full Product Details