Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47871313-47871726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT2A1
Entrez Gene ID
6822
Gene Name
sulfotransferase family 2A member 1
Gene Aliases
DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa
UniGene
Hs.515835
Gene Chromosome Location
Chr.19: 47870466-47886397 on Build GRCh38
Transcripts
NM_003167.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237858 c.746-3C>T p.? Chr.19: 47871570-47871570 on Build GRCh38
1612499 c.833_835delCTC p.P278delP Chr.19: 47871478-47871480 on Build GRCh38
5954685 c.757G>A p.D253N Chr.19: 47871556-47871556 on Build GRCh38
1394972 c.762G>T p.W254C Chr.19: 47871551-47871551 on Build GRCh38
3536387 c.776C>T p.T259I Chr.19: 47871537-47871537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768105841 Chr.19: 47871450-47871450 on Build GRCh38
rs749212128 Chr.19: 47871556-47871556 on Build GRCh38
rs764010986 Chr.19: 47871371-47871371 on Build GRCh38
rs753928755 Chr.19: 47871529-47871529 on Build GRCh38
rs73942749 Chr.19: 47871678-47871678 on Build GRCh38
rs751842572 Chr.19: 47871696-47871696 on Build GRCh38
rs1037716014 Chr.19: 47871649-47871649 on Build GRCh38
rs570625604 Chr.19: 47871521-47871522 on Build GRCh38
rs772645576 Chr.19: 47871435-47871435 on Build GRCh38
rs761236594 Chr.19: 47871448-47871448 on Build GRCh38
rs933420398 Chr.19: 47871376-47871376 on Build GRCh38
rs866659042 Chr.19: 47871549-47871549 on Build GRCh38
rs1032191564 Chr.19: 47871481-47871481 on Build GRCh38
rs1013330593 Chr.19: 47871458-47871458 on Build GRCh38
rs899431218 Chr.19: 47871431-47871431 on Build GRCh38
rs954734465 Chr.19: 47871563-47871563 on Build GRCh38
rs11569677 Chr.19: 47871389-47871389 on Build GRCh38
rs750526898 Chr.19: 47871504-47871504 on Build GRCh38
rs11569679 Chr.19: 47871532-47871532 on Build GRCh38
rs776093287 Chr.19: 47871454-47871454 on Build GRCh38
rs940712109 Chr.19: 47871393-47871393 on Build GRCh38
rs907972306 Chr.19: 47871592-47871592 on Build GRCh38
rs768507982 Chr.19: 47871599-47871599 on Build GRCh38
rs776144203 Chr.19: 47871610-47871610 on Build GRCh38
rs771248364 Chr.19: 47871422-47871422 on Build GRCh38
rs149563641 Chr.19: 47871477-47871477 on Build GRCh38
rs11569678 Chr.19: 47871378-47871378 on Build GRCh38
rs62529880 Chr.19: 47871375-47871375 on Build GRCh38
rs758752472 Chr.19: 47871564-47871564 on Build GRCh38
rs577627993 Chr.19: 47871593-47871593 on Build GRCh38
rs779218418 Chr.19: 47871539-47871539 on Build GRCh38
rs746239667 Chr.19: 47871541-47871541 on Build GRCh38
rs766193413 Chr.19: 47871492-47871492 on Build GRCh38
rs1034520814 Chr.19: 47871453-47871453 on Build GRCh38
rs750577874 Chr.19: 47871396-47871396 on Build GRCh38
rs779450275 Chr.19: 47871426-47871426 on Build GRCh38
rs376834269 Chr.19: 47871443-47871443 on Build GRCh38
rs559862922 Chr.19: 47871657-47871657 on Build GRCh38
rs756265293 Chr.19: 47871669-47871669 on Build GRCh38
rs201140355 Chr.19: 47871470-47871470 on Build GRCh38
rs754922906 Chr.19: 47871522-47871522 on Build GRCh38
rs780449727 Chr.19: 47871576-47871576 on Build GRCh38
rs749572700 Chr.19: 47871421-47871421 on Build GRCh38
rs933609056 Chr.19: 47871347-47871347 on Build GRCh38
rs1020695747 Chr.19: 47871385-47871385 on Build GRCh38
rs746186661 Chr.19: 47871432-47871432 on Build GRCh38
rs761487701 Chr.19: 47871464-47871464 on Build GRCh38
rs764311389 Chr.19: 47871525-47871525 on Build GRCh38
rs375473200 Chr.19: 47871583-47871583 on Build GRCh38
rs764823849 Chr.19: 47871466-47871466 on Build GRCh38
rs374682324 Chr.19: 47871430-47871430 on Build GRCh38
rs933484987 Chr.19: 47871620-47871620 on Build GRCh38
rs368569312 Chr.19: 47871614-47871614 on Build GRCh38
rs62129966 Chr.19: 47871693-47871693 on Build GRCh38
rs945008721 Chr.19: 47871677-47871677 on Build GRCh38
rs1051989842 Chr.19: 47871644-47871644 on Build GRCh38
rs751509523 Chr.19: 47871506-47871506 on Build GRCh38
rs1001671424 Chr.19: 47871445-47871445 on Build GRCh38

View Full Product Details