Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39374342-39374833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNGR1
Entrez Gene ID
9145
Gene Name
synaptogyrin 1
Gene Aliases
-
UniGene
Hs.216226
Gene Chromosome Location
Chr.22: 39349949-39385588 on Build GRCh38
Transcripts
NM_004711.4 NM_145731.3 NM_145738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004841 c.133G>A p.V45M Chr.22: 39374346-39374346 on Build GRCh38
3554508 c.279C>T p.F93F Chr.22: 39374492-39374492 on Build GRCh38
6004840 c.130G>A p.V44M Chr.22: 39374346-39374346 on Build GRCh38
6162241 c.255G>T p.L85L Chr.22: 39374468-39374468 on Build GRCh38
3673284 c.330C>T p.I110I Chr.22: 39374543-39374543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776404998 Chr.22: 39374563-39374563 on Build GRCh38
rs778231606 Chr.22: 39374448-39374448 on Build GRCh38
rs764323044 Chr.22: 39374612-39374612 on Build GRCh38
rs1044896873 Chr.22: 39374544-39374544 on Build GRCh38
rs767447547 Chr.22: 39374672-39374672 on Build GRCh38
rs566040065 Chr.22: 39374724-39374724 on Build GRCh38
rs748528372 Chr.22: 39374388-39374388 on Build GRCh38
rs754188710 Chr.22: 39374619-39374619 on Build GRCh38
rs779450717 Chr.22: 39374624-39374624 on Build GRCh38
rs749080782 Chr.22: 39374656-39374656 on Build GRCh38
rs780778010 Chr.22: 39374641-39374641 on Build GRCh38
rs765177986 Chr.22: 39374494-39374494 on Build GRCh38
rs774197556 Chr.22: 39374604-39374604 on Build GRCh38
rs979683074 Chr.22: 39374737-39374737 on Build GRCh38
rs201445314 Chr.22: 39374502-39374502 on Build GRCh38
rs780313877 Chr.22: 39374574-39374575 on Build GRCh38
rs757601480 Chr.22: 39374620-39374620 on Build GRCh38
rs1055344969 Chr.22: 39374540-39374540 on Build GRCh38
rs747678240 Chr.22: 39374643-39374643 on Build GRCh38
rs745901428 Chr.22: 39374378-39374378 on Build GRCh38
rs771419558 Chr.22: 39374537-39374537 on Build GRCh38
rs897777401 Chr.22: 39374800-39374800 on Build GRCh38
rs937970592 Chr.22: 39374538-39374538 on Build GRCh38
rs753637119 Chr.22: 39374425-39374425 on Build GRCh38
rs775498307 Chr.22: 39374680-39374680 on Build GRCh38
rs756224878 Chr.22: 39374549-39374550 on Build GRCh38
rs770681256 Chr.22: 39374659-39374659 on Build GRCh38
rs771345585 Chr.22: 39374462-39374462 on Build GRCh38
rs373188541 Chr.22: 39374439-39374439 on Build GRCh38
rs759640966 Chr.22: 39374606-39374606 on Build GRCh38
rs1032633162 Chr.22: 39374739-39374739 on Build GRCh38
rs868709703 Chr.22: 39374493-39374493 on Build GRCh38
rs926468845 Chr.22: 39374778-39374778 on Build GRCh38
rs868255177 Chr.22: 39374664-39374664 on Build GRCh38
rs56363153 Chr.22: 39374507-39374507 on Build GRCh38
rs751611540 Chr.22: 39374504-39374504 on Build GRCh38
rs200512399 Chr.22: 39374442-39374442 on Build GRCh38
rs201697745 Chr.22: 39374466-39374466 on Build GRCh38
rs929202963 Chr.22: 39374804-39374804 on Build GRCh38
rs375288974 Chr.22: 39374430-39374430 on Build GRCh38
rs568521101 Chr.22: 39374577-39374577 on Build GRCh38
rs376168336 Chr.22: 39374484-39374484 on Build GRCh38
rs769526625 Chr.22: 39374645-39374645 on Build GRCh38
rs570397080 Chr.22: 39374435-39374435 on Build GRCh38
rs137867913 Chr.22: 39374474-39374474 on Build GRCh38
rs774333802 Chr.22: 39374668-39374668 on Build GRCh38
rs4820377 Chr.22: 39374775-39374775 on Build GRCh38
rs775005783 Chr.22: 39374465-39374465 on Build GRCh38
rs774993653 Chr.22: 39374404-39374404 on Build GRCh38
rs369122831 Chr.22: 39374438-39374438 on Build GRCh38
rs749665196 Chr.22: 39374533-39374533 on Build GRCh38
rs1027868191 Chr.22: 39374444-39374444 on Build GRCh38
rs779317839 Chr.22: 39374545-39374545 on Build GRCh38
rs747309966 Chr.22: 39374381-39374381 on Build GRCh38
rs966205771 Chr.22: 39374695-39374695 on Build GRCh38
rs771662686 Chr.22: 39374403-39374403 on Build GRCh38
rs917834135 Chr.22: 39374761-39374761 on Build GRCh38
rs115437880 Chr.22: 39374375-39374375 on Build GRCh38
rs138689916 Chr.22: 39374791-39374791 on Build GRCh38
rs976225530 Chr.22: 39374717-39374717 on Build GRCh38
rs964313279 Chr.22: 39374440-39374440 on Build GRCh38
rs773088722 Chr.22: 39374583-39374583 on Build GRCh38
rs760308694 Chr.22: 39374409-39374409 on Build GRCh38
rs35429467 Chr.22: 39374495-39374495 on Build GRCh38
rs749852308 Chr.22: 39374458-39374458 on Build GRCh38
rs534706274 Chr.22: 39374552-39374552 on Build GRCh38
rs114689408 Chr.22: 39374483-39374483 on Build GRCh38
rs781363251 Chr.22: 39374508-39374508 on Build GRCh38
rs61637539 Chr.22: 39374750-39374750 on Build GRCh38
rs201803771 Chr.22: 39374369-39374369 on Build GRCh38
rs757491851 Chr.22: 39374742-39374742 on Build GRCh38
rs371659731 Chr.22: 39374429-39374429 on Build GRCh38
rs752966909 Chr.22: 39374510-39374510 on Build GRCh38
rs1037938814 Chr.22: 39374795-39374795 on Build GRCh38
rs756458157 Chr.22: 39374528-39374528 on Build GRCh38
rs754526827 Chr.22: 39374630-39374630 on Build GRCh38
rs990786108 Chr.22: 39374757-39374757 on Build GRCh38
rs539862807 Chr.22: 39374748-39374748 on Build GRCh38
rs770239217 Chr.22: 39374394-39374394 on Build GRCh38
rs375959309 Chr.22: 39374395-39374395 on Build GRCh38
rs746495829 Chr.22: 39374547-39374547 on Build GRCh38
rs778178266 Chr.22: 39374529-39374529 on Build GRCh38
rs773031898 Chr.22: 39374652-39374652 on Build GRCh38
rs759016697 Chr.22: 39374398-39374398 on Build GRCh38
rs779013987 Chr.22: 39374377-39374377 on Build GRCh38
rs769578394 Chr.22: 39374578-39374578 on Build GRCh38
rs746417387 Chr.22: 39374629-39374629 on Build GRCh38
rs138433500 Chr.22: 39374436-39374436 on Build GRCh38
rs941829896 Chr.22: 39374794-39374794 on Build GRCh38
rs1006074865 Chr.22: 39374370-39374370 on Build GRCh38
rs768902694 Chr.22: 39374382-39374382 on Build GRCh38
rs1017476658 Chr.22: 39374691-39374691 on Build GRCh38
rs760123756 Chr.22: 39374472-39374472 on Build GRCh38
rs4821888 Chr.22: 39374592-39374592 on Build GRCh38
rs554127367 Chr.22: 39374608-39374608 on Build GRCh38
rs765460227 Chr.22: 39374743-39374743 on Build GRCh38
rs905008425 Chr.22: 39374579-39374579 on Build GRCh38
rs570834171 Chr.22: 39374498-39374498 on Build GRCh38
rs925161799 Chr.22: 39374514-39374514 on Build GRCh38
rs755842526 Chr.22: 39374364-39374364 on Build GRCh38
rs144438003 Chr.22: 39374447-39374447 on Build GRCh38
rs761730237 Chr.22: 39374427-39374427 on Build GRCh38
rs897488632 Chr.22: 39374626-39374626 on Build GRCh38
rs368987356 Chr.22: 39374414-39374414 on Build GRCh38
rs752913466 Chr.22: 39374607-39374607 on Build GRCh38
rs373970139 Chr.22: 39374556-39374556 on Build GRCh38
rs993219223 Chr.22: 39374637-39374637 on Build GRCh38
rs776991777 Chr.22: 39374387-39374387 on Build GRCh38
rs150117922 Chr.22: 39374379-39374379 on Build GRCh38
rs1007297895 Chr.22: 39374669-39374669 on Build GRCh38
rs759459313 Chr.22: 39374671-39374671 on Build GRCh38
rs757202210 Chr.22: 39374376-39374376 on Build GRCh38
rs758823862 Chr.22: 39374751-39374751 on Build GRCh38
rs149257898 Chr.22: 39374441-39374441 on Build GRCh38

View Full Product Details