Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37516893-37517411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNRG
Entrez Gene ID
11276
Gene Name
synergin, gamma
Gene Aliases
AP1GBP1, SYNG
UniGene
Hs.594647
Gene Chromosome Location
Chr.17: 37514797-37613637 on Build GRCh38
Transcripts
NM_001163544.2 NM_001163545.2 NM_001163546.2 NM_001163547.2 NM_007247.5 NM_080550.4 NM_198882.2 XM_017024076.1 XM_017024077.1 XM_017024078.1 XM_017024079.1 XM_017024080.1 XM_017024081.1 XM_017024082.1 XM_017024083.1 XM_017024084.1 XM_017024085.1 XM_017024086.1 XM_017024087.1 XM_017024088.1 XM_017024089.1 XM_017024090.1 XM_017024091.1 XM_017024092.1 XM_017024093.1 XM_017024097.1 XM_017024099.1 XM_017024102.1 XM_017024106.1

SNP Detail

SNP ID Chromosome Location
rs570057579 Chr.17: 37517245-37517245 on Build GRCh38
rs527873041 Chr.17: 37517108-37517108 on Build GRCh38
rs937863721 Chr.17: 37516958-37516958 on Build GRCh38
rs917786909 Chr.17: 37517130-37517130 on Build GRCh38
rs926512914 Chr.17: 37516957-37516957 on Build GRCh38
rs535623005 Chr.17: 37517258-37517258 on Build GRCh38
rs1029156906 Chr.17: 37516953-37516953 on Build GRCh38
rs914861932 Chr.17: 37516995-37516995 on Build GRCh38
rs541608420 Chr.17: 37517157-37517157 on Build GRCh38
rs539228364 Chr.17: 37517122-37517122 on Build GRCh38
rs571173998 Chr.17: 37516969-37516969 on Build GRCh38
rs542695414 Chr.17: 37517083-37517083 on Build GRCh38
rs890274875 Chr.17: 37517356-37517356 on Build GRCh38
rs946705734 Chr.17: 37516930-37516930 on Build GRCh38
rs937894785 Chr.17: 37517113-37517113 on Build GRCh38
rs968497094 Chr.17: 37517243-37517243 on Build GRCh38
rs572894374 Chr.17: 37517139-37517139 on Build GRCh38
rs140460654 Chr.17: 37517244-37517244 on Build GRCh38
rs191872830 Chr.17: 37517183-37517183 on Build GRCh38
rs1036813871 Chr.17: 37517156-37517156 on Build GRCh38
rs968926965 Chr.17: 37517138-37517138 on Build GRCh38
rs1016159844 Chr.17: 37516923-37516923 on Build GRCh38
rs187299234 Chr.17: 37517087-37517087 on Build GRCh38
rs1057205737 Chr.17: 37517225-37517225 on Build GRCh38
rs536934681 Chr.17: 37517011-37517011 on Build GRCh38
rs931886570 Chr.17: 37517302-37517302 on Build GRCh38
rs111730407 Chr.17: 37516994-37516994 on Build GRCh38
rs556571531 Chr.17: 37516970-37516970 on Build GRCh38
rs907998827 Chr.17: 37517228-37517228 on Build GRCh38
rs551184787 Chr.17: 37516919-37516919 on Build GRCh38
rs572203094 Chr.17: 37517036-37517036 on Build GRCh38
rs902033921 Chr.17: 37517223-37517223 on Build GRCh38
rs1056185930 Chr.17: 37516980-37516980 on Build GRCh38
rs963167331 Chr.17: 37516929-37516929 on Build GRCh38
rs947535044 Chr.17: 37517000-37517000 on Build GRCh38
rs1133543 Chr.17: 37517003-37517003 on Build GRCh38
rs938751834 Chr.17: 37517019-37517019 on Build GRCh38
rs954446828 Chr.17: 37516965-37516965 on Build GRCh38
rs996057156 Chr.17: 37516946-37516946 on Build GRCh38
rs992028286 Chr.17: 37517124-37517124 on Build GRCh38
rs557614217 Chr.17: 37517042-37517042 on Build GRCh38
rs145885514 Chr.17: 37517287-37517287 on Build GRCh38
rs914915832 Chr.17: 37517128-37517128 on Build GRCh38
rs1007848980 Chr.17: 37517207-37517208 on Build GRCh38
rs1057467359 Chr.17: 37517040-37517040 on Build GRCh38
rs35102294 Chr.17: 37516969-37516970 on Build GRCh38
rs1030777846 Chr.17: 37517086-37517086 on Build GRCh38
rs927460862 Chr.17: 37517165-37517165 on Build GRCh38
rs754663391 Chr.17: 37517305-37517305 on Build GRCh38
rs931014871 Chr.17: 37517167-37517167 on Build GRCh38
rs577480036 Chr.17: 37517068-37517068 on Build GRCh38
rs765189279 Chr.17: 37516990-37516990 on Build GRCh38
rs12602776 Chr.17: 37517041-37517041 on Build GRCh38
rs1049386731 Chr.17: 37517174-37517174 on Build GRCh38
rs956345852 Chr.17: 37517106-37517106 on Build GRCh38
rs565769548 Chr.17: 37517372-37517372 on Build GRCh38
rs573556151 Chr.17: 37517073-37517073 on Build GRCh38
rs1040646641 Chr.17: 37517215-37517215 on Build GRCh38
rs142360557 Chr.17: 37517075-37517075 on Build GRCh38
rs988283415 Chr.17: 37517078-37517078 on Build GRCh38
rs151240905 Chr.17: 37517082-37517082 on Build GRCh38
rs564795254 Chr.17: 37517176-37517176 on Build GRCh38
rs996499011 Chr.17: 37517074-37517074 on Build GRCh38
rs1050788888 Chr.17: 37517338-37517338 on Build GRCh38
rs886455330 Chr.17: 37516918-37516918 on Build GRCh38
rs543035980 Chr.17: 37517069-37517069 on Build GRCh38
rs535902631 Chr.17: 37517009-37517009 on Build GRCh38

View Full Product Details