Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84821573-84822067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRABD2A
Entrez Gene ID
129293
Gene Name
TraB domain containing 2A
Gene Aliases
C2orf89, TIKI1
UniGene
Hs.469134
Gene Chromosome Location
Chr.2: 84821650-84881975 on Build GRCh38
Transcripts
NM_001080824.2 NM_001277053.1 XM_011532504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159042 c.1247_1248GG>AT p.R416H Chr.2: 84822040-84822041 on Build GRCh38
3583696 c.1437C>T p.A479A Chr.2: 84821998-84821998 on Build GRCh38
4616003 c.1510delC p.L504fs*>2 Chr.2: 84821925-84821925 on Build GRCh38
219613 c.1251C>T p.L417L Chr.2: 84822037-84822037 on Build GRCh38
4644463 c.1411C>T p.H471Y Chr.2: 84822024-84822024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746036602 Chr.2: 84821967-84821967 on Build GRCh38
rs893550338 Chr.2: 84821637-84821637 on Build GRCh38
rs926454222 Chr.2: 84821951-84821951 on Build GRCh38
rs905317259 Chr.2: 84821655-84821655 on Build GRCh38
rs374787167 Chr.2: 84821892-84821892 on Build GRCh38
rs59922171 Chr.2: 84821632-84821632 on Build GRCh38
rs750468538 Chr.2: 84821803-84821803 on Build GRCh38
rs751587281 Chr.2: 84821952-84821952 on Build GRCh38
rs777472069 Chr.2: 84821916-84821916 on Build GRCh38
rs987699607 Chr.2: 84821731-84821731 on Build GRCh38
rs534732083 Chr.2: 84821619-84821619 on Build GRCh38
rs756446721 Chr.2: 84821979-84821979 on Build GRCh38
rs576224269 Chr.2: 84821714-84821714 on Build GRCh38
rs765273481 Chr.2: 84821927-84821927 on Build GRCh38
rs369918624 Chr.2: 84821923-84821923 on Build GRCh38
rs13000269 Chr.2: 84821717-84821717 on Build GRCh38
rs13024201 Chr.2: 84821811-84821811 on Build GRCh38
rs955134902 Chr.2: 84821657-84821657 on Build GRCh38
rs188546476 Chr.2: 84821858-84821858 on Build GRCh38
rs199868738 Chr.2: 84822029-84822029 on Build GRCh38
rs781542956 Chr.2: 84821871-84821871 on Build GRCh38
rs13000267 Chr.2: 84821716-84821716 on Build GRCh38
rs559465733 Chr.2: 84822041-84822041 on Build GRCh38
rs962217409 Chr.2: 84821741-84821741 on Build GRCh38
rs770224103 Chr.2: 84821875-84821875 on Build GRCh38
rs1046668707 Chr.2: 84821604-84821604 on Build GRCh38
rs949268160 Chr.2: 84821627-84821627 on Build GRCh38
rs13023615 Chr.2: 84821674-84821674 on Build GRCh38
rs781144232 Chr.2: 84821964-84821964 on Build GRCh38
rs991911653 Chr.2: 84821859-84821859 on Build GRCh38
rs371054600 Chr.2: 84822020-84822020 on Build GRCh38
rs747043546 Chr.2: 84821603-84821605 on Build GRCh38
rs754883629 Chr.2: 84821924-84821924 on Build GRCh38
rs369279325 Chr.2: 84821882-84821882 on Build GRCh38
rs973528167 Chr.2: 84821789-84821789 on Build GRCh38
rs1028636265 Chr.2: 84821655-84821655 on Build GRCh38
rs374350995 Chr.2: 84821990-84821990 on Build GRCh38
rs1044911897 Chr.2: 84821927-84821928 on Build GRCh38
rs543721894 Chr.2: 84821740-84821740 on Build GRCh38
rs770565102 Chr.2: 84822033-84822033 on Build GRCh38
rs772631869 Chr.2: 84822032-84822032 on Build GRCh38
rs1053601173 Chr.2: 84821935-84821935 on Build GRCh38
rs778838959 Chr.2: 84821796-84821796 on Build GRCh38
rs908928567 Chr.2: 84821911-84821912 on Build GRCh38
rs749729117 Chr.2: 84821999-84821999 on Build GRCh38
rs766066623 Chr.2: 84822039-84822039 on Build GRCh38
rs377717113 Chr.2: 84822009-84822009 on Build GRCh38
rs757294817 Chr.2: 84821953-84821953 on Build GRCh38
rs892519800 Chr.2: 84822015-84822015 on Build GRCh38
rs568594014 Chr.2: 84821676-84821676 on Build GRCh38
rs773762140 Chr.2: 84822042-84822042 on Build GRCh38
rs373182119 Chr.2: 84821887-84821887 on Build GRCh38
rs369421131 Chr.2: 84821622-84821623 on Build GRCh38
rs1019877297 Chr.2: 84821823-84821823 on Build GRCh38
rs539393364 Chr.2: 84821620-84821620 on Build GRCh38
rs746091619 Chr.2: 84821873-84821873 on Build GRCh38
rs1053804610 Chr.2: 84822007-84822007 on Build GRCh38
rs76277344 Chr.2: 84822030-84822030 on Build GRCh38
rs746331257 Chr.2: 84821920-84821920 on Build GRCh38
rs892467384 Chr.2: 84821739-84821739 on Build GRCh38
rs200837426 Chr.2: 84821881-84821881 on Build GRCh38
rs766944169 Chr.2: 84821915-84821915 on Build GRCh38
rs761457432 Chr.2: 84821906-84821906 on Build GRCh38
rs996269928 Chr.2: 84821651-84821651 on Build GRCh38

View Full Product Details