Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44489860-44490375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPAL
Entrez Gene ID
79183
Gene Name
alpha tocopherol transfer protein like
Gene Aliases
C20orf121
UniGene
Hs.283869
Gene Chromosome Location
Chr.20: 44475886-44494603 on Build GRCh38
Transcripts
NM_001039199.2 NM_001261839.1 NM_024331.4 XM_011529043.2 XM_011529044.2 XM_011529045.2 XM_017028063.1

SNP Detail

SNP ID Chromosome Location
rs931137537 Chr.20: 44490325-44490325 on Build GRCh38
rs760558373 Chr.20: 44490308-44490308 on Build GRCh38
rs966097676 Chr.20: 44490071-44490071 on Build GRCh38
rs73113314 Chr.20: 44489965-44489965 on Build GRCh38
rs953546256 Chr.20: 44490240-44490240 on Build GRCh38
rs1048113653 Chr.20: 44490125-44490125 on Build GRCh38
rs551623271 Chr.20: 44489946-44489946 on Build GRCh38
rs554044429 Chr.20: 44490213-44490214 on Build GRCh38
rs770534044 Chr.20: 44490084-44490084 on Build GRCh38
rs6093984 Chr.20: 44489920-44489920 on Build GRCh38
rs925374012 Chr.20: 44489953-44489953 on Build GRCh38
rs957011501 Chr.20: 44490263-44490263 on Build GRCh38
rs538621017 Chr.20: 44490247-44490247 on Build GRCh38
rs988548145 Chr.20: 44489991-44489991 on Build GRCh38
rs1027290377 Chr.20: 44490294-44490294 on Build GRCh38
rs946177661 Chr.20: 44490194-44490194 on Build GRCh38
rs796462674 Chr.20: 44490242-44490242 on Build GRCh38
rs917797027 Chr.20: 44489997-44489997 on Build GRCh38
rs781565657 Chr.20: 44489928-44489928 on Build GRCh38
rs970599386 Chr.20: 44490288-44490288 on Build GRCh38
rs1004558433 Chr.20: 44490352-44490352 on Build GRCh38
rs1048705096 Chr.20: 44490337-44490337 on Build GRCh38
rs773302663 Chr.20: 44490051-44490051 on Build GRCh38
rs773895603 Chr.20: 44490315-44490315 on Build GRCh38
rs12625842 Chr.20: 44490075-44490075 on Build GRCh38
rs979572112 Chr.20: 44489921-44489921 on Build GRCh38
rs527732592 Chr.20: 44490016-44490016 on Build GRCh38
rs894107747 Chr.20: 44490258-44490258 on Build GRCh38
rs988091662 Chr.20: 44490268-44490268 on Build GRCh38
rs866037363 Chr.20: 44490158-44490158 on Build GRCh38
rs938426786 Chr.20: 44490246-44490246 on Build GRCh38
rs558734691 Chr.20: 44490279-44490279 on Build GRCh38
rs926480525 Chr.20: 44490083-44490083 on Build GRCh38
rs867712488 Chr.20: 44489944-44489944 on Build GRCh38
rs931043375 Chr.20: 44490111-44490111 on Build GRCh38
rs976202002 Chr.20: 44490079-44490079 on Build GRCh38
rs912430871 Chr.20: 44490026-44490026 on Build GRCh38
rs577842592 Chr.20: 44490082-44490082 on Build GRCh38
rs545241023 Chr.20: 44490136-44490136 on Build GRCh38
rs958400686 Chr.20: 44489923-44489923 on Build GRCh38
rs375968871 Chr.20: 44490200-44490200 on Build GRCh38
rs995204010 Chr.20: 44490292-44490292 on Build GRCh38
rs1033974121 Chr.20: 44489891-44489891 on Build GRCh38
rs908278734 Chr.20: 44490348-44490348 on Build GRCh38
rs980683503 Chr.20: 44490291-44490291 on Build GRCh38
rs949335936 Chr.20: 44490042-44490042 on Build GRCh38
rs569587090 Chr.20: 44490231-44490231 on Build GRCh38
rs550029372 Chr.20: 44490208-44490208 on Build GRCh38
rs1026908591 Chr.20: 44489974-44489974 on Build GRCh38
rs567089391 Chr.20: 44490126-44490126 on Build GRCh38
rs917954072 Chr.20: 44490286-44490286 on Build GRCh38
rs982652274 Chr.20: 44490023-44490023 on Build GRCh38
rs144517689 Chr.20: 44490159-44490159 on Build GRCh38
rs112783804 Chr.20: 44490186-44490186 on Build GRCh38
rs368926470 Chr.20: 44490323-44490323 on Build GRCh38

View Full Product Details