Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 31878140-31878648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP6
Entrez Gene ID
55813
Gene Name
UTP6, small subunit processome component
Gene Aliases
C17orf40, HCA66
UniGene
Hs.589862
Gene Chromosome Location
Chr.17: 31862379-31901710 on Build GRCh38
Transcripts
NM_018428.2 XM_005258012.2 XM_011524997.2 XM_017024845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3180152 c.1095G>T p.K365N Chr.17: 31878280-31878280 on Build GRCh38
4065378 c.1087G>C p.E363Q Chr.17: 31878288-31878288 on Build GRCh38
5958776 c.1064delT p.M355fs*10 Chr.17: 31878311-31878311 on Build GRCh38
293104 c.1103C>T p.S368L Chr.17: 31878272-31878272 on Build GRCh38
1302622 c.1099C>G p.L367V Chr.17: 31878276-31878276 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754519191 Chr.17: 31878259-31878259 on Build GRCh38
rs960721659 Chr.17: 31878183-31878184 on Build GRCh38
rs367888410 Chr.17: 31878229-31878229 on Build GRCh38
rs144113207 Chr.17: 31878411-31878411 on Build GRCh38
rs35290456 Chr.17: 31878491-31878492 on Build GRCh38
rs888839639 Chr.17: 31878435-31878435 on Build GRCh38
rs892398222 Chr.17: 31878487-31878487 on Build GRCh38
rs376977101 Chr.17: 31878262-31878262 on Build GRCh38
rs947644982 Chr.17: 31878496-31878496 on Build GRCh38
rs752381434 Chr.17: 31878274-31878274 on Build GRCh38
rs561636597 Chr.17: 31878564-31878564 on Build GRCh38
rs968281645 Chr.17: 31878169-31878169 on Build GRCh38
rs779569120 Chr.17: 31878226-31878227 on Build GRCh38
rs940761515 Chr.17: 31878597-31878597 on Build GRCh38
rs775493371 Chr.17: 31878332-31878332 on Build GRCh38
rs979392583 Chr.17: 31878531-31878531 on Build GRCh38
rs781737053 Chr.17: 31878302-31878302 on Build GRCh38
rs1034160066 Chr.17: 31878180-31878180 on Build GRCh38
rs8080862 Chr.17: 31878439-31878439 on Build GRCh38
rs1033780253 Chr.17: 31878567-31878567 on Build GRCh38
rs539191760 Chr.17: 31878576-31878579 on Build GRCh38
rs551036116 Chr.17: 31878491-31878491 on Build GRCh38
rs1022633495 Chr.17: 31878510-31878510 on Build GRCh38
rs974653921 Chr.17: 31878301-31878301 on Build GRCh38
rs778247568 Chr.17: 31878292-31878292 on Build GRCh38
rs985232115 Chr.17: 31878385-31878385 on Build GRCh38
rs72821922 Chr.17: 31878584-31878584 on Build GRCh38
rs1026665917 Chr.17: 31878330-31878330 on Build GRCh38
rs564942734 Chr.17: 31878391-31878391 on Build GRCh38
rs766265718 Chr.17: 31878367-31878367 on Build GRCh38
rs546916816 Chr.17: 31878578-31878578 on Build GRCh38
rs995816638 Chr.17: 31878289-31878289 on Build GRCh38
rs760575975 Chr.17: 31878250-31878250 on Build GRCh38
rs974390667 Chr.17: 31878618-31878618 on Build GRCh38
rs903710221 Chr.17: 31878539-31878539 on Build GRCh38
rs953819725 Chr.17: 31878378-31878378 on Build GRCh38
rs753239778 Chr.17: 31878253-31878253 on Build GRCh38
rs767290263 Chr.17: 31878241-31878241 on Build GRCh38
rs1057224762 Chr.17: 31878589-31878589 on Build GRCh38
rs908815645 Chr.17: 31878233-31878233 on Build GRCh38
rs148454135 Chr.17: 31878206-31878206 on Build GRCh38
rs1002431635 Chr.17: 31878566-31878566 on Build GRCh38
rs772969811 Chr.17: 31878353-31878353 on Build GRCh38
rs368035634 Chr.17: 31878196-31878196 on Build GRCh38
rs567680923 Chr.17: 31878529-31878529 on Build GRCh38
rs768906041 Chr.17: 31878348-31878348 on Build GRCh38
rs750286373 Chr.17: 31878245-31878245 on Build GRCh38
rs767022331 Chr.17: 31878461-31878461 on Build GRCh38
rs769933589 Chr.17: 31878317-31878317 on Build GRCh38
rs961350235 Chr.17: 31878599-31878599 on Build GRCh38
rs759449083 Chr.17: 31878374-31878374 on Build GRCh38
rs1050215320 Chr.17: 31878407-31878407 on Build GRCh38
rs745871937 Chr.17: 31878306-31878306 on Build GRCh38
rs758191630 Chr.17: 31878290-31878290 on Build GRCh38
rs539342724 Chr.17: 31878593-31878593 on Build GRCh38
rs542486674 Chr.17: 31878189-31878189 on Build GRCh38
rs749524774 Chr.17: 31878342-31878342 on Build GRCh38
rs1040024812 Chr.17: 31878443-31878443 on Build GRCh38
rs188945229 Chr.17: 31878300-31878300 on Build GRCh38
rs530311091 Chr.17: 31878543-31878543 on Build GRCh38
rs746376644 Chr.17: 31878337-31878337 on Build GRCh38
rs760486303 Chr.17: 31878364-31878364 on Build GRCh38
rs774333923 Chr.17: 31878172-31878172 on Build GRCh38
rs776643147 Chr.17: 31878373-31878373 on Build GRCh38
rs747423519 Chr.17: 31878298-31878298 on Build GRCh38

View Full Product Details