Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2550056-2550588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC115110
Entrez Gene ID
115110
Gene Name
uncharacterized LOC115110
Gene Aliases
-
UniGene
Hs.132272
Gene Chromosome Location
Chr.1: 2549920-2557011 on Build GRCh38
Transcripts
NR_037844.2

SNP Detail

SNP ID Chromosome Location
rs1035172893 Chr.1: 2550209-2550209 on Build GRCh38
rs1010798551 Chr.1: 2550177-2550177 on Build GRCh38
rs561805513 Chr.1: 2550408-2550408 on Build GRCh38
rs149465867 Chr.1: 2550318-2550318 on Build GRCh38
rs541718362 Chr.1: 2550108-2550108 on Build GRCh38
rs563990944 Chr.1: 2550172-2550172 on Build GRCh38
rs892345267 Chr.1: 2550176-2550176 on Build GRCh38
rs982403482 Chr.1: 2550228-2550228 on Build GRCh38
rs988173610 Chr.1: 2550544-2550544 on Build GRCh38
rs1015380045 Chr.1: 2550439-2550439 on Build GRCh38
rs946750345 Chr.1: 2550383-2550383 on Build GRCh38
rs527447242 Chr.1: 2550145-2550145 on Build GRCh38
rs1055502 Chr.1: 2550227-2550227 on Build GRCh38
rs768798615 Chr.1: 2550431-2550431 on Build GRCh38
rs911932444 Chr.1: 2550292-2550292 on Build GRCh38
rs535481159 Chr.1: 2550241-2550241 on Build GRCh38
rs1003977251 Chr.1: 2550430-2550430 on Build GRCh38
rs905067921 Chr.1: 2550558-2550558 on Build GRCh38
rs937959491 Chr.1: 2550562-2550562 on Build GRCh38
rs537236676 Chr.1: 2550124-2550124 on Build GRCh38
rs1043607769 Chr.1: 2550386-2550386 on Build GRCh38
rs896198547 Chr.1: 2550411-2550411 on Build GRCh38
rs953662895 Chr.1: 2550510-2550510 on Build GRCh38
rs375079342 Chr.1: 2550543-2550543 on Build GRCh38
rs973774037 Chr.1: 2550480-2550480 on Build GRCh38
rs537126768 Chr.1: 2550302-2550302 on Build GRCh38
rs56371836 Chr.1: 2550528-2550528 on Build GRCh38
rs183059264 Chr.1: 2550204-2550204 on Build GRCh38
rs566367531 Chr.1: 2550250-2550250 on Build GRCh38
rs115511887 Chr.1: 2550263-2550263 on Build GRCh38
rs868703707 Chr.1: 2550173-2550173 on Build GRCh38
rs2259628 Chr.1: 2550079-2550079 on Build GRCh38
rs572320475 Chr.1: 2550475-2550475 on Build GRCh38
rs186998367 Chr.1: 2550210-2550210 on Build GRCh38
rs890584549 Chr.1: 2550142-2550142 on Build GRCh38
rs777048928 Chr.1: 2550456-2550456 on Build GRCh38
rs578135798 Chr.1: 2550295-2550295 on Build GRCh38
rs892481612 Chr.1: 2550353-2550353 on Build GRCh38
rs202076150 Chr.1: 2550388-2550388 on Build GRCh38
rs969616373 Chr.1: 2550189-2550189 on Build GRCh38
rs1043738740 Chr.1: 2550554-2550554 on Build GRCh38
rs986666608 Chr.1: 2550521-2550521 on Build GRCh38
rs541001214 Chr.1: 2550491-2550491 on Build GRCh38
rs780128607 Chr.1: 2550237-2550237 on Build GRCh38
rs371156050 Chr.1: 2550131-2550131 on Build GRCh38
rs367554433 Chr.1: 2550354-2550354 on Build GRCh38
rs1022057469 Chr.1: 2550178-2550178 on Build GRCh38
rs542614825 Chr.1: 2550389-2550389 on Build GRCh38
rs375553053 Chr.1: 2550273-2550273 on Build GRCh38
rs569664299 Chr.1: 2550217-2550217 on Build GRCh38
rs549660111 Chr.1: 2550249-2550249 on Build GRCh38
rs946719210 Chr.1: 2550547-2550547 on Build GRCh38
rs973638709 Chr.1: 2550262-2550262 on Build GRCh38
rs747232222 Chr.1: 2550408-2550408 on Build GRCh38
rs547590823 Chr.1: 2550151-2550151 on Build GRCh38

View Full Product Details