Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28579976-28580520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG12
Entrez Gene ID
85028
Gene Name
small nucleolar RNA host gene 12
Gene Aliases
ASLNC04080, C1orf79, LINC00100, NCRNA00100
UniGene
Hs.632377
Gene Chromosome Location
Chr.1: 28578538-28581854 on Build GRCh38
Transcripts
NR_024127.1 NR_002976.1

Gene Symbol
SNORA44
Entrez Gene ID
677825
Gene Name
small nucleolar RNA, H/ACA box 44
Gene Aliases
ACA44
UniGene
Hs.632377
Gene Chromosome Location
Chr.1: 28580381-28580512 on Build GRCh38
Transcripts
NR_024127.1 NR_002976.1

SNP Detail

SNP ID Chromosome Location
rs184893492 Chr.1: 28580087-28580087 on Build GRCh38
rs192269532 Chr.1: 28580133-28580133 on Build GRCh38
rs1034008999 Chr.1: 28580191-28580191 on Build GRCh38
rs144823378 Chr.1: 28580195-28580195 on Build GRCh38
rs936979757 Chr.1: 28580065-28580065 on Build GRCh38
rs536418574 Chr.1: 28580157-28580157 on Build GRCh38
rs768667774 Chr.1: 28580352-28580352 on Build GRCh38
rs867604349 Chr.1: 28580318-28580318 on Build GRCh38
rs765780432 Chr.1: 28580425-28580425 on Build GRCh38
rs886181099 Chr.1: 28580254-28580254 on Build GRCh38
rs775302441 Chr.1: 28580461-28580461 on Build GRCh38
rs772103192 Chr.1: 28580180-28580180 on Build GRCh38
rs745771322 Chr.1: 28580459-28580459 on Build GRCh38
rs773176446 Chr.1: 28580182-28580182 on Build GRCh38
rs759386495 Chr.1: 28580388-28580388 on Build GRCh38
rs978207471 Chr.1: 28580328-28580328 on Build GRCh38
rs77462506 Chr.1: 28580228-28580228 on Build GRCh38
rs372198770 Chr.1: 28580334-28580334 on Build GRCh38
rs188492028 Chr.1: 28580091-28580091 on Build GRCh38
rs747126800 Chr.1: 28580435-28580436 on Build GRCh38
rs571559813 Chr.1: 28580088-28580088 on Build GRCh38
rs371155351 Chr.1: 28580422-28580422 on Build GRCh38
rs397936635 Chr.1: 28580328-28580329 on Build GRCh38
rs949649937 Chr.1: 28580408-28580408 on Build GRCh38
rs372375354 Chr.1: 28580489-28580489 on Build GRCh38
rs1020114942 Chr.1: 28580319-28580319 on Build GRCh38
rs562742423 Chr.1: 28580456-28580456 on Build GRCh38
rs558263617 Chr.1: 28580197-28580197 on Build GRCh38
rs912539882 Chr.1: 28580022-28580022 on Build GRCh38
rs192623464 Chr.1: 28580026-28580026 on Build GRCh38
rs575128183 Chr.1: 28580209-28580209 on Build GRCh38
rs763535492 Chr.1: 28580413-28580413 on Build GRCh38
rs770791121 Chr.1: 28580373-28580373 on Build GRCh38
rs16837624 Chr.1: 28580472-28580472 on Build GRCh38
rs987289344 Chr.1: 28580171-28580171 on Build GRCh38
rs991249976 Chr.1: 28580357-28580357 on Build GRCh38
rs372383885 Chr.1: 28580312-28580312 on Build GRCh38
rs747224286 Chr.1: 28580351-28580351 on Build GRCh38
rs750963201 Chr.1: 28580426-28580426 on Build GRCh38
rs147786269 Chr.1: 28580307-28580307 on Build GRCh38
rs780210003 Chr.1: 28580349-28580349 on Build GRCh38
rs572835366 Chr.1: 28580175-28580175 on Build GRCh38
rs760275297 Chr.1: 28580465-28580465 on Build GRCh38
rs772909837 Chr.1: 28580120-28580128 on Build GRCh38
rs942057271 Chr.1: 28580321-28580321 on Build GRCh38
rs755192535 Chr.1: 28580433-28580433 on Build GRCh38
rs966727551 Chr.1: 28580322-28580322 on Build GRCh38
rs910548552 Chr.1: 28580321-28580322 on Build GRCh38
rs540407020 Chr.1: 28580302-28580302 on Build GRCh38
rs776723771 Chr.1: 28580359-28580359 on Build GRCh38
rs866442067 Chr.1: 28580070-28580070 on Build GRCh38
rs911330442 Chr.1: 28580337-28580337 on Build GRCh38
rs1050387414 Chr.1: 28580079-28580079 on Build GRCh38
rs758507054 Chr.1: 28580382-28580384 on Build GRCh38
rs560671810 Chr.1: 28580389-28580389 on Build GRCh38
rs936883681 Chr.1: 28580198-28580198 on Build GRCh38
rs781140216 Chr.1: 28580452-28580452 on Build GRCh38
rs1028848558 Chr.1: 28580297-28580297 on Build GRCh38
rs41307828 Chr.1: 28580428-28580428 on Build GRCh38
rs1015963971 Chr.1: 28580109-28580109 on Build GRCh38
rs557246800 Chr.1: 28580110-28580110 on Build GRCh38
rs544510374 Chr.1: 28580220-28580220 on Build GRCh38
rs1034292253 Chr.1: 28580403-28580403 on Build GRCh38
rs899074159 Chr.1: 28580120-28580120 on Build GRCh38
rs199819282 Chr.1: 28580487-28580487 on Build GRCh38
rs780313741 Chr.1: 28580427-28580428 on Build GRCh38
rs1029215749 Chr.1: 28580165-28580165 on Build GRCh38
rs749258893 Chr.1: 28580363-28580363 on Build GRCh38
rs1004941454 Chr.1: 28580102-28580102 on Build GRCh38
rs993978370 Chr.1: 28580330-28580330 on Build GRCh38
rs923508345 Chr.1: 28580308-28580314 on Build GRCh38
rs1037740242 Chr.1: 28580285-28580285 on Build GRCh38
rs907071520 Chr.1: 28580386-28580386 on Build GRCh38
rs898845435 Chr.1: 28580286-28580286 on Build GRCh38
rs996117490 Chr.1: 28580132-28580132 on Build GRCh38
rs1046629016 Chr.1: 28580253-28580253 on Build GRCh38
rs530621258 Chr.1: 28580454-28580454 on Build GRCh38
rs775042761 Chr.1: 28580411-28580411 on Build GRCh38
rs933299502 Chr.1: 28580075-28580075 on Build GRCh38
rs760349496 Chr.1: 28580412-28580412 on Build GRCh38
rs886212450 Chr.1: 28580101-28580101 on Build GRCh38
rs1037775392 Chr.1: 28580326-28580326 on Build GRCh38
rs1050667979 Chr.1: 28580320-28580320 on Build GRCh38
rs954563574 Chr.1: 28580168-28580168 on Build GRCh38
rs774148991 Chr.1: 28580387-28580387 on Build GRCh38
rs1032726779 Chr.1: 28580329-28580329 on Build GRCh38
rs925521055 Chr.1: 28580046-28580046 on Build GRCh38
rs748170696 Chr.1: 28580453-28580453 on Build GRCh38
rs141143333 Chr.1: 28580256-28580256 on Build GRCh38
rs958400400 Chr.1: 28580192-28580192 on Build GRCh38
rs954095869 Chr.1: 28580011-28580011 on Build GRCh38
rs751753027 Chr.1: 28580429-28580429 on Build GRCh38
rs753348390 Chr.1: 28580414-28580414 on Build GRCh38
rs532341748 Chr.1: 28580019-28580019 on Build GRCh38
rs1042422326 Chr.1: 28580027-28580027 on Build GRCh38
rs758875944 Chr.1: 28580427-28580427 on Build GRCh38
rs996472948 Chr.1: 28580289-28580289 on Build GRCh38
rs149431027 Chr.1: 28580172-28580172 on Build GRCh38

View Full Product Details