Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212857319-212857815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR1-AS1
Entrez Gene ID
642946
Gene Name
FLVCR1 antisense RNA 1 (head to head)
Gene Aliases
LQK1, NCRNA00292
UniGene
Hs.552649
Gene Chromosome Location
Chr.1: 212856604-212858138 on Build GRCh38
Transcripts
NR_027285.1 NR_027286.1

SNP Detail

SNP ID Chromosome Location
rs41296676 Chr.1: 212857625-212857625 on Build GRCh38
rs1008165433 Chr.1: 212857394-212857394 on Build GRCh38
rs924281479 Chr.1: 212857661-212857661 on Build GRCh38
rs543160457 Chr.1: 212857461-212857461 on Build GRCh38
rs997691020 Chr.1: 212857714-212857714 on Build GRCh38
rs75028651 Chr.1: 212857384-212857384 on Build GRCh38
rs889050688 Chr.1: 212857681-212857681 on Build GRCh38
rs754516236 Chr.1: 212857544-212857546 on Build GRCh38
rs752519628 Chr.1: 212857369-212857369 on Build GRCh38
rs369205441 Chr.1: 212857698-212857699 on Build GRCh38
rs1012017976 Chr.1: 212857790-212857790 on Build GRCh38
rs980311368 Chr.1: 212857535-212857535 on Build GRCh38
rs41296674 Chr.1: 212857616-212857616 on Build GRCh38
rs546074161 Chr.1: 212857693-212857693 on Build GRCh38
rs888935116 Chr.1: 212857480-212857480 on Build GRCh38
rs1031920405 Chr.1: 212857728-212857728 on Build GRCh38
rs796247634 Chr.1: 212857674-212857674 on Build GRCh38
rs1017799073 Chr.1: 212857398-212857398 on Build GRCh38
rs41296672 Chr.1: 212857353-212857353 on Build GRCh38
rs969901908 Chr.1: 212857532-212857532 on Build GRCh38
rs41296678 Chr.1: 212857639-212857639 on Build GRCh38
rs1041389594 Chr.1: 212857366-212857367 on Build GRCh38
rs764442789 Chr.1: 212857543-212857544 on Build GRCh38
rs76720745 Chr.1: 212857381-212857381 on Build GRCh38
rs71147040 Chr.1: 212857567-212857567 on Build GRCh38
rs1051655068 Chr.1: 212857455-212857455 on Build GRCh38
rs41296680 Chr.1: 212857640-212857640 on Build GRCh38
rs754833101 Chr.1: 212857786-212857786 on Build GRCh38
rs981674269 Chr.1: 212857523-212857523 on Build GRCh38
rs140978949 Chr.1: 212857793-212857793 on Build GRCh38
rs933016523 Chr.1: 212857664-212857664 on Build GRCh38
rs902024933 Chr.1: 212857689-212857689 on Build GRCh38
rs899028449 Chr.1: 212857711-212857711 on Build GRCh38
rs938540338 Chr.1: 212857607-212857607 on Build GRCh38
rs889562518 Chr.1: 212857673-212857673 on Build GRCh38
rs956049201 Chr.1: 212857475-212857475 on Build GRCh38
rs11803558 Chr.1: 212857343-212857343 on Build GRCh38
rs995880386 Chr.1: 212857516-212857516 on Build GRCh38
rs561119655 Chr.1: 212857772-212857772 on Build GRCh38
rs977613039 Chr.1: 212857644-212857644 on Build GRCh38
rs562723278 Chr.1: 212857472-212857472 on Build GRCh38
rs1039157158 Chr.1: 212857679-212857679 on Build GRCh38
rs1031713297 Chr.1: 212857670-212857678 on Build GRCh38
rs943167779 Chr.1: 212857681-212857681 on Build GRCh38
rs1018751728 Chr.1: 212857498-212857498 on Build GRCh38
rs574969976 Chr.1: 212857518-212857518 on Build GRCh38
rs997744712 Chr.1: 212857457-212857457 on Build GRCh38
rs898582619 Chr.1: 212857510-212857510 on Build GRCh38
rs942478757 Chr.1: 212857677-212857677 on Build GRCh38
rs928471311 Chr.1: 212857560-212857560 on Build GRCh38
rs766038448 Chr.1: 212857688-212857688 on Build GRCh38
rs990246382 Chr.1: 212857486-212857486 on Build GRCh38
rs951503172 Chr.1: 212857519-212857519 on Build GRCh38
rs902077464 Chr.1: 212857447-212857447 on Build GRCh38
rs775617837 Chr.1: 212857545-212857546 on Build GRCh38
rs751513759 Chr.1: 212857723-212857723 on Build GRCh38
rs1035976609 Chr.1: 212857549-212857549 on Build GRCh38

View Full Product Details