Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 650972-651516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
UniGene
Hs.432397
Gene Chromosome Location
Chr.10: 274190-689710 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_011519432.1 NR_027151.1 NR_027152.2

Gene Symbol
PRR26
Entrez Gene ID
414235
Gene Name
proline rich 26
Gene Aliases
C10orf108
UniGene
Hs.587103
Gene Chromosome Location
Chr.10: 649948-665169 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_011519432.1 NR_027151.1 NR_027152.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807496 c.663A>G p.Q221Q Chr.10: 651230-651230 on Build GRCh38
3978717 c.595C>A p.L199M Chr.10: 651162-651162 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1031114105 Chr.10: 651023-651023 on Build GRCh38
rs969003965 Chr.10: 651418-651418 on Build GRCh38
rs1000967076 Chr.10: 651022-651022 on Build GRCh38
rs767316167 Chr.10: 651238-651238 on Build GRCh38
rs199575227 Chr.10: 651352-651352 on Build GRCh38
rs571546737 Chr.10: 651407-651407 on Build GRCh38
rs533744356 Chr.10: 651275-651275 on Build GRCh38
rs1034253993 Chr.10: 651342-651342 on Build GRCh38
rs941739051 Chr.10: 651326-651326 on Build GRCh38
rs1035483894 Chr.10: 651162-651162 on Build GRCh38
rs927142678 Chr.10: 651080-651080 on Build GRCh38
rs767066813 Chr.10: 651365-651365 on Build GRCh38
rs747054036 Chr.10: 651300-651300 on Build GRCh38
rs903907827 Chr.10: 651008-651008 on Build GRCh38
rs766526434 Chr.10: 651390-651391 on Build GRCh38
rs576709184 Chr.10: 651150-651150 on Build GRCh38
rs116202578 Chr.10: 651255-651255 on Build GRCh38
rs755586494 Chr.10: 651420-651420 on Build GRCh38
rs1001262700 Chr.10: 651360-651360 on Build GRCh38
rs751372956 Chr.10: 651353-651353 on Build GRCh38
rs1047862213 Chr.10: 651010-651010 on Build GRCh38
rs892517474 Chr.10: 651485-651485 on Build GRCh38
rs960248700 Chr.10: 651171-651171 on Build GRCh38
rs762619179 Chr.10: 651203-651203 on Build GRCh38
rs763683653 Chr.10: 651205-651205 on Build GRCh38
rs1016060696 Chr.10: 651044-651044 on Build GRCh38
rs531429927 Chr.10: 651245-651245 on Build GRCh38
rs771245430 Chr.10: 651126-651126 on Build GRCh38
rs773771522 Chr.10: 651061-651061 on Build GRCh38
rs559987801 Chr.10: 650995-650995 on Build GRCh38
rs779500663 Chr.10: 651421-651421 on Build GRCh38
rs778150029 Chr.10: 651474-651474 on Build GRCh38
rs983352326 Chr.10: 651491-651491 on Build GRCh38
rs868472831 Chr.10: 651067-651067 on Build GRCh38
rs771493167 Chr.10: 651310-651310 on Build GRCh38
rs757188776 Chr.10: 651237-651237 on Build GRCh38
rs911312903 Chr.10: 651246-651246 on Build GRCh38
rs775219395 Chr.10: 651192-651192 on Build GRCh38
rs1039964069 Chr.10: 651434-651434 on Build GRCh38
rs941092129 Chr.10: 651085-651085 on Build GRCh38
rs555531368 Chr.10: 651464-651464 on Build GRCh38
rs145650735 Chr.10: 651457-651457 on Build GRCh38
rs576549722 Chr.10: 651182-651182 on Build GRCh38
rs1018203529 Chr.10: 651424-651424 on Build GRCh38
rs922743953 Chr.10: 651281-651281 on Build GRCh38
rs962509227 Chr.10: 651349-651349 on Build GRCh38
rs568983199 Chr.10: 651156-651156 on Build GRCh38
rs571144152 Chr.10: 651361-651361 on Build GRCh38
rs1006282209 Chr.10: 651041-651041 on Build GRCh38
rs763301096 Chr.10: 651467-651467 on Build GRCh38
rs375895503 Chr.10: 651358-651358 on Build GRCh38
rs992603816 Chr.10: 651037-651037 on Build GRCh38
rs368947867 Chr.10: 650996-650996 on Build GRCh38
rs747499850 Chr.10: 651116-651116 on Build GRCh38
rs971726194 Chr.10: 651050-651050 on Build GRCh38
rs181315415 Chr.10: 651366-651366 on Build GRCh38
rs751598352 Chr.10: 651234-651234 on Build GRCh38
rs942403328 Chr.10: 651425-651425 on Build GRCh38
rs973776584 Chr.10: 651087-651087 on Build GRCh38
rs748677494 Chr.10: 651436-651436 on Build GRCh38
rs750252842 Chr.10: 651247-651247 on Build GRCh38
rs150712991 Chr.10: 651266-651266 on Build GRCh38
rs918368390 Chr.10: 651127-651127 on Build GRCh38
rs764960418 Chr.10: 651183-651183 on Build GRCh38
rs1010470118 Chr.10: 651379-651379 on Build GRCh38
rs1013551168 Chr.10: 651280-651280 on Build GRCh38
rs556715471 Chr.10: 651367-651367 on Build GRCh38
rs964116469 Chr.10: 651448-651448 on Build GRCh38
rs918296242 Chr.10: 651362-651362 on Build GRCh38
rs139428198 Chr.10: 651318-651318 on Build GRCh38
rs986451619 Chr.10: 651373-651373 on Build GRCh38
rs370122680 Chr.10: 651143-651143 on Build GRCh38
rs758528904 Chr.10: 651272-651272 on Build GRCh38
rs760772832 Chr.10: 651073-651074 on Build GRCh38
rs777805339 Chr.10: 651277-651277 on Build GRCh38
rs199865334 Chr.10: 651030-651030 on Build GRCh38
rs1020232234 Chr.10: 651380-651380 on Build GRCh38
rs988717234 Chr.10: 651179-651179 on Build GRCh38
rs746252524 Chr.10: 651319-651319 on Build GRCh38
rs576601241 Chr.10: 651435-651435 on Build GRCh38
rs922676937 Chr.10: 651465-651465 on Build GRCh38

View Full Product Details