Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65502972-65503478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MALAT1
Entrez Gene ID
378938
Gene Name
metastasis associated lung adenocarcinoma transcript 1 (non-protein coding)
Gene Aliases
HCN, LINC00047, NCRNA00047, NEAT2, PRO2853, mascRNA
UniGene
Hs.621695
Gene Chromosome Location
Chr.11: 65497679-65504494 on Build GRCh38
Transcripts
NR_002819.3

SNP Detail

SNP ID Chromosome Location
rs751595960 Chr.11: 65503133-65503133 on Build GRCh38
rs776332890 Chr.11: 65503333-65503333 on Build GRCh38
rs758524824 Chr.11: 65503380-65503381 on Build GRCh38
rs764624639 Chr.11: 65503370-65503370 on Build GRCh38
rs779417050 Chr.11: 65503068-65503068 on Build GRCh38
rs571909713 Chr.11: 65503400-65503400 on Build GRCh38
rs777601312 Chr.11: 65503316-65503316 on Build GRCh38
rs772717107 Chr.11: 65503203-65503203 on Build GRCh38
rs555983322 Chr.11: 65503349-65503349 on Build GRCh38
rs767241666 Chr.11: 65503040-65503040 on Build GRCh38
rs367865679 Chr.11: 65503273-65503273 on Build GRCh38
rs746495814 Chr.11: 65503323-65503323 on Build GRCh38
rs538579405 Chr.11: 65503119-65503119 on Build GRCh38
rs776184770 Chr.11: 65503212-65503212 on Build GRCh38
rs113847292 Chr.11: 65503439-65503439 on Build GRCh38
rs776188206 Chr.11: 65503416-65503416 on Build GRCh38
rs560773874 Chr.11: 65503437-65503437 on Build GRCh38
rs766727457 Chr.11: 65503384-65503384 on Build GRCh38
rs188469226 Chr.11: 65503344-65503344 on Build GRCh38
rs773477843 Chr.11: 65503434-65503434 on Build GRCh38
rs774268908 Chr.11: 65503101-65503101 on Build GRCh38
rs763730705 Chr.11: 65503447-65503447 on Build GRCh38
rs766829373 Chr.11: 65503050-65503050 on Build GRCh38
rs760676580 Chr.11: 65503214-65503214 on Build GRCh38
rs970124896 Chr.11: 65503150-65503150 on Build GRCh38
rs371082259 Chr.11: 65503015-65503015 on Build GRCh38
rs761466407 Chr.11: 65503106-65503106 on Build GRCh38
rs769694231 Chr.11: 65503007-65503007 on Build GRCh38
rs768231116 Chr.11: 65503121-65503122 on Build GRCh38
rs374700152 Chr.11: 65503043-65503043 on Build GRCh38
rs773902830 Chr.11: 65503137-65503138 on Build GRCh38
rs1051608294 Chr.11: 65503293-65503293 on Build GRCh38
rs757173441 Chr.11: 65503399-65503400 on Build GRCh38
rs746005094 Chr.11: 65503238-65503243 on Build GRCh38
rs148133626 Chr.11: 65503027-65503027 on Build GRCh38
rs887177945 Chr.11: 65503002-65503019 on Build GRCh38
rs911351193 Chr.11: 65503298-65503298 on Build GRCh38
rs372419796 Chr.11: 65503217-65503217 on Build GRCh38
rs766858743 Chr.11: 65503416-65503419 on Build GRCh38
rs777801283 Chr.11: 65503386-65503387 on Build GRCh38
rs756344211 Chr.11: 65503380-65503380 on Build GRCh38
rs750755377 Chr.11: 65503110-65503110 on Build GRCh38
rs750144105 Chr.11: 65503041-65503041 on Build GRCh38
rs774465029 Chr.11: 65503105-65503105 on Build GRCh38
rs1013679842 Chr.11: 65503120-65503120 on Build GRCh38
rs764599267 Chr.11: 65503220-65503220 on Build GRCh38
rs747967064 Chr.11: 65503287-65503287 on Build GRCh38
rs373237121 Chr.11: 65503025-65503025 on Build GRCh38
rs936582008 Chr.11: 65503295-65503295 on Build GRCh38
rs979815935 Chr.11: 65503161-65503161 on Build GRCh38
rs569567195 Chr.11: 65503056-65503056 on Build GRCh38
rs768668375 Chr.11: 65503104-65503104 on Build GRCh38
rs754811065 Chr.11: 65503285-65503285 on Build GRCh38
rs769499308 Chr.11: 65503042-65503045 on Build GRCh38
rs752816490 Chr.11: 65503379-65503380 on Build GRCh38
rs763385362 Chr.11: 65503218-65503218 on Build GRCh38
rs924914544 Chr.11: 65503275-65503275 on Build GRCh38
rs575918879 Chr.11: 65503362-65503362 on Build GRCh38
rs762298200 Chr.11: 65503430-65503430 on Build GRCh38
rs768615311 Chr.11: 65503158-65503158 on Build GRCh38
rs754213111 Chr.11: 65503230-65503231 on Build GRCh38
rs748184788 Chr.11: 65503074-65503074 on Build GRCh38
rs759757253 Chr.11: 65503330-65503332 on Build GRCh38
rs888407549 Chr.11: 65503395-65503395 on Build GRCh38
rs747314103 Chr.11: 65503089-65503089 on Build GRCh38
rs757314892 Chr.11: 65503266-65503266 on Build GRCh38
rs184847017 Chr.11: 65503410-65503410 on Build GRCh38
rs758283445 Chr.11: 65503444-65503444 on Build GRCh38
rs755452277 Chr.11: 65503064-65503064 on Build GRCh38
rs1004166415 Chr.11: 65503414-65503414 on Build GRCh38
rs751365619 Chr.11: 65503448-65503448 on Build GRCh38
rs770396686 Chr.11: 65503326-65503326 on Build GRCh38
rs757199237 Chr.11: 65503450-65503450 on Build GRCh38
rs766914316 Chr.11: 65503190-65503191 on Build GRCh38
rs769044265 Chr.11: 65503031-65503031 on Build GRCh38
rs114213656 Chr.11: 65503179-65503179 on Build GRCh38
rs751565708 Chr.11: 65503393-65503396 on Build GRCh38
rs769484169 Chr.11: 65503406-65503406 on Build GRCh38
rs767899509 Chr.11: 65503035-65503035 on Build GRCh38
rs745367365 Chr.11: 65503403-65503403 on Build GRCh38
rs774932005 Chr.11: 65503427-65503427 on Build GRCh38
rs758228699 Chr.11: 65503389-65503389 on Build GRCh38
rs985833932 Chr.11: 65503234-65503234 on Build GRCh38
rs200351093 Chr.11: 65503385-65503385 on Build GRCh38
rs772958453 Chr.11: 65503213-65503213 on Build GRCh38
rs771932551 Chr.11: 65503134-65503134 on Build GRCh38
rs375703167 Chr.11: 65503221-65503221 on Build GRCh38
rs777949106 Chr.11: 65503080-65503080 on Build GRCh38
rs374666797 Chr.11: 65503336-65503336 on Build GRCh38
rs574084712 Chr.11: 65503207-65503207 on Build GRCh38
rs373626664 Chr.11: 65503453-65503453 on Build GRCh38
rs765531162 Chr.11: 65503339-65503339 on Build GRCh38
rs371179067 Chr.11: 65503118-65503118 on Build GRCh38
rs752582849 Chr.11: 65503386-65503386 on Build GRCh38
rs758512610 Chr.11: 65503075-65503075 on Build GRCh38
rs116978099 Chr.11: 65503394-65503394 on Build GRCh38
rs760304882 Chr.11: 65503123-65503123 on Build GRCh38
rs765700554 Chr.11: 65503130-65503130 on Build GRCh38
rs762582136 Chr.11: 65503104-65503110 on Build GRCh38
rs749073597 Chr.11: 65503145-65503145 on Build GRCh38
rs931698482 Chr.11: 65503057-65503057 on Build GRCh38
rs953229225 Chr.11: 65503229-65503229 on Build GRCh38
rs775290253 Chr.11: 65503096-65503096 on Build GRCh38
rs761229689 Chr.11: 65503172-65503174 on Build GRCh38
rs905300778 Chr.11: 65503365-65503365 on Build GRCh38
rs779378082 Chr.11: 65503066-65503066 on Build GRCh38
rs774938572 Chr.11: 65503048-65503049 on Build GRCh38
rs769472985 Chr.11: 65503091-65503091 on Build GRCh38
rs774763117 Chr.11: 65503373-65503373 on Build GRCh38
rs748159274 Chr.11: 65503172-65503172 on Build GRCh38
rs768059903 Chr.11: 65503433-65503433 on Build GRCh38
rs767555323 Chr.11: 65503376-65503376 on Build GRCh38
rs781294493 Chr.11: 65503135-65503135 on Build GRCh38
rs750517358 Chr.11: 65503377-65503377 on Build GRCh38
rs765958835 Chr.11: 65503125-65503125 on Build GRCh38
rs781448780 Chr.11: 65503269-65503269 on Build GRCh38
rs371646684 Chr.11: 65503330-65503330 on Build GRCh38
rs376686732 Chr.11: 65503435-65503435 on Build GRCh38
rs372595584 Chr.11: 65503374-65503374 on Build GRCh38
rs758459500 Chr.11: 65503131-65503131 on Build GRCh38
rs368725127 Chr.11: 65503206-65503206 on Build GRCh38
rs750664653 Chr.11: 65503272-65503272 on Build GRCh38
rs571641196 Chr.11: 65503166-65503166 on Build GRCh38
rs768851482 Chr.11: 65503419-65503419 on Build GRCh38
rs901418118 Chr.11: 65503426-65503426 on Build GRCh38
rs772084040 Chr.11: 65503307-65503307 on Build GRCh38
rs756416948 Chr.11: 65503047-65503047 on Build GRCh38
rs775276724 Chr.11: 65503011-65503011 on Build GRCh38
rs192992046 Chr.11: 65503391-65503391 on Build GRCh38
rs370638337 Chr.11: 65503038-65503038 on Build GRCh38
rs1031483096 Chr.11: 65503124-65503124 on Build GRCh38
rs551783887 Chr.11: 65503127-65503127 on Build GRCh38
rs367742169 Chr.11: 65503099-65503099 on Build GRCh38

View Full Product Details