Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8200211-8200725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66C
Entrez Gene ID
440078
Gene Name
family with sequence similarity 66 member C
Gene Aliases
-
UniGene
Hs.512257
Gene Chromosome Location
Chr.12: 8180209-8201000 on Build GRCh38
Transcripts
NR_026788.1

SNP Detail

SNP ID Chromosome Location
rs911956391 Chr.12: 8200390-8200390 on Build GRCh38
rs766281938 Chr.12: 8200436-8200436 on Build GRCh38
rs200087976 Chr.12: 8200656-8200656 on Build GRCh38
rs180727345 Chr.12: 8200243-8200243 on Build GRCh38
rs12372630 Chr.12: 8200236-8200236 on Build GRCh38
rs1047839294 Chr.12: 8200339-8200339 on Build GRCh38
rs916219451 Chr.12: 8200264-8200264 on Build GRCh38
rs745850335 Chr.12: 8200677-8200677 on Build GRCh38
rs201062268 Chr.12: 8200352-8200352 on Build GRCh38
rs189574351 Chr.12: 8200512-8200512 on Build GRCh38
rs768013901 Chr.12: 8200388-8200388 on Build GRCh38
rs11043934 Chr.12: 8200306-8200306 on Build GRCh38
rs200773126 Chr.12: 8200548-8200548 on Build GRCh38
rs918506196 Chr.12: 8200687-8200687 on Build GRCh38
rs75091444 Chr.12: 8200275-8200275 on Build GRCh38
rs147753917 Chr.12: 8200460-8200460 on Build GRCh38
rs755187681 Chr.12: 8200469-8200469 on Build GRCh38
rs999692826 Chr.12: 8200505-8200505 on Build GRCh38
rs990466142 Chr.12: 8200251-8200251 on Build GRCh38
rs78891334 Chr.12: 8200274-8200274 on Build GRCh38
rs753045722 Chr.12: 8200344-8200354 on Build GRCh38
rs568652611 Chr.12: 8200452-8200452 on Build GRCh38
rs200792551 Chr.12: 8200411-8200411 on Build GRCh38
rs1023724427 Chr.12: 8200575-8200575 on Build GRCh38
rs534421207 Chr.12: 8200570-8200570 on Build GRCh38
rs201493975 Chr.12: 8200265-8200265 on Build GRCh38
rs112277666 Chr.12: 8200306-8200306 on Build GRCh38
rs774599269 Chr.12: 8200321-8200321 on Build GRCh38
rs753472541 Chr.12: 8200287-8200287 on Build GRCh38
rs918423309 Chr.12: 8200290-8200293 on Build GRCh38
rs950873175 Chr.12: 8200391-8200391 on Build GRCh38
rs771979759 Chr.12: 8200643-8200643 on Build GRCh38
rs7297314 Chr.12: 8200405-8200405 on Build GRCh38
rs752051160 Chr.12: 8200431-8200434 on Build GRCh38
rs201513401 Chr.12: 8200467-8200467 on Build GRCh38
rs900478659 Chr.12: 8200451-8200451 on Build GRCh38
rs201926381 Chr.12: 8200579-8200579 on Build GRCh38
rs75021043 Chr.12: 8200308-8200308 on Build GRCh38
rs778587555 Chr.12: 8200522-8200522 on Build GRCh38
rs902717589 Chr.12: 8200470-8200470 on Build GRCh38
rs199807633 Chr.12: 8200360-8200360 on Build GRCh38
rs376164034 Chr.12: 8200493-8200493 on Build GRCh38
rs757047451 Chr.12: 8200527-8200527 on Build GRCh38
rs962371352 Chr.12: 8200577-8200577 on Build GRCh38
rs951238512 Chr.12: 8200688-8200688 on Build GRCh38
rs201543007 Chr.12: 8200305-8200306 on Build GRCh38
rs185165803 Chr.12: 8200446-8200446 on Build GRCh38

View Full Product Details