Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 130041808-130042338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100190940
Entrez Gene ID
100190940
Gene Name
uncharacterized LOC100190940
Gene Aliases
-
UniGene
Hs.625993
Gene Chromosome Location
Chr.12: 130033812-130042342 on Build GRCh38
Transcripts
NR_024457.2

SNP Detail

SNP ID Chromosome Location
rs1001218460 Chr.12: 130041991-130041991 on Build GRCh38
rs886644745 Chr.12: 130041872-130041872 on Build GRCh38
rs1027877333 Chr.12: 130041989-130041989 on Build GRCh38
rs995506179 Chr.12: 130041971-130041971 on Build GRCh38
rs1052014592 Chr.12: 130041846-130041846 on Build GRCh38
rs967646490 Chr.12: 130042114-130042114 on Build GRCh38
rs886720345 Chr.12: 130042319-130042319 on Build GRCh38
rs762579153 Chr.12: 130042317-130042317 on Build GRCh38
rs747341982 Chr.12: 130041946-130041946 on Build GRCh38
rs1016453346 Chr.12: 130041966-130041966 on Build GRCh38
rs944443494 Chr.12: 130042266-130042266 on Build GRCh38
rs771334444 Chr.12: 130041980-130041980 on Build GRCh38
rs921970120 Chr.12: 130042185-130042185 on Build GRCh38
rs571405224 Chr.12: 130042104-130042105 on Build GRCh38
rs1038748400 Chr.12: 130042272-130042272 on Build GRCh38
rs866048269 Chr.12: 130042280-130042280 on Build GRCh38
rs933320849 Chr.12: 130042241-130042241 on Build GRCh38
rs1004863873 Chr.12: 130042050-130042050 on Build GRCh38
rs1034970500 Chr.12: 130042092-130042092 on Build GRCh38
rs960811322 Chr.12: 130042113-130042113 on Build GRCh38
rs76852761 Chr.12: 130042115-130042115 on Build GRCh38
rs759214648 Chr.12: 130042126-130042126 on Build GRCh38
rs111951983 Chr.12: 130042163-130042163 on Build GRCh38
rs959688922 Chr.12: 130042044-130042044 on Build GRCh38
rs764733993 Chr.12: 130042141-130042141 on Build GRCh38
rs1020184676 Chr.12: 130042096-130042096 on Build GRCh38
rs984253740 Chr.12: 130042291-130042300 on Build GRCh38
rs1052468841 Chr.12: 130042267-130042267 on Build GRCh38
rs1696428 Chr.12: 130042181-130042181 on Build GRCh38
rs1005031649 Chr.12: 130041879-130041879 on Build GRCh38
rs911559358 Chr.12: 130042265-130042265 on Build GRCh38
rs765011814 Chr.12: 130041959-130041959 on Build GRCh38
rs900681861 Chr.12: 130042287-130042287 on Build GRCh38
rs963093983 Chr.12: 130041972-130041972 on Build GRCh38
rs879944445 Chr.12: 130042022-130042022 on Build GRCh38
rs1049677510 Chr.12: 130042318-130042318 on Build GRCh38
rs1039650796 Chr.12: 130041923-130041923 on Build GRCh38
rs1008645679 Chr.12: 130041855-130041855 on Build GRCh38
rs113122791 Chr.12: 130042006-130042006 on Build GRCh38
rs898924882 Chr.12: 130042306-130042306 on Build GRCh38
rs979097799 Chr.12: 130042157-130042157 on Build GRCh38
rs900630898 Chr.12: 130041963-130041963 on Build GRCh38
rs552680500 Chr.12: 130042178-130042178 on Build GRCh38
rs868168945 Chr.12: 130042198-130042198 on Build GRCh38
rs915730367 Chr.12: 130042121-130042121 on Build GRCh38

View Full Product Details