Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51814813-51815320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00282
Entrez Gene ID
283521
Gene Name
long intergenic non-protein coding RNA 282
Gene Aliases
NCRNA00282
UniGene
Hs.649896
Gene Chromosome Location
Chr.13: 51813347-51845150 on Build GRCh38
Transcripts
NR_027047.1 NR_027048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764479 c.444C>T p.P148P Chr.13: 51815045-51815045 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs114784176 Chr.13: 51815013-51815013 on Build GRCh38
rs186718672 Chr.13: 51815195-51815195 on Build GRCh38
rs945549609 Chr.13: 51815173-51815173 on Build GRCh38
rs773406280 Chr.13: 51814839-51814839 on Build GRCh38
rs570290509 Chr.13: 51815117-51815117 on Build GRCh38
rs182934574 Chr.13: 51815104-51815104 on Build GRCh38
rs117201520 Chr.13: 51815040-51815040 on Build GRCh38
rs565358840 Chr.13: 51814995-51814995 on Build GRCh38
rs1024171841 Chr.13: 51815162-51815162 on Build GRCh38
rs1024556983 Chr.13: 51814901-51814901 on Build GRCh38
rs962391201 Chr.13: 51814966-51814966 on Build GRCh38
rs144644351 Chr.13: 51814838-51814838 on Build GRCh38
rs558911477 Chr.13: 51815176-51815176 on Build GRCh38
rs970035512 Chr.13: 51814919-51814919 on Build GRCh38
rs751768067 Chr.13: 51815205-51815205 on Build GRCh38
rs935346502 Chr.13: 51815100-51815100 on Build GRCh38
rs573305130 Chr.13: 51815283-51815283 on Build GRCh38
rs373070731 Chr.13: 51815080-51815080 on Build GRCh38
rs933697429 Chr.13: 51814989-51814989 on Build GRCh38
rs761421297 Chr.13: 51815254-51815254 on Build GRCh38
rs1016634159 Chr.13: 51814930-51814930 on Build GRCh38
rs1016876715 Chr.13: 51815182-51815182 on Build GRCh38
rs1025727576 Chr.13: 51815226-51815226 on Build GRCh38
rs980003156 Chr.13: 51814920-51814920 on Build GRCh38
rs150063336 Chr.13: 51815294-51815294 on Build GRCh38
rs1014078679 Chr.13: 51815267-51815267 on Build GRCh38
rs972365002 Chr.13: 51814982-51814982 on Build GRCh38
rs118154526 Chr.13: 51814845-51814845 on Build GRCh38
rs767725064 Chr.13: 51815230-51815230 on Build GRCh38
rs139390540 Chr.13: 51815257-51815257 on Build GRCh38
rs1009765135 Chr.13: 51814858-51814858 on Build GRCh38
rs531524675 Chr.13: 51814928-51814928 on Build GRCh38
rs1033739530 Chr.13: 51814916-51814916 on Build GRCh38
rs1026239397 Chr.13: 51815029-51815029 on Build GRCh38
rs544909617 Chr.13: 51814961-51814961 on Build GRCh38
rs971255455 Chr.13: 51815005-51815005 on Build GRCh38
rs918349303 Chr.13: 51814987-51814987 on Build GRCh38
rs868601809 Chr.13: 51815220-51815220 on Build GRCh38
rs760955604 Chr.13: 51815014-51815027 on Build GRCh38
rs902044701 Chr.13: 51814848-51814848 on Build GRCh38
rs1042656073 Chr.13: 51814878-51814878 on Build GRCh38
rs903822230 Chr.13: 51814899-51814899 on Build GRCh38
rs369431587 Chr.13: 51815208-51815208 on Build GRCh38
rs865966524 Chr.13: 51815265-51815265 on Build GRCh38
rs896609682 Chr.13: 51815132-51815132 on Build GRCh38
rs147954980 Chr.13: 51815031-51815031 on Build GRCh38
rs753280858 Chr.13: 51815286-51815286 on Build GRCh38
rs539204754 Chr.13: 51815140-51815140 on Build GRCh38
rs4290380 Chr.13: 51815211-51815211 on Build GRCh38
rs567675979 Chr.13: 51815038-51815038 on Build GRCh38
rs1013693602 Chr.13: 51814952-51814952 on Build GRCh38
rs923437304 Chr.13: 51815125-51815125 on Build GRCh38
rs756545918 Chr.13: 51815289-51815289 on Build GRCh38
rs559846475 Chr.13: 51815032-51815032 on Build GRCh38
rs767223590 Chr.13: 51815292-51815292 on Build GRCh38
rs533656523 Chr.13: 51815252-51815252 on Build GRCh38
rs905674562 Chr.13: 51815165-51815165 on Build GRCh38
rs1024545302 Chr.13: 51814964-51814964 on Build GRCh38
rs943096744 Chr.13: 51815085-51815085 on Build GRCh38

View Full Product Details