Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99197262-99197807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAC2-AS1
Entrez Gene ID
100289373
Gene Name
UBAC2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.225914
Gene Chromosome Location
Chr.13: 99196374-99200757 on Build GRCh38
Transcripts
NR_036531.1 NR_036532.1

SNP Detail

SNP ID Chromosome Location
rs187083292 Chr.13: 99197663-99197663 on Build GRCh38
rs974496076 Chr.13: 99197739-99197739 on Build GRCh38
rs1027053809 Chr.13: 99197455-99197455 on Build GRCh38
rs61968334 Chr.13: 99197527-99197527 on Build GRCh38
rs960724889 Chr.13: 99197560-99197560 on Build GRCh38
rs913710688 Chr.13: 99197346-99197346 on Build GRCh38
rs73563969 Chr.13: 99197314-99197314 on Build GRCh38
rs552174269 Chr.13: 99197703-99197703 on Build GRCh38
rs945163704 Chr.13: 99197531-99197531 on Build GRCh38
rs759994360 Chr.13: 99197570-99197570 on Build GRCh38
rs141179199 Chr.13: 99197405-99197405 on Build GRCh38
rs938203161 Chr.13: 99197326-99197326 on Build GRCh38
rs142981700 Chr.13: 99197582-99197582 on Build GRCh38
rs759900858 Chr.13: 99197488-99197488 on Build GRCh38
rs72648066 Chr.13: 99197719-99197719 on Build GRCh38
rs562032924 Chr.13: 99197566-99197566 on Build GRCh38
rs767654968 Chr.13: 99197510-99197510 on Build GRCh38
rs922933673 Chr.13: 99197745-99197745 on Build GRCh38
rs553283603 Chr.13: 99197301-99197301 on Build GRCh38
rs1004385093 Chr.13: 99197473-99197473 on Build GRCh38
rs1015773489 Chr.13: 99197391-99197391 on Build GRCh38
rs962836956 Chr.13: 99197499-99197499 on Build GRCh38
rs998382065 Chr.13: 99197438-99197438 on Build GRCh38
rs1043829250 Chr.13: 99197353-99197353 on Build GRCh38
rs926495793 Chr.13: 99197329-99197329 on Build GRCh38
rs72648065 Chr.13: 99197375-99197375 on Build GRCh38
rs1001898857 Chr.13: 99197344-99197346 on Build GRCh38
rs992099505 Chr.13: 99197341-99197341 on Build GRCh38
rs912545533 Chr.13: 99197491-99197491 on Build GRCh38
rs150748246 Chr.13: 99197454-99197454 on Build GRCh38
rs1016239188 Chr.13: 99197674-99197674 on Build GRCh38
rs892259680 Chr.13: 99197307-99197307 on Build GRCh38
rs139341674 Chr.13: 99197575-99197575 on Build GRCh38
rs1027106069 Chr.13: 99197710-99197710 on Build GRCh38
rs976365149 Chr.13: 99197514-99197514 on Build GRCh38
rs901542472 Chr.13: 99197418-99197418 on Build GRCh38
rs201148799 Chr.13: 99197452-99197452 on Build GRCh38
rs11069357 Chr.13: 99197376-99197376 on Build GRCh38
rs533809183 Chr.13: 99197733-99197733 on Build GRCh38
rs774929900 Chr.13: 99197448-99197448 on Build GRCh38
rs1017397507 Chr.13: 99197483-99197483 on Build GRCh38
rs1043472004 Chr.13: 99197322-99197322 on Build GRCh38
rs1013842955 Chr.13: 99197645-99197645 on Build GRCh38
rs528462595 Chr.13: 99197526-99197526 on Build GRCh38
rs760710876 Chr.13: 99197547-99197547 on Build GRCh38
rs569424214 Chr.13: 99197583-99197583 on Build GRCh38
rs979302610 Chr.13: 99197696-99197696 on Build GRCh38
rs960631149 Chr.13: 99197338-99197338 on Build GRCh38
rs184048673 Chr.13: 99197352-99197352 on Build GRCh38

View Full Product Details