Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 67899145-67899643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC643542
Entrez Gene ID
643542
Gene Name
uncharacterized LOC643542
Gene Aliases
-
UniGene
Hs.652901
Gene Chromosome Location
Chr.18: 67516546-67899619 on Build GRCh38
Transcripts
NR_033921.1

SNP Detail

SNP ID Chromosome Location
rs760352268 Chr.18: 67899478-67899478 on Build GRCh38
rs983798768 Chr.18: 67899323-67899323 on Build GRCh38
rs1040344160 Chr.18: 67899528-67899528 on Build GRCh38
rs1042465996 Chr.18: 67899214-67899214 on Build GRCh38
rs557309755 Chr.18: 67899232-67899232 on Build GRCh38
rs1041595400 Chr.18: 67899580-67899580 on Build GRCh38
rs765943106 Chr.18: 67899566-67899566 on Build GRCh38
rs902509455 Chr.18: 67899216-67899216 on Build GRCh38
rs1034438326 Chr.18: 67899404-67899404 on Build GRCh38
rs539859988 Chr.18: 67899331-67899331 on Build GRCh38
rs965481679 Chr.18: 67899239-67899239 on Build GRCh38
rs1019797103 Chr.18: 67899227-67899227 on Build GRCh38
rs539395703 Chr.18: 67899226-67899226 on Build GRCh38
rs971048541 Chr.18: 67899381-67899381 on Build GRCh38
rs915398132 Chr.18: 67899204-67899204 on Build GRCh38
rs151228012 Chr.18: 67899581-67899581 on Build GRCh38
rs948017671 Chr.18: 67899352-67899352 on Build GRCh38
rs938757443 Chr.18: 67899461-67899461 on Build GRCh38
rs372344815 Chr.18: 67899606-67899606 on Build GRCh38
rs773044168 Chr.18: 67899215-67899215 on Build GRCh38
rs891832588 Chr.18: 67899175-67899175 on Build GRCh38
rs1014705256 Chr.18: 67899374-67899374 on Build GRCh38
rs533662079 Chr.18: 67899591-67899591 on Build GRCh38
rs1043051092 Chr.18: 67899542-67899542 on Build GRCh38
rs190661877 Chr.18: 67899614-67899614 on Build GRCh38
rs944703512 Chr.18: 67899513-67899513 on Build GRCh38
rs1025304270 Chr.18: 67899248-67899248 on Build GRCh38
rs750044100 Chr.18: 67899545-67899545 on Build GRCh38
rs866654703 Chr.18: 67899414-67899414 on Build GRCh38
rs978130890 Chr.18: 67899491-67899491 on Build GRCh38
rs931886563 Chr.18: 67899549-67899549 on Build GRCh38
rs1049477727 Chr.18: 67899567-67899567 on Build GRCh38
rs887620435 Chr.18: 67899343-67899343 on Build GRCh38
rs143691308 Chr.18: 67899408-67899408 on Build GRCh38
rs756759305 Chr.18: 67899244-67899244 on Build GRCh38
rs1004619837 Chr.18: 67899364-67899364 on Build GRCh38
rs924069515 Chr.18: 67899505-67899505 on Build GRCh38
rs1023331243 Chr.18: 67899608-67899608 on Build GRCh38
rs756465963 Chr.18: 67899577-67899577 on Build GRCh38
rs1005200589 Chr.18: 67899605-67899605 on Build GRCh38
rs946779698 Chr.18: 67899208-67899208 on Build GRCh38
rs17078262 Chr.18: 67899593-67899593 on Build GRCh38
rs1053170636 Chr.18: 67899498-67899498 on Build GRCh38
rs982336208 Chr.18: 67899371-67899372 on Build GRCh38
rs996507089 Chr.18: 67899329-67899329 on Build GRCh38
rs959611796 Chr.18: 67899450-67899450 on Build GRCh38
rs145730436 Chr.18: 67899473-67899473 on Build GRCh38
rs972536501 Chr.18: 67899247-67899247 on Build GRCh38
rs148503995 Chr.18: 67899481-67899481 on Build GRCh38
rs947355648 Chr.18: 67899538-67899538 on Build GRCh38
rs1008878628 Chr.18: 67899221-67899221 on Build GRCh38
rs915384659 Chr.18: 67899475-67899475 on Build GRCh38
rs981165221 Chr.18: 67899386-67899386 on Build GRCh38
rs969698273 Chr.18: 67899609-67899609 on Build GRCh38
rs1033996280 Chr.18: 67899599-67899599 on Build GRCh38
rs537768758 Chr.18: 67899245-67899245 on Build GRCh38
rs141405447 Chr.18: 67899603-67899603 on Build GRCh38
rs572541565 Chr.18: 67899324-67899324 on Build GRCh38
rs368528103 Chr.18: 67899320-67899320 on Build GRCh38
rs1003037098 Chr.18: 67899595-67899595 on Build GRCh38

View Full Product Details