Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5558297-5558837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TINCR
Entrez Gene ID
257000
Gene Name
tissue differentiation-inducing non-protein coding RNA
Gene Aliases
LINC00036, NCRNA00036, PLAC2, onco-lncRNA-16
UniGene
Hs.515575
Gene Chromosome Location
Chr.19: 5558167-5568034 on Build GRCh38
Transcripts
NR_027064.2

SNP Detail

SNP ID Chromosome Location
rs569117682 Chr.19: 5558584-5558584 on Build GRCh38
rs758154234 Chr.19: 5558348-5558348 on Build GRCh38
rs751202761 Chr.19: 5558606-5558606 on Build GRCh38
rs533339705 Chr.19: 5558459-5558459 on Build GRCh38
rs533739617 Chr.19: 5558721-5558721 on Build GRCh38
rs866340486 Chr.19: 5558374-5558374 on Build GRCh38
rs765950274 Chr.19: 5558546-5558546 on Build GRCh38
rs944965832 Chr.19: 5558411-5558411 on Build GRCh38
rs922144332 Chr.19: 5558713-5558713 on Build GRCh38
rs898088667 Chr.19: 5558766-5558766 on Build GRCh38
rs923595705 Chr.19: 5558633-5558633 on Build GRCh38
rs532440092 Chr.19: 5558371-5558371 on Build GRCh38
rs985672314 Chr.19: 5558590-5558590 on Build GRCh38
rs189297333 Chr.19: 5558421-5558421 on Build GRCh38
rs908079659 Chr.19: 5558750-5558750 on Build GRCh38
rs34657688 Chr.19: 5558786-5558786 on Build GRCh38
rs895505346 Chr.19: 5558537-5558537 on Build GRCh38
rs1049227664 Chr.19: 5558463-5558463 on Build GRCh38
rs976687643 Chr.19: 5558607-5558607 on Build GRCh38
rs1036686748 Chr.19: 5558506-5558506 on Build GRCh38
rs888168139 Chr.19: 5558487-5558487 on Build GRCh38
rs953490765 Chr.19: 5558567-5558567 on Build GRCh38
rs1039730231 Chr.19: 5558413-5558413 on Build GRCh38
rs947843166 Chr.19: 5558670-5558670 on Build GRCh38
rs150308546 Chr.19: 5558649-5558649 on Build GRCh38
rs766984892 Chr.19: 5558765-5558765 on Build GRCh38
rs956119795 Chr.19: 5558366-5558366 on Build GRCh38
rs994755741 Chr.19: 5558780-5558780 on Build GRCh38
rs906750196 Chr.19: 5558361-5558361 on Build GRCh38
rs1003942028 Chr.19: 5558500-5558500 on Build GRCh38
rs999999204 Chr.19: 5558581-5558583 on Build GRCh38
rs1049200438 Chr.19: 5558744-5558744 on Build GRCh38
rs974774389 Chr.19: 5558678-5558678 on Build GRCh38
rs930774204 Chr.19: 5558450-5558452 on Build GRCh38
rs1003747252 Chr.19: 5558727-5558727 on Build GRCh38
rs1027660857 Chr.19: 5558552-5558552 on Build GRCh38
rs746929794 Chr.19: 5558695-5558695 on Build GRCh38
rs1027630185 Chr.19: 5558792-5558792 on Build GRCh38
rs956091992 Chr.19: 5558620-5558620 on Build GRCh38
rs560370815 Chr.19: 5558712-5558712 on Build GRCh38
rs966481015 Chr.19: 5558674-5558674 on Build GRCh38
rs186478254 Chr.19: 5558363-5558363 on Build GRCh38
rs1023263199 Chr.19: 5558561-5558561 on Build GRCh38
rs895568353 Chr.19: 5558794-5558794 on Build GRCh38
rs1014168469 Chr.19: 5558560-5558560 on Build GRCh38
rs1032861681 Chr.19: 5558618-5558618 on Build GRCh38
rs771015517 Chr.19: 5558444-5558447 on Build GRCh38
rs549923802 Chr.19: 5558522-5558522 on Build GRCh38
rs911907929 Chr.19: 5558408-5558408 on Build GRCh38
rs911069838 Chr.19: 5558604-5558604 on Build GRCh38
rs140195060 Chr.19: 5558634-5558634 on Build GRCh38
rs988790244 Chr.19: 5558368-5558368 on Build GRCh38
rs180759742 Chr.19: 5558349-5558349 on Build GRCh38
rs372679389 Chr.19: 5558683-5558683 on Build GRCh38
rs151177308 Chr.19: 5558345-5558345 on Build GRCh38
rs914951149 Chr.19: 5558664-5558664 on Build GRCh38

View Full Product Details