Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 94881028-94881522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC442028
Entrez Gene ID
442028
Gene Name
uncharacterized LOC442028
Gene Aliases
-
UniGene
Hs.611545
Gene Chromosome Location
Chr.2: 94868685-94947342 on Build GRCh38
Transcripts
NR_037597.1

SNP Detail

SNP ID Chromosome Location
rs113221661 Chr.2: 94881491-94881491 on Build GRCh38
rs111684625 Chr.2: 94881245-94881245 on Build GRCh38
rs36041838 Chr.2: 94881248-94881248 on Build GRCh38
rs78663674 Chr.2: 94881366-94881366 on Build GRCh38
rs4991395 Chr.2: 94881140-94881140 on Build GRCh38
rs183411120 Chr.2: 94881058-94881058 on Build GRCh38
rs187467715 Chr.2: 94881410-94881410 on Build GRCh38
rs4991397 Chr.2: 94881150-94881150 on Build GRCh38
rs113521632 Chr.2: 94881336-94881336 on Build GRCh38
rs79417723 Chr.2: 94881456-94881456 on Build GRCh38
rs576346655 Chr.2: 94881181-94881181 on Build GRCh38
rs111358379 Chr.2: 94881196-94881196 on Build GRCh38
rs4991394 Chr.2: 94881080-94881080 on Build GRCh38
rs181714854 Chr.2: 94881100-94881100 on Build GRCh38
rs782080030 Chr.2: 94881484-94881484 on Build GRCh38
rs182876031 Chr.2: 94881395-94881395 on Build GRCh38
rs572455863 Chr.2: 94881094-94881094 on Build GRCh38
rs4996374 Chr.2: 94881182-94881182 on Build GRCh38
rs561686730 Chr.2: 94881208-94881208 on Build GRCh38
rs74572584 Chr.2: 94881078-94881078 on Build GRCh38
rs74843260 Chr.2: 94881394-94881394 on Build GRCh38
rs112610415 Chr.2: 94881351-94881351 on Build GRCh38
rs189017683 Chr.2: 94881101-94881101 on Build GRCh38
rs111440755 Chr.2: 94881276-94881276 on Build GRCh38
rs4991396 Chr.2: 94881143-94881143 on Build GRCh38
rs782068177 Chr.2: 94881111-94881111 on Build GRCh38
rs112481563 Chr.2: 94881346-94881346 on Build GRCh38
rs118061020 Chr.2: 94881341-94881341 on Build GRCh38
rs148300914 Chr.2: 94881490-94881490 on Build GRCh38
rs782017569 Chr.2: 94881110-94881110 on Build GRCh38
rs112190834 Chr.2: 94881224-94881224 on Build GRCh38
rs77583739 Chr.2: 94881307-94881307 on Build GRCh38
rs781973721 Chr.2: 94881388-94881388 on Build GRCh38
rs369125106 Chr.2: 94881377-94881377 on Build GRCh38
rs111291161 Chr.2: 94881420-94881420 on Build GRCh38
rs75165292 Chr.2: 94881295-94881295 on Build GRCh38
rs200012734 Chr.2: 94881195-94881203 on Build GRCh38
rs75557949 Chr.2: 94881361-94881361 on Build GRCh38
rs80315028 Chr.2: 94881443-94881443 on Build GRCh38
rs112137058 Chr.2: 94881374-94881374 on Build GRCh38
rs112318266 Chr.2: 94881325-94881325 on Build GRCh38
rs782414468 Chr.2: 94881106-94881106 on Build GRCh38
rs554158144 Chr.2: 94881070-94881070 on Build GRCh38
rs192513081 Chr.2: 94881151-94881151 on Build GRCh38

View Full Product Details