Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 144768236-144768747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX41
Entrez Gene ID
401014
Gene Name
testis expressed 41 (non-protein coding)
Gene Aliases
DKFZp686O1327, LINC00953
UniGene
Hs.742290
Gene Chromosome Location
Chr.2: 144667967-145076729 on Build GRCh38
Transcripts
NR_033870.2

SNP Detail

SNP ID Chromosome Location
rs946228045 Chr.2: 144768618-144768618 on Build GRCh38
rs1010252188 Chr.2: 144768335-144768335 on Build GRCh38
rs1037915218 Chr.2: 144768300-144768300 on Build GRCh38
rs767548587 Chr.2: 144768537-144768537 on Build GRCh38
rs140397817 Chr.2: 144768495-144768495 on Build GRCh38
rs931872734 Chr.2: 144768699-144768699 on Build GRCh38
rs373912088 Chr.2: 144768283-144768283 on Build GRCh38
rs764411334 Chr.2: 144768316-144768316 on Build GRCh38
rs936010539 Chr.2: 144768703-144768703 on Build GRCh38
rs752571352 Chr.2: 144768560-144768560 on Build GRCh38
rs147153519 Chr.2: 144768333-144768333 on Build GRCh38
rs973512351 Chr.2: 144768494-144768494 on Build GRCh38
rs1041744431 Chr.2: 144768624-144768624 on Build GRCh38
rs941683954 Chr.2: 144768280-144768280 on Build GRCh38
rs562353149 Chr.2: 144768323-144768323 on Build GRCh38
rs533721635 Chr.2: 144768261-144768261 on Build GRCh38
rs1013796402 Chr.2: 144768489-144768489 on Build GRCh38
rs188271129 Chr.2: 144768671-144768671 on Build GRCh38
rs368062111 Chr.2: 144768392-144768392 on Build GRCh38
rs1030195310 Chr.2: 144768417-144768417 on Build GRCh38
rs1002468247 Chr.2: 144768503-144768503 on Build GRCh38
rs758349766 Chr.2: 144768469-144768469 on Build GRCh38
rs572720017 Chr.2: 144768669-144768669 on Build GRCh38
rs762018293 Chr.2: 144768512-144768512 on Build GRCh38
rs183784332 Chr.2: 144768477-144768477 on Build GRCh38
rs750519552 Chr.2: 144768453-144768453 on Build GRCh38
rs114535102 Chr.2: 144768372-144768372 on Build GRCh38
rs887466196 Chr.2: 144768422-144768422 on Build GRCh38
rs1032733333 Chr.2: 144768513-144768513 on Build GRCh38
rs377442013 Chr.2: 144768270-144768270 on Build GRCh38
rs931900586 Chr.2: 144768374-144768374 on Build GRCh38
rs996012546 Chr.2: 144768405-144768405 on Build GRCh38
rs537453129 Chr.2: 144768353-144768353 on Build GRCh38
rs1040258642 Chr.2: 144768368-144768368 on Build GRCh38
rs750418859 Chr.2: 144768643-144768644 on Build GRCh38
rs756010122 Chr.2: 144768632-144768632 on Build GRCh38
rs975847787 Chr.2: 144768657-144768657 on Build GRCh38
rs768115385 Chr.2: 144768277-144768282 on Build GRCh38
rs917387784 Chr.2: 144768599-144768599 on Build GRCh38
rs1057333689 Chr.2: 144768565-144768565 on Build GRCh38
rs925314668 Chr.2: 144768298-144768298 on Build GRCh38
rs115155718 Chr.2: 144768347-144768347 on Build GRCh38
rs762047467 Chr.2: 144768371-144768371 on Build GRCh38
rs55964560 Chr.2: 144768490-144768490 on Build GRCh38
rs1057490859 Chr.2: 144768328-144768328 on Build GRCh38
rs954548207 Chr.2: 144768434-144768434 on Build GRCh38

View Full Product Details