Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46109113-46109611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIRLET7BHG
Entrez Gene ID
400931
Gene Name
MIRLET7B host gene
Gene Aliases
linc-Ppara
UniGene
Hs.235838
Gene Chromosome Location
Chr.22: 46085997-46113928 on Build GRCh38
Transcripts
NR_027033.2 NR_110479.1

SNP Detail

SNP ID Chromosome Location
rs374130636 Chr.22: 46109390-46109390 on Build GRCh38
rs966019887 Chr.22: 46109368-46109368 on Build GRCh38
rs960276715 Chr.22: 46109230-46109230 on Build GRCh38
rs865915825 Chr.22: 46109325-46109325 on Build GRCh38
rs1051928711 Chr.22: 46109156-46109156 on Build GRCh38
rs1014691578 Chr.22: 46109456-46109456 on Build GRCh38
rs529021497 Chr.22: 46109215-46109215 on Build GRCh38
rs922875287 Chr.22: 46109538-46109538 on Build GRCh38
rs912891907 Chr.22: 46109552-46109552 on Build GRCh38
rs73886706 Chr.22: 46109178-46109178 on Build GRCh38
rs939019294 Chr.22: 46109474-46109474 on Build GRCh38
rs267606277 Chr.22: 46109326-46109326 on Build GRCh38
rs569720462 Chr.22: 46109196-46109196 on Build GRCh38
rs558096824 Chr.22: 46109311-46109311 on Build GRCh38
rs1010574410 Chr.22: 46109497-46109497 on Build GRCh38
rs987582590 Chr.22: 46109543-46109543 on Build GRCh38
rs866679744 Chr.22: 46109157-46109157 on Build GRCh38
rs1034658959 Chr.22: 46109431-46109431 on Build GRCh38
rs769104702 Chr.22: 46109316-46109317 on Build GRCh38
rs895832592 Chr.22: 46109449-46109449 on Build GRCh38
rs12159783 Chr.22: 46109561-46109561 on Build GRCh38
rs766374093 Chr.22: 46109357-46109357 on Build GRCh38
rs944830566 Chr.22: 46109141-46109141 on Build GRCh38
rs776925340 Chr.22: 46109303-46109303 on Build GRCh38
rs183161934 Chr.22: 46109409-46109409 on Build GRCh38
rs73177018 Chr.22: 46109293-46109293 on Build GRCh38
rs964535393 Chr.22: 46109518-46109518 on Build GRCh38
rs563705315 Chr.22: 46109166-46109166 on Build GRCh38
rs1038730288 Chr.22: 46109138-46109138 on Build GRCh38
rs11090905 Chr.22: 46109457-46109457 on Build GRCh38
rs771062062 Chr.22: 46109267-46109267 on Build GRCh38
rs758432830 Chr.22: 46109317-46109317 on Build GRCh38
rs779095523 Chr.22: 46109531-46109531 on Build GRCh38
rs375375108 Chr.22: 46109533-46109534 on Build GRCh38
rs968647400 Chr.22: 46109193-46109193 on Build GRCh38
rs894907685 Chr.22: 46109492-46109492 on Build GRCh38
rs939971579 Chr.22: 46109438-46109438 on Build GRCh38
rs745420007 Chr.22: 46109280-46109280 on Build GRCh38
rs757239279 Chr.22: 46109394-46109394 on Build GRCh38
rs547695867 Chr.22: 46109162-46109162 on Build GRCh38
rs1034363995 Chr.22: 46109207-46109207 on Build GRCh38
rs777440037 Chr.22: 46109246-46109246 on Build GRCh38
rs952111103 Chr.22: 46109579-46109580 on Build GRCh38
rs988277196 Chr.22: 46109533-46109533 on Build GRCh38
rs992948017 Chr.22: 46109233-46109233 on Build GRCh38
rs762274849 Chr.22: 46109306-46109306 on Build GRCh38
rs951475137 Chr.22: 46109461-46109461 on Build GRCh38
rs1022481418 Chr.22: 46109502-46109502 on Build GRCh38
rs975703792 Chr.22: 46109536-46109536 on Build GRCh38
rs377146971 Chr.22: 46109308-46109308 on Build GRCh38
rs976030208 Chr.22: 46109372-46109372 on Build GRCh38
rs576470962 Chr.22: 46109406-46109406 on Build GRCh38
rs999807961 Chr.22: 46109546-46109546 on Build GRCh38
rs532788318 Chr.22: 46109167-46109167 on Build GRCh38
rs1018956611 Chr.22: 46109366-46109366 on Build GRCh38
rs1033372114 Chr.22: 46109260-46109260 on Build GRCh38
rs921633313 Chr.22: 46109373-46109373 on Build GRCh38
rs142762314 Chr.22: 46109363-46109363 on Build GRCh38
rs749233337 Chr.22: 46109259-46109259 on Build GRCh38
rs934243068 Chr.22: 46109540-46109540 on Build GRCh38
rs906162970 Chr.22: 46109243-46109243 on Build GRCh38
rs774050131 Chr.22: 46109560-46109560 on Build GRCh38
rs947217485 Chr.22: 46109160-46109172 on Build GRCh38
rs879677971 Chr.22: 46109146-46109146 on Build GRCh38
rs755868628 Chr.22: 46109217-46109217 on Build GRCh38
rs11090903 Chr.22: 46109151-46109151 on Build GRCh38
rs749078209 Chr.22: 46109391-46109391 on Build GRCh38
rs760252579 Chr.22: 46109297-46109297 on Build GRCh38
rs139490021 Chr.22: 46109569-46109569 on Build GRCh38
rs543007224 Chr.22: 46109562-46109562 on Build GRCh38
rs56411695 Chr.22: 46109365-46109365 on Build GRCh38
rs1036702924 Chr.22: 46109205-46109205 on Build GRCh38
rs768339062 Chr.22: 46109490-46109490 on Build GRCh38
rs369080965 Chr.22: 46109422-46109422 on Build GRCh38
rs755211833 Chr.22: 46109381-46109381 on Build GRCh38
rs367888840 Chr.22: 46109279-46109279 on Build GRCh38
rs764465058 Chr.22: 46109299-46109299 on Build GRCh38
rs555890571 Chr.22: 46109415-46109415 on Build GRCh38
rs1019594480 Chr.22: 46109557-46109557 on Build GRCh38
rs528934356 Chr.22: 46109581-46109581 on Build GRCh38
rs1001463552 Chr.22: 46109416-46109416 on Build GRCh38
rs895358720 Chr.22: 46109262-46109262 on Build GRCh38
rs984325763 Chr.22: 46109346-46109346 on Build GRCh38
rs1017115663 Chr.22: 46109512-46109512 on Build GRCh38
rs138696056 Chr.22: 46109302-46109302 on Build GRCh38

View Full Product Details