Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 55657393-55657936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERC2
Entrez Gene ID
26059
Gene Name
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases
CAST, CAST1, ELKSL, SPBC110, Spc110
UniGene
Hs.476389
Gene Chromosome Location
Chr.3: 55508311-56468473 on Build GRCh38
Transcripts
NM_015576.2 NR_132749.1 XM_017006138.1 XM_017006139.1 XM_017006140.1 XM_017006141.1 XM_017006142.1 XM_017006143.1 XM_017006144.1 XM_017006145.1 XM_017006146.1 XM_017006147.1 XM_017006148.1 XM_017006149.1 XM_017006150.1 XM_017006151.1 XM_017006152.1 XM_017006153.1 XM_017006154.1 XM_017006155.1 XM_017006156.1 XM_017006157.1 XM_017006158.1 XM_017006159.1 NR_024615.1

Gene Symbol
ERC2-IT1
Entrez Gene ID
711
Gene Name
ERC2 intronic transcript 1
Gene Aliases
C1orf1, C3orf51, Po42
UniGene
Hs.667161
Gene Chromosome Location
Chr.3: 55657206-55659469 on Build GRCh38
Transcripts
NM_015576.2 NR_132749.1 XM_017006138.1 XM_017006139.1 XM_017006140.1 XM_017006141.1 XM_017006142.1 XM_017006143.1 XM_017006144.1 XM_017006145.1 XM_017006146.1 XM_017006147.1 XM_017006148.1 XM_017006149.1 XM_017006150.1 XM_017006151.1 XM_017006152.1 XM_017006153.1 XM_017006154.1 XM_017006155.1 XM_017006156.1 XM_017006157.1 XM_017006158.1 XM_017006159.1 NR_024615.1

SNP Detail

SNP ID Chromosome Location
rs963911851 Chr.3: 55657576-55657576 on Build GRCh38
rs1046701779 Chr.3: 55657467-55657468 on Build GRCh38
rs770713858 Chr.3: 55657705-55657705 on Build GRCh38
rs999808783 Chr.3: 55657594-55657594 on Build GRCh38
rs1054905543 Chr.3: 55657818-55657818 on Build GRCh38
rs551413779 Chr.3: 55657658-55657658 on Build GRCh38
rs538177385 Chr.3: 55657763-55657763 on Build GRCh38
rs547027618 Chr.3: 55657608-55657608 on Build GRCh38
rs751824503 Chr.3: 55657431-55657431 on Build GRCh38
rs1019533488 Chr.3: 55657540-55657540 on Build GRCh38
rs990621734 Chr.3: 55657630-55657630 on Build GRCh38
rs771952739 Chr.3: 55657755-55657755 on Build GRCh38
rs937520166 Chr.3: 55657862-55657862 on Build GRCh38
rs558189933 Chr.3: 55657831-55657831 on Build GRCh38
rs749382441 Chr.3: 55657680-55657680 on Build GRCh38
rs753027172 Chr.3: 55657753-55657753 on Build GRCh38
rs951665860 Chr.3: 55657730-55657730 on Build GRCh38
rs62251548 Chr.3: 55657574-55657574 on Build GRCh38
rs149357668 Chr.3: 55657749-55657749 on Build GRCh38
rs916321293 Chr.3: 55657886-55657886 on Build GRCh38
rs902445465 Chr.3: 55657560-55657560 on Build GRCh38
rs928853394 Chr.3: 55657860-55657860 on Build GRCh38
rs778931706 Chr.3: 55657743-55657743 on Build GRCh38
rs1055278790 Chr.3: 55657434-55657434 on Build GRCh38
rs1008173064 Chr.3: 55657526-55657526 on Build GRCh38
rs753014332 Chr.3: 55657555-55657555 on Build GRCh38
rs991590818 Chr.3: 55657896-55657896 on Build GRCh38
rs778061634 Chr.3: 55657651-55657651 on Build GRCh38
rs577532977 Chr.3: 55657840-55657840 on Build GRCh38
rs185989603 Chr.3: 55657718-55657718 on Build GRCh38
rs535652652 Chr.3: 55657626-55657626 on Build GRCh38
rs564475594 Chr.3: 55657473-55657473 on Build GRCh38
rs976900431 Chr.3: 55657717-55657717 on Build GRCh38
rs756244512 Chr.3: 55657617-55657617 on Build GRCh38
rs867312855 Chr.3: 55657616-55657616 on Build GRCh38
rs1018293480 Chr.3: 55657690-55657690 on Build GRCh38
rs1024926753 Chr.3: 55657497-55657497 on Build GRCh38
rs533184748 Chr.3: 55657494-55657494 on Build GRCh38
rs781236881 Chr.3: 55657495-55657495 on Build GRCh38
rs566630641 Chr.3: 55657623-55657623 on Build GRCh38
rs1020313831 Chr.3: 55657644-55657644 on Build GRCh38

View Full Product Details