Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3677428-3677922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100133461
Entrez Gene ID
100133461
Gene Name
uncharacterized LOC100133461
Gene Aliases
-
UniGene
Hs.554274
Gene Chromosome Location
Chr.4: 3673593-3677855 on Build GRCh38
Transcripts
NR_034136.1

SNP Detail

SNP ID Chromosome Location
rs556162201 Chr.4: 3677544-3677544 on Build GRCh38
rs946346859 Chr.4: 3677762-3677762 on Build GRCh38
rs974861859 Chr.4: 3677473-3677473 on Build GRCh38
rs896298383 Chr.4: 3677798-3677798 on Build GRCh38
rs989035774 Chr.4: 3677713-3677713 on Build GRCh38
rs996343210 Chr.4: 3677826-3677826 on Build GRCh38
rs931164922 Chr.4: 3677549-3677549 on Build GRCh38
rs779073720 Chr.4: 3677569-3677569 on Build GRCh38
rs775485823 Chr.4: 3677661-3677661 on Build GRCh38
rs1046497693 Chr.4: 3677584-3677584 on Build GRCh38
rs954794699 Chr.4: 3677620-3677620 on Build GRCh38
rs115930152 Chr.4: 3677496-3677496 on Build GRCh38
rs996333312 Chr.4: 3677609-3677609 on Build GRCh38
rs374402766 Chr.4: 3677844-3677844 on Build GRCh38
rs553342712 Chr.4: 3677512-3677512 on Build GRCh38
rs961032482 Chr.4: 3677460-3677460 on Build GRCh38
rs781778632 Chr.4: 3677461-3677461 on Build GRCh38
rs58062370 Chr.4: 3677777-3677777 on Build GRCh38
rs114472420 Chr.4: 3677634-3677634 on Build GRCh38
rs192754849 Chr.4: 3677712-3677712 on Build GRCh38
rs937725600 Chr.4: 3677778-3677778 on Build GRCh38
rs999227895 Chr.4: 3677801-3677801 on Build GRCh38
rs765079948 Chr.4: 3677629-3677629 on Build GRCh38
rs778230573 Chr.4: 3677503-3677503 on Build GRCh38
rs759580875 Chr.4: 3677608-3677608 on Build GRCh38
rs965284982 Chr.4: 3677675-3677675 on Build GRCh38
rs377623479 Chr.4: 3677631-3677631 on Build GRCh38
rs1032939227 Chr.4: 3677456-3677456 on Build GRCh38
rs905048449 Chr.4: 3677574-3677574 on Build GRCh38
rs933397594 Chr.4: 3677488-3677488 on Build GRCh38
rs1018201621 Chr.4: 3677510-3677510 on Build GRCh38
rs184914427 Chr.4: 3677572-3677572 on Build GRCh38
rs542056634 Chr.4: 3677589-3677589 on Build GRCh38
rs900588365 Chr.4: 3677865-3677865 on Build GRCh38
rs993452516 Chr.4: 3677472-3677472 on Build GRCh38
rs997987308 Chr.4: 3677893-3677893 on Build GRCh38
rs563911615 Chr.4: 3677782-3677782 on Build GRCh38
rs913601563 Chr.4: 3677739-3677739 on Build GRCh38
rs1034926536 Chr.4: 3677451-3677451 on Build GRCh38
rs952131293 Chr.4: 3677482-3677482 on Build GRCh38
rs1047624761 Chr.4: 3677813-3677813 on Build GRCh38
rs776646109 Chr.4: 3677592-3677592 on Build GRCh38
rs574640607 Chr.4: 3677527-3677527 on Build GRCh38
rs1029124992 Chr.4: 3677833-3677833 on Build GRCh38
rs567966334 Chr.4: 3677799-3677799 on Build GRCh38
rs148601821 Chr.4: 3677839-3677839 on Build GRCh38
rs555361642 Chr.4: 3677601-3677601 on Build GRCh38
rs535121191 Chr.4: 3677897-3677897 on Build GRCh38
rs773110120 Chr.4: 3677761-3677761 on Build GRCh38
rs553595519 Chr.4: 3677457-3677457 on Build GRCh38
rs1053853722 Chr.4: 3677810-3677810 on Build GRCh38
rs187786288 Chr.4: 3677677-3677677 on Build GRCh38
rs993334643 Chr.4: 3677618-3677618 on Build GRCh38
rs1054957905 Chr.4: 3677552-3677552 on Build GRCh38
rs10001474 Chr.4: 3677481-3677481 on Build GRCh38
rs6827536 Chr.4: 3677764-3677764 on Build GRCh38
rs976763094 Chr.4: 3677513-3677513 on Build GRCh38
rs528610170 Chr.4: 3677822-3677822 on Build GRCh38
rs923542983 Chr.4: 3677520-3677520 on Build GRCh38
rs572175868 Chr.4: 3677602-3677602 on Build GRCh38
rs149639714 Chr.4: 3677650-3677650 on Build GRCh38
rs59916646 Chr.4: 3677707-3677707 on Build GRCh38
rs1000205982 Chr.4: 3677595-3677595 on Build GRCh38
rs1043483383 Chr.4: 3677573-3677573 on Build GRCh38
rs528130323 Chr.4: 3677832-3677832 on Build GRCh38
rs749562033 Chr.4: 3677518-3677518 on Build GRCh38
rs539272121 Chr.4: 3677891-3677891 on Build GRCh38
rs934867269 Chr.4: 3677525-3677525 on Build GRCh38

View Full Product Details