Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105569514-105570045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_011532050.2 XM_011532051.2 NR_046840.1

Gene Symbol
ARHGEF38-IT1
Entrez Gene ID
100874374
Gene Name
ARHGEF38 intronic transcript 1
Gene Aliases
-
UniGene
Hs.668908
Gene Chromosome Location
Chr.4: 105561591-105570238 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_011532050.2 XM_011532051.2 NR_046840.1

SNP Detail

SNP ID Chromosome Location
rs998011876 Chr.4: 105569926-105569926 on Build GRCh38
rs990407243 Chr.4: 105569647-105569647 on Build GRCh38
rs1048960551 Chr.4: 105569825-105569825 on Build GRCh38
rs77167945 Chr.4: 105569616-105569616 on Build GRCh38
rs568188332 Chr.4: 105569995-105569995 on Build GRCh38
rs945260678 Chr.4: 105569547-105569547 on Build GRCh38
rs1046309240 Chr.4: 105569834-105569834 on Build GRCh38
rs956015614 Chr.4: 105570009-105570009 on Build GRCh38
rs1007385096 Chr.4: 105569566-105569566 on Build GRCh38
rs769390960 Chr.4: 105569644-105569644 on Build GRCh38
rs1027591913 Chr.4: 105569672-105569672 on Build GRCh38
rs766037903 Chr.4: 105569789-105569789 on Build GRCh38
rs975417703 Chr.4: 105569766-105569766 on Build GRCh38
rs774492058 Chr.4: 105569670-105569670 on Build GRCh38
rs1019826829 Chr.4: 105569755-105569755 on Build GRCh38
rs951890736 Chr.4: 105569563-105569563 on Build GRCh38
rs1056112141 Chr.4: 105569777-105569777 on Build GRCh38
rs887718095 Chr.4: 105569826-105569826 on Build GRCh38
rs995677503 Chr.4: 105569651-105569651 on Build GRCh38
rs1044982743 Chr.4: 105569731-105569731 on Build GRCh38
rs531424138 Chr.4: 105569580-105569580 on Build GRCh38
rs924625053 Chr.4: 105569604-105569604 on Build GRCh38
rs966129083 Chr.4: 105569867-105569867 on Build GRCh38
rs547992439 Chr.4: 105569796-105569796 on Build GRCh38
rs182744687 Chr.4: 105569898-105569898 on Build GRCh38
rs190494574 Chr.4: 105569759-105569759 on Build GRCh38
rs927907071 Chr.4: 105569857-105569857 on Build GRCh38
rs74510421 Chr.4: 105569790-105569790 on Build GRCh38
rs546065453 Chr.4: 105569713-105569713 on Build GRCh38
rs546694257 Chr.4: 105569829-105569829 on Build GRCh38
rs906167260 Chr.4: 105569601-105569601 on Build GRCh38
rs948864794 Chr.4: 105569685-105569685 on Build GRCh38
rs990189502 Chr.4: 105570021-105570021 on Build GRCh38
rs188074390 Chr.4: 105570016-105570016 on Build GRCh38
rs1031637078 Chr.4: 105569958-105569958 on Build GRCh38
rs376003759 Chr.4: 105569957-105569957 on Build GRCh38
rs926379876 Chr.4: 105569748-105569748 on Build GRCh38
rs1007332712 Chr.4: 105569856-105569856 on Build GRCh38
rs10019536 Chr.4: 105569857-105569857 on Build GRCh38
rs1017668246 Chr.4: 105569573-105569573 on Build GRCh38
rs914858798 Chr.4: 105569652-105569652 on Build GRCh38
rs373073942 Chr.4: 105569693-105569693 on Build GRCh38
rs951597354 Chr.4: 105569716-105569716 on Build GRCh38
rs754419374 Chr.4: 105569980-105569980 on Build GRCh38
rs976078505 Chr.4: 105569592-105569592 on Build GRCh38
rs762603830 Chr.4: 105569779-105569779 on Build GRCh38
rs567774480 Chr.4: 105569824-105569824 on Build GRCh38

View Full Product Details