Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 88541873-88542407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00461
Entrez Gene ID
645323
Gene Name
long intergenic non-protein coding RNA 461
Gene Aliases
EyeLinc1, VISC, Visc-1a, Visc-1b, Visc-2
UniGene
Hs.12827
Gene Chromosome Location
Chr.5: 88540779-88684803 on Build GRCh38
Transcripts
NR_015436.1

SNP Detail

SNP ID Chromosome Location
rs73173597 Chr.5: 88542248-88542248 on Build GRCh38
rs189163702 Chr.5: 88542179-88542179 on Build GRCh38
rs34424549 Chr.5: 88542074-88542075 on Build GRCh38
rs913232553 Chr.5: 88541933-88541933 on Build GRCh38
rs930064459 Chr.5: 88542300-88542300 on Build GRCh38
rs374081470 Chr.5: 88542050-88542050 on Build GRCh38
rs376568744 Chr.5: 88541942-88541942 on Build GRCh38
rs1041100511 Chr.5: 88542140-88542140 on Build GRCh38
rs994162332 Chr.5: 88541898-88541898 on Build GRCh38
rs932144195 Chr.5: 88542061-88542061 on Build GRCh38
rs936929838 Chr.5: 88542101-88542101 on Build GRCh38
rs183908949 Chr.5: 88542342-88542342 on Build GRCh38
rs558067685 Chr.5: 88542285-88542285 on Build GRCh38
rs905591247 Chr.5: 88542198-88542198 on Build GRCh38
rs1034609889 Chr.5: 88541940-88541940 on Build GRCh38
rs901293841 Chr.5: 88542143-88542143 on Build GRCh38
rs1005362017 Chr.5: 88541935-88541936 on Build GRCh38
rs539431299 Chr.5: 88542268-88542268 on Build GRCh38
rs967145622 Chr.5: 88542238-88542238 on Build GRCh38
rs912010652 Chr.5: 88542287-88542287 on Build GRCh38
rs992581982 Chr.5: 88541970-88541970 on Build GRCh38
rs540109370 Chr.5: 88542334-88542334 on Build GRCh38
rs921977415 Chr.5: 88542267-88542267 on Build GRCh38
rs940946507 Chr.5: 88542144-88542144 on Build GRCh38
rs751211561 Chr.5: 88541908-88541909 on Build GRCh38
rs911391556 Chr.5: 88542122-88542122 on Build GRCh38
rs989627823 Chr.5: 88542104-88542104 on Build GRCh38
rs57977764 Chr.5: 88541941-88541942 on Build GRCh38
rs888385234 Chr.5: 88541926-88541926 on Build GRCh38
rs1048109614 Chr.5: 88542194-88542194 on Build GRCh38
rs977083468 Chr.5: 88542239-88542239 on Build GRCh38
rs958410683 Chr.5: 88541969-88541969 on Build GRCh38
rs1001322648 Chr.5: 88542230-88542230 on Build GRCh38
rs757001669 Chr.5: 88541993-88541993 on Build GRCh38
rs1034148157 Chr.5: 88542174-88542174 on Build GRCh38
rs1025547044 Chr.5: 88541926-88541927 on Build GRCh38
rs547234434 Chr.5: 88541988-88541988 on Build GRCh38
rs776623958 Chr.5: 88542273-88542273 on Build GRCh38
rs1052344037 Chr.5: 88542069-88542069 on Build GRCh38
rs901179023 Chr.5: 88541897-88541897 on Build GRCh38
rs193120388 Chr.5: 88542246-88542246 on Build GRCh38
rs756026842 Chr.5: 88541907-88541908 on Build GRCh38
rs148292196 Chr.5: 88542019-88542019 on Build GRCh38
rs958273590 Chr.5: 88542244-88542244 on Build GRCh38
rs534733589 Chr.5: 88542221-88542222 on Build GRCh38
rs888342310 Chr.5: 88542157-88542157 on Build GRCh38
rs747589607 Chr.5: 88542359-88542359 on Build GRCh38
rs1024101415 Chr.5: 88541974-88541974 on Build GRCh38
rs376633259 Chr.5: 88542229-88542230 on Build GRCh38
rs141401967 Chr.5: 88542325-88542325 on Build GRCh38
rs978774513 Chr.5: 88542265-88542265 on Build GRCh38
rs977522934 Chr.5: 88542032-88542032 on Build GRCh38
rs922021762 Chr.5: 88542022-88542022 on Build GRCh38
rs924201272 Chr.5: 88542090-88542090 on Build GRCh38
rs978804199 Chr.5: 88542010-88542010 on Build GRCh38
rs920864343 Chr.5: 88542374-88542374 on Build GRCh38
rs573490380 Chr.5: 88542366-88542366 on Build GRCh38
rs145305281 Chr.5: 88541906-88541907 on Build GRCh38
rs535964548 Chr.5: 88542243-88542243 on Build GRCh38
rs779108456 Chr.5: 88542231-88542231 on Build GRCh38
rs912099055 Chr.5: 88542094-88542094 on Build GRCh38
rs555857068 Chr.5: 88542036-88542036 on Build GRCh38
rs968675630 Chr.5: 88542251-88542251 on Build GRCh38
rs1047025281 Chr.5: 88542347-88542347 on Build GRCh38
rs942910280 Chr.5: 88542133-88542133 on Build GRCh38
rs1014132989 Chr.5: 88542220-88542220 on Build GRCh38
rs112127173 Chr.5: 88541942-88541942 on Build GRCh38
rs1047524464 Chr.5: 88542148-88542148 on Build GRCh38
rs182832847 Chr.5: 88542147-88542147 on Build GRCh38
rs896795091 Chr.5: 88542173-88542173 on Build GRCh38

View Full Product Details