Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 88664347-88664855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00461
Entrez Gene ID
645323
Gene Name
long intergenic non-protein coding RNA 461
Gene Aliases
EyeLinc1, VISC, Visc-1a, Visc-1b, Visc-2
UniGene
Hs.12827
Gene Chromosome Location
Chr.5: 88540779-88684803 on Build GRCh38
Transcripts
NR_015436.1 NR_024383.1 NR_024384.1

SNP Detail

SNP ID Chromosome Location
rs899142238 Chr.5: 88664770-88664770 on Build GRCh38
rs931799542 Chr.5: 88664646-88664646 on Build GRCh38
rs1036298802 Chr.5: 88664717-88664717 on Build GRCh38
rs1030499237 Chr.5: 88664431-88664431 on Build GRCh38
rs927831811 Chr.5: 88664651-88664651 on Build GRCh38
rs912862194 Chr.5: 88664392-88664392 on Build GRCh38
rs954963567 Chr.5: 88664432-88664432 on Build GRCh38
rs890707190 Chr.5: 88664772-88664772 on Build GRCh38
rs142047230 Chr.5: 88664667-88664667 on Build GRCh38
rs982112408 Chr.5: 88664596-88664596 on Build GRCh38
rs947092645 Chr.5: 88664430-88664430 on Build GRCh38
rs183652495 Chr.5: 88664648-88664648 on Build GRCh38
rs978904633 Chr.5: 88664440-88664440 on Build GRCh38
rs747099552 Chr.5: 88664390-88664390 on Build GRCh38
rs1023670008 Chr.5: 88664796-88664796 on Build GRCh38
rs761848899 Chr.5: 88664808-88664808 on Build GRCh38
rs538728263 Chr.5: 88664677-88664677 on Build GRCh38
rs1015087722 Chr.5: 88664459-88664459 on Build GRCh38
rs1024102050 Chr.5: 88664812-88664812 on Build GRCh38
rs997878676 Chr.5: 88664416-88664416 on Build GRCh38
rs1039836611 Chr.5: 88664727-88664727 on Build GRCh38
rs994877940 Chr.5: 88664790-88664790 on Build GRCh38
rs536389542 Chr.5: 88664504-88664504 on Build GRCh38
rs957137056 Chr.5: 88664377-88664377 on Build GRCh38
rs935275301 Chr.5: 88664652-88664652 on Build GRCh38
rs973925321 Chr.5: 88664481-88664481 on Build GRCh38
rs555045695 Chr.5: 88664561-88664561 on Build GRCh38
rs537463660 Chr.5: 88664771-88664771 on Build GRCh38
rs899581686 Chr.5: 88664744-88664744 on Build GRCh38
rs1005001531 Chr.5: 88664456-88664456 on Build GRCh38
rs933615743 Chr.5: 88664767-88664767 on Build GRCh38
rs1015035211 Chr.5: 88664779-88664779 on Build GRCh38
rs1005117928 Chr.5: 88664775-88664775 on Build GRCh38
rs963479114 Chr.5: 88664472-88664472 on Build GRCh38
rs946583323 Chr.5: 88664719-88664719 on Build GRCh38
rs969771798 Chr.5: 88664516-88664516 on Build GRCh38

View Full Product Details