Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 88672791-88673333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00461
Entrez Gene ID
645323
Gene Name
long intergenic non-protein coding RNA 461
Gene Aliases
EyeLinc1, VISC, Visc-1a, Visc-1b, Visc-2
UniGene
Hs.12827
Gene Chromosome Location
Chr.5: 88540779-88684803 on Build GRCh38
Transcripts
NR_015436.1 NR_024383.1 NR_024384.1

SNP Detail

SNP ID Chromosome Location
rs1027904301 Chr.5: 88673250-88673250 on Build GRCh38
rs1056717868 Chr.5: 88672831-88672831 on Build GRCh38
rs1000231915 Chr.5: 88673181-88673181 on Build GRCh38
rs564694309 Chr.5: 88673040-88673040 on Build GRCh38
rs548392617 Chr.5: 88672856-88672856 on Build GRCh38
rs763384563 Chr.5: 88673088-88673088 on Build GRCh38
rs981072738 Chr.5: 88672878-88672878 on Build GRCh38
rs1009186608 Chr.5: 88672984-88672984 on Build GRCh38
rs1029127348 Chr.5: 88672944-88672944 on Build GRCh38
rs1018757260 Chr.5: 88673274-88673274 on Build GRCh38
rs867934453 Chr.5: 88672818-88672818 on Build GRCh38
rs561428817 Chr.5: 88673225-88673225 on Build GRCh38
rs1043974175 Chr.5: 88673145-88673145 on Build GRCh38
rs1007876876 Chr.5: 88672869-88672869 on Build GRCh38
rs903656825 Chr.5: 88672819-88672819 on Build GRCh38
rs939460074 Chr.5: 88673036-88673036 on Build GRCh38
rs945631979 Chr.5: 88673143-88673143 on Build GRCh38
rs374243033 Chr.5: 88673285-88673285 on Build GRCh38
rs750914129 Chr.5: 88672993-88672993 on Build GRCh38
rs755817963 Chr.5: 88673165-88673165 on Build GRCh38
rs903815535 Chr.5: 88673095-88673102 on Build GRCh38
rs1050093620 Chr.5: 88673191-88673191 on Build GRCh38
rs553169964 Chr.5: 88673062-88673062 on Build GRCh38
rs920613325 Chr.5: 88673026-88673026 on Build GRCh38
rs1008634621 Chr.5: 88673261-88673261 on Build GRCh38
rs1015706899 Chr.5: 88672870-88672870 on Build GRCh38
rs570120096 Chr.5: 88672862-88672862 on Build GRCh38
rs892760163 Chr.5: 88672851-88672851 on Build GRCh38
rs112029600 Chr.5: 88672903-88672903 on Build GRCh38
rs961087478 Chr.5: 88672901-88672901 on Build GRCh38
rs878927581 Chr.5: 88673095-88673096 on Build GRCh38
rs996643897 Chr.5: 88672855-88672855 on Build GRCh38
rs1009812675 Chr.5: 88672852-88672852 on Build GRCh38
rs6882046 Chr.5: 88673046-88673046 on Build GRCh38
rs1056678562 Chr.5: 88673115-88673115 on Build GRCh38
rs3814424 Chr.5: 88673135-88673135 on Build GRCh38
rs920851912 Chr.5: 88672942-88672942 on Build GRCh38
rs952295117 Chr.5: 88673271-88673271 on Build GRCh38
rs926659757 Chr.5: 88673058-88673058 on Build GRCh38
rs952181485 Chr.5: 88672947-88672947 on Build GRCh38
rs1043590882 Chr.5: 88673095-88673101 on Build GRCh38
rs953243074 Chr.5: 88672971-88672971 on Build GRCh38
rs758467101 Chr.5: 88673095-88673095 on Build GRCh38
rs971936638 Chr.5: 88673023-88673023 on Build GRCh38
rs374453020 Chr.5: 88672927-88672927 on Build GRCh38
rs556670733 Chr.5: 88672863-88672863 on Build GRCh38
rs972287495 Chr.5: 88672937-88672937 on Build GRCh38
rs1018726168 Chr.5: 88672992-88672992 on Build GRCh38
rs535590202 Chr.5: 88673197-88673197 on Build GRCh38
rs552573182 Chr.5: 88673035-88673035 on Build GRCh38
rs995203931 Chr.5: 88673226-88673226 on Build GRCh38
rs1050048787 Chr.5: 88673129-88673129 on Build GRCh38

View Full Product Details