Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173020564-173020965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0E1
Entrez Gene ID
8992
Gene Name
ATPase H+ transporting V0 subunit e1
Gene Aliases
ATP6H, ATP6V0E, M9.2, Vma21, Vma21p
UniGene
Hs.484188
Gene Chromosome Location
Chr.5: 172983760-173034897 on Build GRCh38
Transcripts
NM_003945.3 NR_002988.1

Gene Symbol
SNORA74B
Entrez Gene ID
677841
Gene Name
small nucleolar RNA, H/ACA box 74B
Gene Aliases
U19-2
UniGene
Hs.692720
Gene Chromosome Location
Chr.5: 173020726-173020929 on Build GRCh38
Transcripts
NM_003945.3 NR_002988.1

SNP Detail

SNP ID Chromosome Location
rs761038210 Chr.5: 173020893-173020893 on Build GRCh38
rs74759042 Chr.5: 173020635-173020635 on Build GRCh38
rs767035981 Chr.5: 173020682-173020682 on Build GRCh38
rs17075508 Chr.5: 173020759-173020759 on Build GRCh38
rs372762917 Chr.5: 173020821-173020821 on Build GRCh38
rs369090145 Chr.5: 173020886-173020886 on Build GRCh38
rs758184319 Chr.5: 173020743-173020743 on Build GRCh38
rs549343891 Chr.5: 173020749-173020749 on Build GRCh38
rs764696391 Chr.5: 173020820-173020820 on Build GRCh38
rs934430642 Chr.5: 173020818-173020818 on Build GRCh38
rs990913004 Chr.5: 173020856-173020856 on Build GRCh38
rs779766546 Chr.5: 173020745-173020745 on Build GRCh38
rs911515684 Chr.5: 173020636-173020636 on Build GRCh38
rs749394818 Chr.5: 173020757-173020757 on Build GRCh38
rs774054553 Chr.5: 173020873-173020874 on Build GRCh38
rs775775261 Chr.5: 173020801-173020801 on Build GRCh38
rs185838457 Chr.5: 173020758-173020758 on Build GRCh38
rs756188549 Chr.5: 173020928-173020929 on Build GRCh38
rs772256697 Chr.5: 173020905-173020905 on Build GRCh38
rs763578412 Chr.5: 173020691-173020691 on Build GRCh38
rs761643289 Chr.5: 173020923-173020924 on Build GRCh38
rs750317274 Chr.5: 173020926-173020927 on Build GRCh38
rs767375160 Chr.5: 173020924-173020925 on Build GRCh38
rs941932740 Chr.5: 173020742-173020742 on Build GRCh38
rs375904134 Chr.5: 173020624-173020624 on Build GRCh38
rs778908608 Chr.5: 173020870-173020870 on Build GRCh38
rs766390354 Chr.5: 173020933-173020934 on Build GRCh38
rs891222648 Chr.5: 173020787-173020787 on Build GRCh38
rs762882276 Chr.5: 173020934-173020934 on Build GRCh38
rs763796980 Chr.5: 173020867-173020867 on Build GRCh38
rs907710119 Chr.5: 173020901-173020901 on Build GRCh38
rs746371416 Chr.5: 173020873-173020873 on Build GRCh38
rs983186022 Chr.5: 173020872-173020872 on Build GRCh38
rs376016727 Chr.5: 173020836-173020836 on Build GRCh38
rs1016795113 Chr.5: 173020883-173020883 on Build GRCh38
rs189365736 Chr.5: 173020642-173020642 on Build GRCh38
rs752363279 Chr.5: 173020684-173020684 on Build GRCh38
rs763736200 Chr.5: 173020809-173020809 on Build GRCh38
rs560387869 Chr.5: 173020727-173020727 on Build GRCh38
rs756778919 Chr.5: 173020819-173020819 on Build GRCh38
rs749971740 Chr.5: 173020920-173020920 on Build GRCh38
rs778165480 Chr.5: 173020837-173020837 on Build GRCh38
rs757116279 Chr.5: 173020714-173020714 on Build GRCh38
rs768245246 Chr.5: 173020903-173020903 on Build GRCh38
rs898492929 Chr.5: 173020752-173020753 on Build GRCh38
rs764794790 Chr.5: 173020917-173020917 on Build GRCh38
rs758329349 Chr.5: 173020825-173020825 on Build GRCh38
rs767900917 Chr.5: 173020799-173020799 on Build GRCh38
rs771101216 Chr.5: 173020859-173020859 on Build GRCh38
rs753408749 Chr.5: 173020705-173020705 on Build GRCh38
rs200759323 Chr.5: 173020748-173020748 on Build GRCh38
rs762569921 Chr.5: 173020762-173020762 on Build GRCh38
rs924084577 Chr.5: 173020817-173020817 on Build GRCh38
rs779898834 Chr.5: 173020829-173020829 on Build GRCh38
rs773359318 Chr.5: 173020753-173020753 on Build GRCh38
rs796568343 Chr.5: 173020628-173020628 on Build GRCh38
rs772517659 Chr.5: 173020876-173020876 on Build GRCh38
rs181186513 Chr.5: 173020690-173020690 on Build GRCh38
rs371698779 Chr.5: 173020734-173020734 on Build GRCh38
rs751405612 Chr.5: 173020832-173020832 on Build GRCh38
rs773133360 Chr.5: 173020620-173020620 on Build GRCh38
rs755812076 Chr.5: 173020688-173020688 on Build GRCh38
rs905783426 Chr.5: 173020869-173020869 on Build GRCh38
rs367721000 Chr.5: 173020785-173020785 on Build GRCh38
rs200037818 Chr.5: 173020790-173020790 on Build GRCh38
rs776446593 Chr.5: 173020810-173020810 on Build GRCh38

View Full Product Details