Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 22146547-22147044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC15
Entrez Gene ID
401237
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
UniGene
Hs.712707
Gene Chromosome Location
Chr.6: 21666444-22194400 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

Gene Symbol
NBAT1
Entrez Gene ID
729177
Gene Name
neuroblastoma associated transcript 1
Gene Aliases
CASC14, NBAT-1
UniGene
Hs.730673
Gene Chromosome Location
Chr.6: 22134602-22147193 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

SNP Detail

SNP ID Chromosome Location
rs997461816 Chr.6: 22146926-22146926 on Build GRCh38
rs887774309 Chr.6: 22146912-22146912 on Build GRCh38
rs1057235974 Chr.6: 22146775-22146775 on Build GRCh38
rs75959788 Chr.6: 22146745-22146745 on Build GRCh38
rs966410449 Chr.6: 22146783-22146783 on Build GRCh38
rs756433912 Chr.6: 22146801-22146801 on Build GRCh38
rs70996637 Chr.6: 22146590-22146591 on Build GRCh38
rs929340104 Chr.6: 22146874-22146874 on Build GRCh38
rs959035849 Chr.6: 22146796-22146796 on Build GRCh38
rs369437432 Chr.6: 22146569-22146569 on Build GRCh38
rs869266216 Chr.6: 22146601-22146601 on Build GRCh38
rs5874858 Chr.6: 22146590-22146590 on Build GRCh38
rs775501641 Chr.6: 22146685-22146685 on Build GRCh38
rs1053464166 Chr.6: 22146842-22146842 on Build GRCh38
rs926413966 Chr.6: 22146974-22146974 on Build GRCh38
rs950690295 Chr.6: 22146935-22146935 on Build GRCh38
rs1008005643 Chr.6: 22146913-22146913 on Build GRCh38
rs145098007 Chr.6: 22146590-22146590 on Build GRCh38
rs545256535 Chr.6: 22146774-22146774 on Build GRCh38
rs773371245 Chr.6: 22146620-22146620 on Build GRCh38
rs919306827 Chr.6: 22146933-22146933 on Build GRCh38
rs367784883 Chr.6: 22146617-22146617 on Build GRCh38
rs966525854 Chr.6: 22146937-22146937 on Build GRCh38
rs750654141 Chr.6: 22146782-22146782 on Build GRCh38
rs1017977326 Chr.6: 22146750-22146750 on Build GRCh38
rs950234792 Chr.6: 22146826-22146826 on Build GRCh38
rs55997687 Chr.6: 22146587-22146587 on Build GRCh38
rs540665082 Chr.6: 22146721-22146721 on Build GRCh38
rs529457154 Chr.6: 22146987-22146987 on Build GRCh38
rs945030348 Chr.6: 22147014-22147014 on Build GRCh38
rs562156523 Chr.6: 22146963-22146963 on Build GRCh38
rs959026429 Chr.6: 22146960-22146960 on Build GRCh38
rs1018162347 Chr.6: 22146911-22146911 on Build GRCh38
rs991754707 Chr.6: 22146811-22146811 on Build GRCh38
rs537274293 Chr.6: 22146660-22146660 on Build GRCh38
rs552885353 Chr.6: 22146739-22146739 on Build GRCh38
rs765296589 Chr.6: 22146666-22146666 on Build GRCh38
rs753994564 Chr.6: 22147019-22147019 on Build GRCh38
rs756476503 Chr.6: 22146786-22146786 on Build GRCh38
rs544905898 Chr.6: 22146573-22146573 on Build GRCh38
rs767690349 Chr.6: 22146778-22146783 on Build GRCh38
rs766333591 Chr.6: 22146991-22146991 on Build GRCh38
rs973443449 Chr.6: 22146622-22146622 on Build GRCh38
rs563599632 Chr.6: 22146823-22146823 on Build GRCh38
rs751690384 Chr.6: 22146569-22146570 on Build GRCh38
rs965898771 Chr.6: 22146755-22146755 on Build GRCh38
rs1021178855 Chr.6: 22146927-22146927 on Build GRCh38
rs374098798 Chr.6: 22146587-22146589 on Build GRCh38
rs919435889 Chr.6: 22146873-22146873 on Build GRCh38
rs1001161609 Chr.6: 22146841-22146841 on Build GRCh38
rs901780919 Chr.6: 22146919-22146919 on Build GRCh38
rs56389486 Chr.6: 22146589-22146589 on Build GRCh38
rs764066431 Chr.6: 22146695-22146695 on Build GRCh38
rs35735817 Chr.6: 22146824-22146825 on Build GRCh38
rs766745529 Chr.6: 22146845-22146845 on Build GRCh38
rs55720019 Chr.6: 22146713-22146713 on Build GRCh38
rs1038937405 Chr.6: 22146917-22146917 on Build GRCh38
rs905125769 Chr.6: 22146828-22146828 on Build GRCh38
rs963943442 Chr.6: 22146569-22146569 on Build GRCh38
rs892165271 Chr.6: 22147002-22147002 on Build GRCh38
rs555027820 Chr.6: 22146668-22146668 on Build GRCh38
rs530705087 Chr.6: 22146902-22146902 on Build GRCh38
rs184457870 Chr.6: 22146949-22146949 on Build GRCh38
rs1046293223 Chr.6: 22146857-22146857 on Build GRCh38
rs757392363 Chr.6: 22146754-22146754 on Build GRCh38
rs749702885 Chr.6: 22147002-22147002 on Build GRCh38
rs750808990 Chr.6: 22146787-22146787 on Build GRCh38
rs763106405 Chr.6: 22146694-22146694 on Build GRCh38
rs1049175708 Chr.6: 22146910-22146910 on Build GRCh38
rs980641338 Chr.6: 22146965-22146965 on Build GRCh38
rs149970179 Chr.6: 22146588-22146588 on Build GRCh38
rs540995324 Chr.6: 22146571-22146572 on Build GRCh38
rs917791112 Chr.6: 22146780-22146780 on Build GRCh38
rs997787773 Chr.6: 22146781-22146781 on Build GRCh38
rs1032093123 Chr.6: 22146954-22146954 on Build GRCh38
rs781583164 Chr.6: 22146570-22146571 on Build GRCh38
rs573441503 Chr.6: 22146692-22146692 on Build GRCh38

View Full Product Details