Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 22572938-22573467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506178
Entrez Gene ID
100506178
Gene Name
uncharacterized LOC100506178
Gene Aliases
-
UniGene
Hs.668122
Gene Chromosome Location
Chr.7: 22563337-22573998 on Build GRCh38
Transcripts
NR_038393.1

SNP Detail

SNP ID Chromosome Location
rs74932277 Chr.7: 22572987-22572987 on Build GRCh38
rs149726744 Chr.7: 22573352-22573352 on Build GRCh38
rs1050542388 Chr.7: 22573428-22573428 on Build GRCh38
rs1009992232 Chr.7: 22573325-22573325 on Build GRCh38
rs896541018 Chr.7: 22573317-22573317 on Build GRCh38
rs373468808 Chr.7: 22573264-22573264 on Build GRCh38
rs979314540 Chr.7: 22573145-22573145 on Build GRCh38
rs971941086 Chr.7: 22573232-22573232 on Build GRCh38
rs970695122 Chr.7: 22573284-22573284 on Build GRCh38
rs913821901 Chr.7: 22573118-22573118 on Build GRCh38
rs923019410 Chr.7: 22573381-22573381 on Build GRCh38
rs113614314 Chr.7: 22573013-22573013 on Build GRCh38
rs928976275 Chr.7: 22573196-22573196 on Build GRCh38
rs945554464 Chr.7: 22573356-22573356 on Build GRCh38
rs926718952 Chr.7: 22573014-22573014 on Build GRCh38
rs1041239633 Chr.7: 22573376-22573376 on Build GRCh38
rs1041354850 Chr.7: 22573087-22573087 on Build GRCh38
rs116229638 Chr.7: 22573147-22573147 on Build GRCh38
rs943192623 Chr.7: 22573156-22573156 on Build GRCh38
rs901450671 Chr.7: 22573398-22573398 on Build GRCh38
rs528932193 Chr.7: 22573200-22573200 on Build GRCh38
rs964532435 Chr.7: 22573370-22573370 on Build GRCh38
rs1002325686 Chr.7: 22573086-22573086 on Build GRCh38
rs11514749 Chr.7: 22573342-22573342 on Build GRCh38
rs923019173 Chr.7: 22573184-22573184 on Build GRCh38
rs1035099884 Chr.7: 22573100-22573100 on Build GRCh38
rs76804061 Chr.7: 22573004-22573004 on Build GRCh38
rs887358405 Chr.7: 22573229-22573229 on Build GRCh38
rs975827773 Chr.7: 22573160-22573160 on Build GRCh38
rs773577194 Chr.7: 22573201-22573201 on Build GRCh38
rs953198381 Chr.7: 22572980-22572980 on Build GRCh38
rs182168349 Chr.7: 22573353-22573353 on Build GRCh38
rs905391764 Chr.7: 22573276-22573276 on Build GRCh38
rs1016533719 Chr.7: 22573195-22573196 on Build GRCh38
rs967959075 Chr.7: 22573139-22573139 on Build GRCh38
rs560887712 Chr.7: 22573331-22573331 on Build GRCh38
rs1002356916 Chr.7: 22573303-22573303 on Build GRCh38
rs916731 Chr.7: 22573217-22573217 on Build GRCh38
rs774426449 Chr.7: 22573031-22573031 on Build GRCh38
rs551682830 Chr.7: 22573267-22573267 on Build GRCh38
rs539839927 Chr.7: 22573053-22573053 on Build GRCh38
rs1548415 Chr.7: 22572998-22572998 on Build GRCh38
rs936827540 Chr.7: 22573017-22573017 on Build GRCh38
rs115935938 Chr.7: 22572972-22572972 on Build GRCh38
rs867066408 Chr.7: 22573406-22573406 on Build GRCh38
rs527966304 Chr.7: 22573412-22573412 on Build GRCh38
rs762204466 Chr.7: 22573188-22573188 on Build GRCh38
rs1040091235 Chr.7: 22572992-22572992 on Build GRCh38
rs568875582 Chr.7: 22573235-22573235 on Build GRCh38
rs555252276 Chr.7: 22572993-22572993 on Build GRCh38
rs1017076021 Chr.7: 22573030-22573030 on Build GRCh38
rs111671105 Chr.7: 22573429-22573429 on Build GRCh38
rs190990264 Chr.7: 22573309-22573309 on Build GRCh38
rs552430392 Chr.7: 22573248-22573248 on Build GRCh38
rs748474336 Chr.7: 22573107-22573107 on Build GRCh38
rs945673788 Chr.7: 22573057-22573057 on Build GRCh38
rs975909651 Chr.7: 22573374-22573374 on Build GRCh38
rs936777227 Chr.7: 22573338-22573338 on Build GRCh38
rs959678305 Chr.7: 22573333-22573333 on Build GRCh38
rs139562749 Chr.7: 22573003-22573003 on Build GRCh38
rs888875205 Chr.7: 22573138-22573138 on Build GRCh38
rs988136262 Chr.7: 22573108-22573108 on Build GRCh38
rs968012946 Chr.7: 22573349-22573349 on Build GRCh38

View Full Product Details